Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 103298029-103298539 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FZD6
Entrez Gene ID
8323
Gene Name
frizzled class receptor 6
Gene Aliases
FZ-6, FZ6, HFZ6, NDNC10
UniGene
Hs.591863
Gene Chromosome Location
Chr.8: 103298433-103332866 on Build GRCh38
Transcripts
NM_001164615.1

SNP Detail

SNP ID Chromosome Location
rs766661509 Chr.8: 103298343-103298343 on Build GRCh38
rs909393573 Chr.8: 103298506-103298506 on Build GRCh38
rs1009597281 Chr.8: 103298515-103298515 on Build GRCh38
rs1000674146 Chr.8: 103298359-103298359 on Build GRCh38
rs536524164 Chr.8: 103298493-103298493 on Build GRCh38
rs764033958 Chr.8: 103298246-103298246 on Build GRCh38
rs940895733 Chr.8: 103298516-103298516 on Build GRCh38
rs908944270 Chr.8: 103298215-103298215 on Build GRCh38
rs182026182 Chr.8: 103298314-103298314 on Build GRCh38
rs564153240 Chr.8: 103298356-103298356 on Build GRCh38
rs147010363 Chr.8: 103298371-103298371 on Build GRCh38
rs971370331 Chr.8: 103298414-103298414 on Build GRCh38
rs190632224 Chr.8: 103298504-103298504 on Build GRCh38
rs981880858 Chr.8: 103298421-103298421 on Build GRCh38
rs148270136 Chr.8: 103298295-103298295 on Build GRCh38
rs535824163 Chr.8: 103298225-103298228 on Build GRCh38
rs971424077 Chr.8: 103298074-103298074 on Build GRCh38
rs548634924 Chr.8: 103298128-103298128 on Build GRCh38
rs941087119 Chr.8: 103298367-103298367 on Build GRCh38
rs889060001 Chr.8: 103298329-103298329 on Build GRCh38
rs1020298233 Chr.8: 103298062-103298062 on Build GRCh38
rs550113463 Chr.8: 103298460-103298460 on Build GRCh38
rs892689704 Chr.8: 103298514-103298515 on Build GRCh38
rs185780520 Chr.8: 103298412-103298412 on Build GRCh38
rs367593363 Chr.8: 103298219-103298219 on Build GRCh38
rs530382431 Chr.8: 103298079-103298079 on Build GRCh38
rs1010319914 Chr.8: 103298410-103298410 on Build GRCh38
rs3758096 Chr.8: 103298265-103298265 on Build GRCh38
rs927561374 Chr.8: 103298126-103298126 on Build GRCh38
rs827524 Chr.8: 103298160-103298160 on Build GRCh38
rs929634238 Chr.8: 103298301-103298301 on Build GRCh38
rs113841273 Chr.8: 103298456-103298456 on Build GRCh38
rs1034579792 Chr.8: 103298451-103298451 on Build GRCh38
rs982842626 Chr.8: 103298324-103298324 on Build GRCh38
rs934246757 Chr.8: 103298427-103298427 on Build GRCh38
rs968523345 Chr.8: 103298173-103298173 on Build GRCh38
rs981717106 Chr.8: 103298112-103298112 on Build GRCh38
rs893184737 Chr.8: 103298406-103298406 on Build GRCh38
rs901508222 Chr.8: 103298268-103298268 on Build GRCh38
rs1010087301 Chr.8: 103298137-103298137 on Build GRCh38
rs141302959 Chr.8: 103298299-103298299 on Build GRCh38
rs909667193 Chr.8: 103298331-103298331 on Build GRCh38
rs940447586 Chr.8: 103298237-103298237 on Build GRCh38
rs1051332148 Chr.8: 103298475-103298475 on Build GRCh38
rs78475112 Chr.8: 103298187-103298187 on Build GRCh38
rs892440432 Chr.8: 103298107-103298107 on Build GRCh38
rs568394626 Chr.8: 103298478-103298478 on Build GRCh38

View Full Product Details