Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 143214921-143215415 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GPIHBP1
Entrez Gene ID
338328
Gene Name
glycosylphosphatidylinositol anchored high density lipoprotein binding protein 1
Gene Aliases
GPI-HBP1, HYPL1D
UniGene
Hs.426410
Gene Chromosome Location
Chr.8: 143213193-143217170 on Build GRCh38
Transcripts
NM_001301772.1 NM_178172.5

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3897358 c.258C>T p.G86G Chr.8: 143215089-143215089 on Build GRCh38
6451834 c.360G>A p.T120T Chr.8: 143215323-143215323 on Build GRCh38
3897357 c.194G>A p.C65Y Chr.8: 143215025-143215025 on Build GRCh38
6180244 c.293C>A p.T98N Chr.8: 143215124-143215124 on Build GRCh38
6180243 c.434G>T p.R145L Chr.8: 143215397-143215397 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1008833185 Chr.8: 143215289-143215289 on Build GRCh38
rs147455726 Chr.8: 143215126-143215126 on Build GRCh38
rs776509101 Chr.8: 143215002-143215002 on Build GRCh38
rs764329208 Chr.8: 143215311-143215311 on Build GRCh38
rs201591245 Chr.8: 143215248-143215248 on Build GRCh38
rs1019659098 Chr.8: 143215316-143215316 on Build GRCh38
rs756751413 Chr.8: 143215129-143215129 on Build GRCh38
rs531971333 Chr.8: 143215165-143215165 on Build GRCh38
rs545434518 Chr.8: 143215089-143215089 on Build GRCh38
rs368250860 Chr.8: 143214982-143214982 on Build GRCh38
rs781028625 Chr.8: 143215149-143215149 on Build GRCh38
rs150327266 Chr.8: 143214958-143214958 on Build GRCh38
rs746715981 Chr.8: 143215381-143215381 on Build GRCh38
rs201558206 Chr.8: 143215154-143215154 on Build GRCh38
rs777392774 Chr.8: 143215356-143215356 on Build GRCh38
rs756671297 Chr.8: 143215057-143215057 on Build GRCh38
rs370523141 Chr.8: 143215220-143215220 on Build GRCh38
rs201685731 Chr.8: 143215331-143215331 on Build GRCh38
rs987327940 Chr.8: 143214995-143214995 on Build GRCh38
rs1040945521 Chr.8: 143214970-143214970 on Build GRCh38
rs751011642 Chr.8: 143215056-143215056 on Build GRCh38
rs587777643 Chr.8: 143215283-143215283 on Build GRCh38
rs141874363 Chr.8: 143215178-143215178 on Build GRCh38
rs766823940 Chr.8: 143215235-143215235 on Build GRCh38
rs548047112 Chr.8: 143215194-143215194 on Build GRCh38
rs78123380 Chr.8: 143215145-143215145 on Build GRCh38
rs587777638 Chr.8: 143215025-143215025 on Build GRCh38
rs567736240 Chr.8: 143215014-143215014 on Build GRCh38
rs538358896 Chr.8: 143215020-143215020 on Build GRCh38
rs770702398 Chr.8: 143215392-143215392 on Build GRCh38
rs759437603 Chr.8: 143215008-143215008 on Build GRCh38
rs1014854435 Chr.8: 143215034-143215034 on Build GRCh38
rs56046179 Chr.8: 143215153-143215153 on Build GRCh38
rs1044562943 Chr.8: 143215260-143215260 on Build GRCh38
rs149305582 Chr.8: 143215058-143215058 on Build GRCh38
rs587777641 Chr.8: 143215294-143215294 on Build GRCh38
rs1033049419 Chr.8: 143215083-143215083 on Build GRCh38
rs778853798 Chr.8: 143215344-143215344 on Build GRCh38
rs375091775 Chr.8: 143215167-143215167 on Build GRCh38
rs536044071 Chr.8: 143214959-143214959 on Build GRCh38
rs142861814 Chr.8: 143215285-143215285 on Build GRCh38
rs754340438 Chr.8: 143215211-143215211 on Build GRCh38
rs761064228 Chr.8: 143215178-143215178 on Build GRCh38
rs547403983 Chr.8: 143215168-143215168 on Build GRCh38
rs755487011 Chr.8: 143215215-143215215 on Build GRCh38
rs377573186 Chr.8: 143215137-143215137 on Build GRCh38
rs144587519 Chr.8: 143215071-143215071 on Build GRCh38
rs763489745 Chr.8: 143215176-143215176 on Build GRCh38
rs762969345 Chr.8: 143215295-143215295 on Build GRCh38
rs375004793 Chr.8: 143215054-143215054 on Build GRCh38
rs369934389 Chr.8: 143214996-143214996 on Build GRCh38
rs369222108 Chr.8: 143215342-143215342 on Build GRCh38
rs199743701 Chr.8: 143215238-143215238 on Build GRCh38
rs766013621 Chr.8: 143214994-143214994 on Build GRCh38
rs759883512 Chr.8: 143215396-143215396 on Build GRCh38
rs371690913 Chr.8: 143215051-143215051 on Build GRCh38
rs779017546 Chr.8: 143215143-143215143 on Build GRCh38
rs777910164 Chr.8: 143215225-143215225 on Build GRCh38
rs565995398 Chr.8: 143215221-143215221 on Build GRCh38
rs587777642 Chr.8: 143215380-143215396 on Build GRCh38
rs78632677 Chr.8: 143215209-143215209 on Build GRCh38
rs745795326 Chr.8: 143215000-143215000 on Build GRCh38
rs781490909 Chr.8: 143214983-143214983 on Build GRCh38
rs976611935 Chr.8: 143215205-143215205 on Build GRCh38
rs868788719 Chr.8: 143215353-143215353 on Build GRCh38
rs201256271 Chr.8: 143215250-143215250 on Build GRCh38
rs138215873 Chr.8: 143215116-143215116 on Build GRCh38
rs930554936 Chr.8: 143215187-143215187 on Build GRCh38
rs376082089 Chr.8: 143215151-143215151 on Build GRCh38
rs751821739 Chr.8: 143215312-143215312 on Build GRCh38
rs187913047 Chr.8: 143214941-143214941 on Build GRCh38
rs948589033 Chr.8: 143215217-143215217 on Build GRCh38
rs768095273 Chr.8: 143215159-143215159 on Build GRCh38
rs531136098 Chr.8: 143215255-143215255 on Build GRCh38
rs745681552 Chr.8: 143215233-143215233 on Build GRCh38
rs751963282 Chr.8: 143215050-143215050 on Build GRCh38
rs562028937 Chr.8: 143215135-143215135 on Build GRCh38
rs78367243 Chr.8: 143215394-143215394 on Build GRCh38
rs776388699 Chr.8: 143215098-143215098 on Build GRCh38
rs530025316 Chr.8: 143215192-143215192 on Build GRCh38
rs536657163 Chr.8: 143215242-143215242 on Build GRCh38
rs780340378 Chr.8: 143215061-143215061 on Build GRCh38
rs542034857 Chr.8: 143215103-143215103 on Build GRCh38
rs747635399 Chr.8: 143215345-143215345 on Build GRCh38
rs771469195 Chr.8: 143215270-143215270 on Build GRCh38
rs1019027365 Chr.8: 143215186-143215186 on Build GRCh38
rs912544443 Chr.8: 143214996-143214996 on Build GRCh38
rs375588884 Chr.8: 143215240-143215240 on Build GRCh38
rs587777637 Chr.8: 143215307-143215307 on Build GRCh38
rs587777639 Chr.8: 143215033-143215033 on Build GRCh38
rs1010225016 Chr.8: 143215380-143215380 on Build GRCh38
rs761154174 Chr.8: 143215265-143215265 on Build GRCh38
rs565814494 Chr.8: 143215185-143215185 on Build GRCh38
rs768749238 Chr.8: 143214998-143214998 on Build GRCh38
rs749374488 Chr.8: 143215070-143215070 on Build GRCh38
rs752728823 Chr.8: 143215286-143215286 on Build GRCh38
rs866602255 Chr.8: 143215379-143215379 on Build GRCh38
rs773126617 Chr.8: 143214999-143214999 on Build GRCh38
rs769712017 Chr.8: 143215109-143215109 on Build GRCh38
rs999099101 Chr.8: 143214971-143214971 on Build GRCh38
rs199622782 Chr.8: 143215136-143215136 on Build GRCh38
rs142959160 Chr.8: 143215125-143215125 on Build GRCh38
rs773838142 Chr.8: 143215118-143215118 on Build GRCh38
rs774982418 Chr.8: 143215113-143215113 on Build GRCh38
rs200649178 Chr.8: 143215219-143215219 on Build GRCh38
rs748863132 Chr.8: 143215157-143215157 on Build GRCh38
rs536960226 Chr.8: 143215347-143215347 on Build GRCh38
rs777039010 Chr.8: 143215282-143215282 on Build GRCh38
rs370422727 Chr.8: 143215166-143215166 on Build GRCh38
rs973149211 Chr.8: 143215177-143215177 on Build GRCh38
rs192966895 Chr.8: 143214988-143214988 on Build GRCh38
rs371650837 Chr.8: 143215322-143215322 on Build GRCh38
rs373101330 Chr.8: 143215117-143215117 on Build GRCh38
rs578073937 Chr.8: 143215022-143215022 on Build GRCh38
rs371050017 Chr.8: 143215144-143215144 on Build GRCh38
rs200196582 Chr.8: 143215267-143215267 on Build GRCh38
rs556559035 Chr.8: 143215021-143215021 on Build GRCh38
rs375903045 Chr.8: 143215251-143215251 on Build GRCh38
rs377237094 Chr.8: 143214989-143214989 on Build GRCh38
rs200709949 Chr.8: 143215268-143215268 on Build GRCh38
rs778945628 Chr.8: 143215076-143215076 on Build GRCh38
rs746851401 Chr.8: 143215082-143215082 on Build GRCh38
rs587777640 Chr.8: 143215097-143215097 on Build GRCh38
rs779774750 Chr.8: 143215323-143215323 on Build GRCh38
rs987970176 Chr.8: 143214944-143214944 on Build GRCh38

View Full Product Details