Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 22447056-22447552 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DMRTA1
Entrez Gene ID
63951
Gene Name
DMRT like family A1
Gene Aliases
DMO, DMRT4
UniGene
Hs.371976
Gene Chromosome Location
Chr.9: 22446841-22452473 on Build GRCh38
Transcripts
NM_022160.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4678152 c.117delG p.M41fs*44 Chr.9: 22447182-22447182 on Build GRCh38
7115235 c.116C>T p.S39L Chr.9: 22447181-22447181 on Build GRCh38
4678151 c.64delG p.L23fs*1 Chr.9: 22447129-22447129 on Build GRCh38
4620873 c.150G>A p.P50P Chr.9: 22447215-22447215 on Build GRCh38
5876077 c.32G>A p.R11Q Chr.9: 22447097-22447097 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs367897183 Chr.9: 22447099-22447099 on Build GRCh38
rs773714453 Chr.9: 22447155-22447155 on Build GRCh38
rs770303277 Chr.9: 22447436-22447436 on Build GRCh38
rs772704733 Chr.9: 22447154-22447154 on Build GRCh38
rs531593966 Chr.9: 22447505-22447505 on Build GRCh38
rs866228763 Chr.9: 22447333-22447333 on Build GRCh38
rs544960245 Chr.9: 22447289-22447289 on Build GRCh38
rs777563284 Chr.9: 22447178-22447178 on Build GRCh38
rs747359394 Chr.9: 22447254-22447255 on Build GRCh38
rs766471458 Chr.9: 22447159-22447159 on Build GRCh38
rs3207249 Chr.9: 22447183-22447183 on Build GRCh38
rs777562758 Chr.9: 22447112-22447112 on Build GRCh38
rs375102960 Chr.9: 22447167-22447167 on Build GRCh38
rs535115502 Chr.9: 22447199-22447199 on Build GRCh38
rs759526828 Chr.9: 22447227-22447227 on Build GRCh38
rs759047301 Chr.9: 22447475-22447475 on Build GRCh38
rs943476278 Chr.9: 22447329-22447329 on Build GRCh38
rs755146229 Chr.9: 22447080-22447080 on Build GRCh38
rs757218772 Chr.9: 22447182-22447182 on Build GRCh38
rs754075701 Chr.9: 22447079-22447079 on Build GRCh38
rs778251211 Chr.9: 22447255-22447260 on Build GRCh38
rs182619176 Chr.9: 22447429-22447429 on Build GRCh38
rs982473444 Chr.9: 22447328-22447328 on Build GRCh38
rs759797418 Chr.9: 22447165-22447165 on Build GRCh38
rs746326120 Chr.9: 22447326-22447326 on Build GRCh38
rs779073439 Chr.9: 22447246-22447248 on Build GRCh38
rs144513092 Chr.9: 22447123-22447123 on Build GRCh38
rs764010857 Chr.9: 22447253-22447253 on Build GRCh38
rs749690923 Chr.9: 22447525-22447525 on Build GRCh38
rs371967563 Chr.9: 22447124-22447124 on Build GRCh38
rs372741047 Chr.9: 22447215-22447215 on Build GRCh38
rs765840976 Chr.9: 22447375-22447375 on Build GRCh38
rs748905882 Chr.9: 22447203-22447203 on Build GRCh38
rs961368316 Chr.9: 22447278-22447278 on Build GRCh38
rs773022500 Chr.9: 22447463-22447463 on Build GRCh38
rs370894033 Chr.9: 22447241-22447241 on Build GRCh38
rs752365919 Chr.9: 22447478-22447478 on Build GRCh38
rs752600308 Chr.9: 22447169-22447169 on Build GRCh38
rs747356238 Chr.9: 22447315-22447315 on Build GRCh38
rs549110334 Chr.9: 22447468-22447468 on Build GRCh38
rs760179259 Chr.9: 22447392-22447392 on Build GRCh38
rs767354118 Chr.9: 22447357-22447357 on Build GRCh38
rs767550081 Chr.9: 22447288-22447288 on Build GRCh38
rs754621439 Chr.9: 22447309-22447309 on Build GRCh38
rs771491779 Chr.9: 22447320-22447320 on Build GRCh38
rs765290110 Chr.9: 22447232-22447232 on Build GRCh38
rs759531750 Chr.9: 22447464-22447464 on Build GRCh38
rs1021322554 Chr.9: 22447337-22447338 on Build GRCh38
rs961304940 Chr.9: 22447107-22447107 on Build GRCh38
rs374668199 Chr.9: 22447479-22447479 on Build GRCh38
rs111465355 Chr.9: 22447337-22447337 on Build GRCh38
rs750776463 Chr.9: 22447495-22447495 on Build GRCh38
rs755110527 Chr.9: 22447209-22447242 on Build GRCh38
rs771400248 Chr.9: 22447220-22447220 on Build GRCh38
rs1054207836 Chr.9: 22447186-22447186 on Build GRCh38
rs1037059968 Chr.9: 22447454-22447454 on Build GRCh38
rs777123187 Chr.9: 22447322-22447322 on Build GRCh38
rs750343241 Chr.9: 22447360-22447360 on Build GRCh38
rs752276044 Chr.9: 22447414-22447414 on Build GRCh38
rs749373349 Chr.9: 22447446-22447446 on Build GRCh38
rs34336274 Chr.9: 22447524-22447524 on Build GRCh38
rs779568105 Chr.9: 22447202-22447202 on Build GRCh38
rs1029541487 Chr.9: 22447299-22447299 on Build GRCh38
rs753360659 Chr.9: 22447389-22447389 on Build GRCh38
rs575127664 Chr.9: 22447212-22447212 on Build GRCh38
rs748918900 Chr.9: 22447302-22447302 on Build GRCh38
rs757627293 Chr.9: 22447425-22447425 on Build GRCh38
rs1053839191 Chr.9: 22447193-22447193 on Build GRCh38
rs552671652 Chr.9: 22447150-22447150 on Build GRCh38
rs746390595 Chr.9: 22447435-22447435 on Build GRCh38
rs774344629 Chr.9: 22447353-22447353 on Build GRCh38
rs775585334 Chr.9: 22447237-22447237 on Build GRCh38
rs961025232 Chr.9: 22447276-22447276 on Build GRCh38
rs756031690 Chr.9: 22447366-22447366 on Build GRCh38
rs1042656565 Chr.9: 22447238-22447238 on Build GRCh38
rs769782737 Chr.9: 22447332-22447332 on Build GRCh38
rs748435300 Chr.9: 22447245-22447246 on Build GRCh38
rs369146134 Chr.9: 22447453-22447453 on Build GRCh38
rs945369353 Chr.9: 22447378-22447380 on Build GRCh38
rs915211979 Chr.9: 22447391-22447391 on Build GRCh38
rs771288515 Chr.9: 22447490-22447491 on Build GRCh38
rs780798416 Chr.9: 22447131-22447131 on Build GRCh38
rs763122411 Chr.9: 22447346-22447346 on Build GRCh38
rs751368754 Chr.9: 22447260-22447260 on Build GRCh38
rs952454027 Chr.9: 22447311-22447311 on Build GRCh38
rs1015142186 Chr.9: 22447272-22447272 on Build GRCh38
rs779559391 Chr.9: 22447301-22447301 on Build GRCh38
rs758130740 Chr.9: 22447173-22447173 on Build GRCh38
rs555297669 Chr.9: 22447210-22447210 on Build GRCh38
rs993830673 Chr.9: 22447280-22447280 on Build GRCh38
rs890603910 Chr.9: 22447257-22447257 on Build GRCh38
rs201950652 Chr.9: 22447184-22447184 on Build GRCh38
rs201444816 Chr.9: 22447095-22447095 on Build GRCh38
rs756637667 Chr.9: 22447499-22447499 on Build GRCh38
rs755586405 Chr.9: 22447292-22447292 on Build GRCh38
rs564754414 Chr.9: 22447344-22447344 on Build GRCh38
rs776744543 Chr.9: 22447226-22447226 on Build GRCh38
rs529345917 Chr.9: 22447455-22447455 on Build GRCh38
rs896875470 Chr.9: 22447488-22447488 on Build GRCh38
rs916420544 Chr.9: 22447319-22447319 on Build GRCh38
rs745546150 Chr.9: 22447135-22447135 on Build GRCh38
rs570229256 Chr.9: 22447528-22447528 on Build GRCh38
rs754640625 Chr.9: 22447407-22447407 on Build GRCh38
rs769299301 Chr.9: 22447138-22447138 on Build GRCh38
rs935354573 Chr.9: 22447367-22447367 on Build GRCh38
rs753878586 Chr.9: 22447164-22447164 on Build GRCh38
rs578231901 Chr.9: 22447398-22447398 on Build GRCh38
rs752407569 Chr.9: 22447101-22447101 on Build GRCh38
rs751563180 Chr.9: 22447180-22447180 on Build GRCh38
rs201252188 Chr.9: 22447111-22447111 on Build GRCh38
rs985274648 Chr.9: 22447172-22447172 on Build GRCh38
rs748414619 Chr.9: 22447532-22447532 on Build GRCh38
rs868676600 Chr.9: 22447196-22447196 on Build GRCh38
rs753272129 Chr.9: 22447477-22447477 on Build GRCh38
rs774829612 Chr.9: 22447145-22447145 on Build GRCh38
rs1003716955 Chr.9: 22447251-22447251 on Build GRCh38
rs764389162 Chr.9: 22447350-22447350 on Build GRCh38
rs1050428620 Chr.9: 22447400-22447400 on Build GRCh38
rs865992039 Chr.9: 22447361-22447361 on Build GRCh38
rs772016586 Chr.9: 22447458-22447458 on Build GRCh38
rs775598674 Chr.9: 22447340-22447340 on Build GRCh38
rs779087748 Chr.9: 22447530-22447530 on Build GRCh38
rs761115757 Chr.9: 22447158-22447158 on Build GRCh38
rs778155730 Chr.9: 22447314-22447314 on Build GRCh38
rs770857162 Chr.9: 22447263-22447263 on Build GRCh38
rs1017034609 Chr.9: 22447269-22447269 on Build GRCh38
rs763183946 Chr.9: 22447242-22447242 on Build GRCh38
rs761697575 Chr.9: 22447354-22447354 on Build GRCh38
rs560582163 Chr.9: 22447445-22447445 on Build GRCh38

View Full Product Details