Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 35661844-35662355 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ARHGEF39
Entrez Gene ID
84904
Gene Name
Rho guanine nucleotide exchange factor 39
Gene Aliases
C9orf100
UniGene
Hs.534579
Gene Chromosome Location
Chr.9: 35658875-35665281 on Build GRCh38
Transcripts
NM_032818.2 XM_011518057.2 XM_017015225.1 XM_017015226.1 XM_017015227.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1108686 c.545C>T p.P182L Chr.9: 35662267-35662267 on Build GRCh38
1701069 c.946C>A p.Q316K Chr.9: 35662225-35662225 on Build GRCh38
1199061 c.548T>A p.V183D Chr.9: 35662264-35662264 on Build GRCh38
4513625 c.945C>T p.D315D Chr.9: 35662226-35662226 on Build GRCh38
3907189 c.949G>C p.E317Q Chr.9: 35662222-35662222 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs372474273 Chr.9: 35662290-35662290 on Build GRCh38
rs773937794 Chr.9: 35662258-35662258 on Build GRCh38
rs918137615 Chr.9: 35662305-35662305 on Build GRCh38
rs752538916 Chr.9: 35661993-35661993 on Build GRCh38
rs201388388 Chr.9: 35662202-35662202 on Build GRCh38
rs145102414 Chr.9: 35662210-35662210 on Build GRCh38
rs202026693 Chr.9: 35662021-35662021 on Build GRCh38
rs769112868 Chr.9: 35662232-35662232 on Build GRCh38
rs571432251 Chr.9: 35661944-35661945 on Build GRCh38
rs72727021 Chr.9: 35661946-35661946 on Build GRCh38
rs563309170 Chr.9: 35662123-35662123 on Build GRCh38
rs749338640 Chr.9: 35662052-35662052 on Build GRCh38
rs900326609 Chr.9: 35662157-35662157 on Build GRCh38
rs958410728 Chr.9: 35661877-35661877 on Build GRCh38
rs962658159 Chr.9: 35662099-35662099 on Build GRCh38
rs754565633 Chr.9: 35662269-35662270 on Build GRCh38
rs759040206 Chr.9: 35662196-35662196 on Build GRCh38
rs368516952 Chr.9: 35662169-35662169 on Build GRCh38
rs755282083 Chr.9: 35662310-35662310 on Build GRCh38
rs543653898 Chr.9: 35662209-35662209 on Build GRCh38
rs757218113 Chr.9: 35662212-35662212 on Build GRCh38
rs375320699 Chr.9: 35662273-35662273 on Build GRCh38
rs753354206 Chr.9: 35662046-35662046 on Build GRCh38
rs188416193 Chr.9: 35662160-35662160 on Build GRCh38
rs750723950 Chr.9: 35662202-35662203 on Build GRCh38
rs767209570 Chr.9: 35662201-35662201 on Build GRCh38
rs761893192 Chr.9: 35662159-35662167 on Build GRCh38
rs371981293 Chr.9: 35662207-35662207 on Build GRCh38
rs778898090 Chr.9: 35662215-35662215 on Build GRCh38
rs183746919 Chr.9: 35662147-35662147 on Build GRCh38
rs777072431 Chr.9: 35662176-35662177 on Build GRCh38
rs549593972 Chr.9: 35662020-35662020 on Build GRCh38
rs542783733 Chr.9: 35662137-35662137 on Build GRCh38
rs982687996 Chr.9: 35662070-35662070 on Build GRCh38
rs757130272 Chr.9: 35662283-35662283 on Build GRCh38
rs146744438 Chr.9: 35662217-35662217 on Build GRCh38
rs747201515 Chr.9: 35662294-35662294 on Build GRCh38
rs761157168 Chr.9: 35662165-35662165 on Build GRCh38
rs2297879 Chr.9: 35662254-35662254 on Build GRCh38
rs192748579 Chr.9: 35662164-35662164 on Build GRCh38
rs749939324 Chr.9: 35661939-35661939 on Build GRCh38
rs902251022 Chr.9: 35662122-35662122 on Build GRCh38
rs770426400 Chr.9: 35662256-35662256 on Build GRCh38
rs369229378 Chr.9: 35661897-35661897 on Build GRCh38
rs768660374 Chr.9: 35662017-35662020 on Build GRCh38
rs570847750 Chr.9: 35662185-35662185 on Build GRCh38
rs761321558 Chr.9: 35662248-35662249 on Build GRCh38
rs775265694 Chr.9: 35662274-35662274 on Build GRCh38
rs986824429 Chr.9: 35661878-35661878 on Build GRCh38
rs920113208 Chr.9: 35661978-35661978 on Build GRCh38
rs991254555 Chr.9: 35661909-35661909 on Build GRCh38
rs373020136 Chr.9: 35662096-35662096 on Build GRCh38
rs777152998 Chr.9: 35662242-35662242 on Build GRCh38
rs763662178 Chr.9: 35662035-35662036 on Build GRCh38
rs1000462230 Chr.9: 35662211-35662211 on Build GRCh38
rs747304228 Chr.9: 35662226-35662226 on Build GRCh38
rs780388609 Chr.9: 35662225-35662225 on Build GRCh38
rs760271171 Chr.9: 35662275-35662275 on Build GRCh38
rs767004564 Chr.9: 35662250-35662251 on Build GRCh38
rs765219350 Chr.9: 35662285-35662285 on Build GRCh38
rs750463750 Chr.9: 35662289-35662289 on Build GRCh38
rs764672860 Chr.9: 35661937-35661937 on Build GRCh38
rs565886755 Chr.9: 35662276-35662276 on Build GRCh38
rs369314807 Chr.9: 35662288-35662288 on Build GRCh38
rs376843558 Chr.9: 35661984-35661984 on Build GRCh38
rs374982403 Chr.9: 35662162-35662162 on Build GRCh38
rs971248927 Chr.9: 35662037-35662037 on Build GRCh38
rs762618751 Chr.9: 35661940-35661940 on Build GRCh38
rs781634794 Chr.9: 35662312-35662312 on Build GRCh38
rs372984038 Chr.9: 35662183-35662183 on Build GRCh38
rs755995169 Chr.9: 35662029-35662029 on Build GRCh38
rs780116952 Chr.9: 35662291-35662291 on Build GRCh38
rs765935230 Chr.9: 35662186-35662186 on Build GRCh38
rs191189324 Chr.9: 35662128-35662128 on Build GRCh38
rs754678407 Chr.9: 35661976-35661976 on Build GRCh38
rs373614212 Chr.9: 35662319-35662319 on Build GRCh38
rs370610213 Chr.9: 35662030-35662030 on Build GRCh38
rs374038098 Chr.9: 35662138-35662138 on Build GRCh38
rs754330389 Chr.9: 35662251-35662252 on Build GRCh38
rs762587408 Chr.9: 35661887-35661887 on Build GRCh38
rs962916833 Chr.9: 35662327-35662327 on Build GRCh38
rs751301439 Chr.9: 35661971-35661971 on Build GRCh38
rs774026852 Chr.9: 35662193-35662193 on Build GRCh38
rs755980580 Chr.9: 35662205-35662205 on Build GRCh38
rs973776195 Chr.9: 35662105-35662105 on Build GRCh38

View Full Product Details