Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 41343873-41344376 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DDX3X
Entrez Gene ID
1654
Gene Name
DEAD-box helicase 3, X-linked
Gene Aliases
CAP-Rf, DBX, DDX14, DDX3, HLP2, MRX102
UniGene
Hs.728563
Gene Chromosome Location
Chr.X: 41333308-41364472 on Build GRCh38
Transcripts
NM_001193416.2 NM_001193417.2 NM_001356.4 NR_126093.1 XM_011543892.2 XM_017029313.1 XM_017029314.1

SNP Detail

SNP ID Chromosome Location
rs1042619114 Chr.X: 41343935-41343935 on Build GRCh38
rs769385948 Chr.X: 41344014-41344018 on Build GRCh38
rs768636608 Chr.X: 41344235-41344235 on Build GRCh38
rs752332279 Chr.X: 41344103-41344103 on Build GRCh38
rs770973350 Chr.X: 41344179-41344179 on Build GRCh38
rs778328870 Chr.X: 41344331-41344331 on Build GRCh38
rs780646477 Chr.X: 41343989-41343990 on Build GRCh38
rs1046285813 Chr.X: 41344229-41344229 on Build GRCh38
rs1021585207 Chr.X: 41344051-41344051 on Build GRCh38
rs777089000 Chr.X: 41344164-41344164 on Build GRCh38
rs763958253 Chr.X: 41344043-41344043 on Build GRCh38
rs762591825 Chr.X: 41344223-41344223 on Build GRCh38
rs370318891 Chr.X: 41344216-41344216 on Build GRCh38
rs758861097 Chr.X: 41344185-41344185 on Build GRCh38
rs760617639 Chr.X: 41344037-41344037 on Build GRCh38
rs753360586 Chr.X: 41344062-41344062 on Build GRCh38
rs754843761 Chr.X: 41343938-41343939 on Build GRCh38
rs761300398 Chr.X: 41344067-41344067 on Build GRCh38
rs752063399 Chr.X: 41343905-41343907 on Build GRCh38
rs368366114 Chr.X: 41344343-41344343 on Build GRCh38
rs749374119 Chr.X: 41343985-41343985 on Build GRCh38
rs986153207 Chr.X: 41344183-41344183 on Build GRCh38
rs869320681 Chr.X: 41344247-41344247 on Build GRCh38
rs771151855 Chr.X: 41343991-41343991 on Build GRCh38
rs759391220 Chr.X: 41343994-41343994 on Build GRCh38
rs910542744 Chr.X: 41344184-41344184 on Build GRCh38
rs747348474 Chr.X: 41344209-41344209 on Build GRCh38
rs767359806 Chr.X: 41343996-41343996 on Build GRCh38
rs753107889 Chr.X: 41344150-41344150 on Build GRCh38
rs746016142 Chr.X: 41344203-41344203 on Build GRCh38
rs756500897 Chr.X: 41344327-41344327 on Build GRCh38
rs776493154 Chr.X: 41344244-41344244 on Build GRCh38
rs875989802 Chr.X: 41344278-41344278 on Build GRCh38
rs769044936 Chr.X: 41344322-41344322 on Build GRCh38
rs774779977 Chr.X: 41343992-41343992 on Build GRCh38
rs12011185 Chr.X: 41343946-41343946 on Build GRCh38
rs111399541 Chr.X: 41344129-41344129 on Build GRCh38
rs754240311 Chr.X: 41344335-41344335 on Build GRCh38
rs75228950 Chr.X: 41343974-41343974 on Build GRCh38
rs758891758 Chr.X: 41344165-41344165 on Build GRCh38
rs796052225 Chr.X: 41344318-41344318 on Build GRCh38
rs757557752 Chr.X: 41343922-41343922 on Build GRCh38
rs956044475 Chr.X: 41344009-41344011 on Build GRCh38
rs758050366 Chr.X: 41344128-41344128 on Build GRCh38
rs374274692 Chr.X: 41344268-41344268 on Build GRCh38
rs867967504 Chr.X: 41344052-41344052 on Build GRCh38
rs762545072 Chr.X: 41344311-41344311 on Build GRCh38
rs748834275 Chr.X: 41344167-41344167 on Build GRCh38
rs749967337 Chr.X: 41344307-41344307 on Build GRCh38
rs745409903 Chr.X: 41343991-41343994 on Build GRCh38
rs775426247 Chr.X: 41344001-41344001 on Build GRCh38
rs751362480 Chr.X: 41343930-41343930 on Build GRCh38
rs775218288 Chr.X: 41344222-41344223 on Build GRCh38
rs374463509 Chr.X: 41344340-41344340 on Build GRCh38
rs368761223 Chr.X: 41344286-41344286 on Build GRCh38
rs772451101 Chr.X: 41344206-41344206 on Build GRCh38
rs188024690 Chr.X: 41344201-41344201 on Build GRCh38
rs797045025 Chr.X: 41344351-41344351 on Build GRCh38
rs753083462 Chr.X: 41344155-41344155 on Build GRCh38
rs766426082 Chr.X: 41343904-41343904 on Build GRCh38
rs755636737 Chr.X: 41344113-41344113 on Build GRCh38

View Full Product Details