Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 108164799-108165308 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
COL4A6
Entrez Gene ID
1288
Gene Name
collagen type IV alpha 6 chain
Gene Aliases
CXDELq22.3, DELXq22.3, DFNX6
UniGene
Hs.145586
Gene Chromosome Location
Chr.X: 108155607-108439497 on Build GRCh38
Transcripts
NM_001287758.1 NM_001287759.1 NM_001287760.1 NM_001847.3 NM_033641.3 XM_006724617.3 XM_011530852.2 XM_011530853.2

SNP Detail

SNP ID Chromosome Location
rs759096492 Chr.X: 108165081-108165081 on Build GRCh38
rs368443205 Chr.X: 108164982-108164982 on Build GRCh38
rs760642708 Chr.X: 108165070-108165070 on Build GRCh38
rs924409073 Chr.X: 108165110-108165110 on Build GRCh38
rs947130162 Chr.X: 108165094-108165094 on Build GRCh38
rs1044393095 Chr.X: 108165214-108165214 on Build GRCh38
rs961524119 Chr.X: 108165242-108165242 on Build GRCh38
rs764946097 Chr.X: 108165025-108165025 on Build GRCh38
rs771079240 Chr.X: 108164849-108164849 on Build GRCh38
rs774910875 Chr.X: 108165161-108165161 on Build GRCh38
rs896111053 Chr.X: 108164898-108164898 on Build GRCh38
rs766448373 Chr.X: 108165078-108165078 on Build GRCh38
rs201920507 Chr.X: 108164976-108164976 on Build GRCh38
rs758764486 Chr.X: 108164962-108164962 on Build GRCh38
rs188298998 Chr.X: 108165238-108165238 on Build GRCh38
rs764779930 Chr.X: 108165083-108165083 on Build GRCh38
rs371882231 Chr.X: 108164855-108164855 on Build GRCh38
rs748653548 Chr.X: 108164826-108164826 on Build GRCh38
rs762477564 Chr.X: 108165239-108165239 on Build GRCh38
rs768951810 Chr.X: 108165106-108165106 on Build GRCh38
rs767347024 Chr.X: 108165008-108165008 on Build GRCh38
rs766126341 Chr.X: 108164923-108164923 on Build GRCh38
rs759314157 Chr.X: 108164829-108164829 on Build GRCh38
rs771942739 Chr.X: 108165063-108165063 on Build GRCh38
rs779096584 Chr.X: 108164959-108164959 on Build GRCh38
rs771062602 Chr.X: 108164977-108164977 on Build GRCh38
rs781637030 Chr.X: 108165200-108165200 on Build GRCh38
rs756910677 Chr.X: 108165035-108165035 on Build GRCh38
rs765172549 Chr.X: 108165068-108165068 on Build GRCh38
rs760233418 Chr.X: 108165011-108165011 on Build GRCh38
rs201356626 Chr.X: 108164858-108164858 on Build GRCh38
rs375109347 Chr.X: 108164864-108164864 on Build GRCh38
rs375561639 Chr.X: 108164983-108164983 on Build GRCh38
rs971170375 Chr.X: 108165150-108165150 on Build GRCh38
rs145634519 Chr.X: 108164905-108164905 on Build GRCh38
rs761686276 Chr.X: 108164993-108164993 on Build GRCh38
rs934823710 Chr.X: 108165145-108165145 on Build GRCh38
rs756095492 Chr.X: 108164873-108164873 on Build GRCh38
rs747890668 Chr.X: 108165053-108165053 on Build GRCh38
rs368126530 Chr.X: 108164835-108164835 on Build GRCh38
rs753586099 Chr.X: 108164928-108164928 on Build GRCh38
rs751394543 Chr.X: 108164865-108164865 on Build GRCh38
rs374158244 Chr.X: 108164834-108164834 on Build GRCh38
rs1022981436 Chr.X: 108164930-108164930 on Build GRCh38
rs778764545 Chr.X: 108165044-108165044 on Build GRCh38
rs766036752 Chr.X: 108165015-108165015 on Build GRCh38
rs182434004 Chr.X: 108165234-108165234 on Build GRCh38
rs34132805 Chr.X: 108164992-108164992 on Build GRCh38
rs752461306 Chr.X: 108165084-108165084 on Build GRCh38
rs138267158 Chr.X: 108164961-108164961 on Build GRCh38
rs1024978295 Chr.X: 108165146-108165146 on Build GRCh38
rs763195874 Chr.X: 108165085-108165085 on Build GRCh38
rs368876473 Chr.X: 108165009-108165009 on Build GRCh38
rs376219034 Chr.X: 108165064-108165064 on Build GRCh38
rs966910222 Chr.X: 108164832-108164832 on Build GRCh38
rs1048778473 Chr.X: 108165002-108165002 on Build GRCh38
rs373308389 Chr.X: 108164981-108164981 on Build GRCh38
rs749296554 Chr.X: 108164901-108164901 on Build GRCh38
rs770726601 Chr.X: 108164872-108164872 on Build GRCh38
rs1012628704 Chr.X: 108165091-108165091 on Build GRCh38
rs757302174 Chr.X: 108165036-108165036 on Build GRCh38
rs1043185258 Chr.X: 108165108-108165108 on Build GRCh38
rs1021123578 Chr.X: 108164831-108164831 on Build GRCh38
rs923188604 Chr.X: 108165031-108165031 on Build GRCh38
rs559631296 Chr.X: 108165270-108165270 on Build GRCh38
rs192754010 Chr.X: 108165052-108165052 on Build GRCh38
rs755374057 Chr.X: 108164934-108164934 on Build GRCh38
rs752945513 Chr.X: 108165028-108165028 on Build GRCh38
rs113329155 Chr.X: 108165024-108165024 on Build GRCh38
rs896038821 Chr.X: 108165176-108165176 on Build GRCh38
rs755743892 Chr.X: 108164921-108164921 on Build GRCh38
rs778357288 Chr.X: 108164967-108164967 on Build GRCh38
rs776211548 Chr.X: 108165079-108165079 on Build GRCh38
rs1038702288 Chr.X: 108165016-108165016 on Build GRCh38
rs758851710 Chr.X: 108165034-108165034 on Build GRCh38
rs767454751 Chr.X: 108164850-108164850 on Build GRCh38
rs1039232364 Chr.X: 108165277-108165277 on Build GRCh38

View Full Product Details