Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 202962565-202963067 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CYB5R1
Entrez Gene ID
51706
Gene Name
cytochrome b5 reductase 1
Gene Aliases
B5R.1, B5R1, B5R2, NQO3A2, humb5R2
UniGene
Hs.334832
Gene Chromosome Location
Chr.1: 202961873-202967276 on Build GRCh38
Transcripts
NM_016243.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6124174 c.851T>A p.V284E Chr.1: 202962594-202962594 on Build GRCh38
6124173 c.801C>T p.P267P Chr.1: 202962644-202962644 on Build GRCh38
4844218 c.746-1G>C p.? Chr.1: 202962700-202962700 on Build GRCh38
2213271 c.778G>A p.D260N Chr.1: 202962667-202962667 on Build GRCh38
6900780 c.803C>T p.A268V Chr.1: 202962642-202962642 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs147500937 Chr.1: 202962955-202962955 on Build GRCh38
rs760465028 Chr.1: 202962763-202962763 on Build GRCh38
rs768907769 Chr.1: 202962640-202962640 on Build GRCh38
rs775569567 Chr.1: 202962613-202962614 on Build GRCh38
rs757165233 Chr.1: 202962917-202962917 on Build GRCh38
rs778167846 Chr.1: 202963027-202963027 on Build GRCh38
rs139281039 Chr.1: 202962611-202962611 on Build GRCh38
rs916861412 Chr.1: 202962775-202962775 on Build GRCh38
rs768714535 Chr.1: 202962693-202962693 on Build GRCh38
rs140109989 Chr.1: 202962985-202962985 on Build GRCh38
rs2232852 Chr.1: 202962870-202962870 on Build GRCh38
rs747308777 Chr.1: 202963033-202963033 on Build GRCh38
rs781467504 Chr.1: 202962643-202962643 on Build GRCh38
rs2232855 Chr.1: 202962706-202962706 on Build GRCh38
rs776768068 Chr.1: 202962811-202962811 on Build GRCh38
rs761628880 Chr.1: 202962997-202962997 on Build GRCh38
rs763702340 Chr.1: 202962995-202962995 on Build GRCh38
rs201672974 Chr.1: 202962705-202962705 on Build GRCh38
rs142160530 Chr.1: 202962610-202962610 on Build GRCh38
rs776228063 Chr.1: 202962720-202962720 on Build GRCh38
rs370774212 Chr.1: 202962644-202962644 on Build GRCh38
rs762569843 Chr.1: 202962900-202962900 on Build GRCh38
rs200152783 Chr.1: 202962598-202962598 on Build GRCh38
rs774504672 Chr.1: 202962752-202962752 on Build GRCh38
rs558186954 Chr.1: 202962615-202962615 on Build GRCh38
rs906320379 Chr.1: 202962609-202962609 on Build GRCh38
rs2232851 Chr.1: 202963026-202963026 on Build GRCh38
rs777279598 Chr.1: 202962976-202962976 on Build GRCh38
rs1018832464 Chr.1: 202962942-202962942 on Build GRCh38
rs749419789 Chr.1: 202962689-202962689 on Build GRCh38
rs780075925 Chr.1: 202962683-202962683 on Build GRCh38
rs550808555 Chr.1: 202962927-202962927 on Build GRCh38
rs2232856 Chr.1: 202962625-202962625 on Build GRCh38
rs1029225839 Chr.1: 202962907-202962907 on Build GRCh38
rs543752468 Chr.1: 202963005-202963005 on Build GRCh38
rs748397908 Chr.1: 202962702-202962702 on Build GRCh38
rs769875330 Chr.1: 202962631-202962631 on Build GRCh38
rs753561339 Chr.1: 202962595-202962595 on Build GRCh38
rs978056408 Chr.1: 202962969-202962969 on Build GRCh38
rs761262526 Chr.1: 202962709-202962709 on Build GRCh38
rs767377771 Chr.1: 202962998-202962998 on Build GRCh38
rs752391510 Chr.1: 202962667-202962667 on Build GRCh38
rs967232813 Chr.1: 202962755-202962755 on Build GRCh38
rs747396746 Chr.1: 202962966-202962966 on Build GRCh38
rs749505973 Chr.1: 202962634-202962634 on Build GRCh38
rs764960509 Chr.1: 202962658-202962658 on Build GRCh38
rs762634579 Chr.1: 202962986-202962986 on Build GRCh38
rs113277602 Chr.1: 202962787-202962787 on Build GRCh38
rs771520187 Chr.1: 202962974-202962974 on Build GRCh38
rs962903352 Chr.1: 202962696-202962696 on Build GRCh38
rs765978281 Chr.1: 202962651-202962651 on Build GRCh38
rs764726910 Chr.1: 202962884-202962884 on Build GRCh38
rs774083961 Chr.1: 202962996-202962996 on Build GRCh38
rs1004465991 Chr.1: 202962837-202962837 on Build GRCh38
rs953395552 Chr.1: 202962674-202962674 on Build GRCh38
rs755090666 Chr.1: 202962933-202962933 on Build GRCh38
rs777511762 Chr.1: 202962668-202962668 on Build GRCh38
rs750376582 Chr.1: 202963002-202963002 on Build GRCh38
rs766067115 Chr.1: 202962585-202962585 on Build GRCh38
rs770988478 Chr.1: 202962773-202962773 on Build GRCh38
rs752406347 Chr.1: 202962888-202962888 on Build GRCh38
rs199641680 Chr.1: 202962656-202962656 on Build GRCh38
rs745867958 Chr.1: 202962626-202962626 on Build GRCh38
rs773777113 Chr.1: 202962650-202962650 on Build GRCh38
rs774558313 Chr.1: 202962641-202962641 on Build GRCh38
rs371377002 Chr.1: 202962680-202962680 on Build GRCh38
rs752252253 Chr.1: 202962952-202962952 on Build GRCh38
rs751395799 Chr.1: 202962912-202962912 on Build GRCh38
rs573326198 Chr.1: 202962953-202962953 on Build GRCh38
rs936518828 Chr.1: 202962831-202962831 on Build GRCh38
rs568446633 Chr.1: 202962867-202962867 on Build GRCh38
rs528172097 Chr.1: 202962698-202962698 on Build GRCh38
rs753876399 Chr.1: 202962930-202962930 on Build GRCh38
rs760706628 Chr.1: 202962920-202962922 on Build GRCh38
rs537378591 Chr.1: 202962737-202962737 on Build GRCh38
rs770198268 Chr.1: 202962697-202962697 on Build GRCh38
rs186129618 Chr.1: 202962908-202962908 on Build GRCh38
rs978191075 Chr.1: 202962782-202962782 on Build GRCh38
rs751483688 Chr.1: 202962671-202962671 on Build GRCh38
rs768573443 Chr.1: 202962738-202962738 on Build GRCh38
rs1051400584 Chr.1: 202962962-202962962 on Build GRCh38
rs375092634 Chr.1: 202963025-202963025 on Build GRCh38
rs906205809 Chr.1: 202962833-202962833 on Build GRCh38
rs554829576 Chr.1: 202962984-202962984 on Build GRCh38
rs2232854 Chr.1: 202962711-202962711 on Build GRCh38
rs765359335 Chr.1: 202963012-202963012 on Build GRCh38
rs994700134 Chr.1: 202962871-202962871 on Build GRCh38
rs778538611 Chr.1: 202962608-202962608 on Build GRCh38
rs767465631 Chr.1: 202962922-202962922 on Build GRCh38
rs2232853 Chr.1: 202962830-202962830 on Build GRCh38
rs200700150 Chr.1: 202962652-202962652 on Build GRCh38

View Full Product Details