Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 205062304-205062810 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CNTN2
Entrez Gene ID
6900
Gene Name
contactin 2
Gene Aliases
AXT, FAME5, TAG-1, TAX, TAX1
UniGene
Hs.519220
Gene Chromosome Location
Chr.1: 205043160-205078043 on Build GRCh38
Transcripts
NM_005076.3 XM_017002198.1 XM_017002199.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
902706 c.1171G>A p.G391S Chr.1: 205062500-205062500 on Build GRCh38
6124046 c.1180C>T p.Q394* Chr.1: 205062509-205062509 on Build GRCh38
7138983 c.1165G>A p.D389N Chr.1: 205062494-205062494 on Build GRCh38
902705 c.1170G>A p.S390S Chr.1: 205062499-205062499 on Build GRCh38
902707 c.1215C>T p.Y405Y Chr.1: 205062544-205062544 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs370954288 Chr.1: 205062448-205062448 on Build GRCh38
rs563230395 Chr.1: 205062635-205062635 on Build GRCh38
rs765988318 Chr.1: 205062522-205062522 on Build GRCh38
rs771512123 Chr.1: 205062584-205062584 on Build GRCh38
rs558496903 Chr.1: 205062567-205062567 on Build GRCh38
rs1038459326 Chr.1: 205062628-205062628 on Build GRCh38
rs956480070 Chr.1: 205062774-205062774 on Build GRCh38
rs963748900 Chr.1: 205062734-205062734 on Build GRCh38
rs764907069 Chr.1: 205062544-205062544 on Build GRCh38
rs369658946 Chr.1: 205062701-205062701 on Build GRCh38
rs758300776 Chr.1: 205062454-205062454 on Build GRCh38
rs946086711 Chr.1: 205062354-205062354 on Build GRCh38
rs748305651 Chr.1: 205062562-205062562 on Build GRCh38
rs774607784 Chr.1: 205062479-205062479 on Build GRCh38
rs559161918 Chr.1: 205062660-205062660 on Build GRCh38
rs115882464 Chr.1: 205062498-205062498 on Build GRCh38
rs768968401 Chr.1: 205062393-205062393 on Build GRCh38
rs994385604 Chr.1: 205062375-205062375 on Build GRCh38
rs367637735 Chr.1: 205062582-205062582 on Build GRCh38
rs747527753 Chr.1: 205062578-205062578 on Build GRCh38
rs753497215 Chr.1: 205062433-205062433 on Build GRCh38
rs925052873 Chr.1: 205062763-205062763 on Build GRCh38
rs113709347 Chr.1: 205062570-205062570 on Build GRCh38
rs779243225 Chr.1: 205062561-205062561 on Build GRCh38
rs910917774 Chr.1: 205062415-205062415 on Build GRCh38
rs752262787 Chr.1: 205062545-205062545 on Build GRCh38
rs528366084 Chr.1: 205062761-205062761 on Build GRCh38
rs558556897 Chr.1: 205062470-205062470 on Build GRCh38
rs749341276 Chr.1: 205062555-205062555 on Build GRCh38
rs370344142 Chr.1: 205062502-205062502 on Build GRCh38
rs749478140 Chr.1: 205062471-205062471 on Build GRCh38
rs773641285 Chr.1: 205062577-205062577 on Build GRCh38
rs962876679 Chr.1: 205062367-205062367 on Build GRCh38
rs559405248 Chr.1: 205062664-205062664 on Build GRCh38
rs574939411 Chr.1: 205062590-205062590 on Build GRCh38
rs375976844 Chr.1: 205062397-205062397 on Build GRCh38
rs377487667 Chr.1: 205062476-205062476 on Build GRCh38
rs2242000 Chr.1: 205062641-205062641 on Build GRCh38
rs765974056 Chr.1: 205062429-205062429 on Build GRCh38
rs760589422 Chr.1: 205062694-205062694 on Build GRCh38
rs766319971 Chr.1: 205062786-205062786 on Build GRCh38
rs759129965 Chr.1: 205062532-205062532 on Build GRCh38
rs550015659 Chr.1: 205062394-205062394 on Build GRCh38
rs548368851 Chr.1: 205062352-205062352 on Build GRCh38
rs760311129 Chr.1: 205062419-205062419 on Build GRCh38
rs763866888 Chr.1: 205062547-205062547 on Build GRCh38
rs987923005 Chr.1: 205062783-205062783 on Build GRCh38
rs762201248 Chr.1: 205062598-205062598 on Build GRCh38
rs1008513392 Chr.1: 205062648-205062648 on Build GRCh38
rs536703590 Chr.1: 205062370-205062370 on Build GRCh38
rs761256115 Chr.1: 205062521-205062521 on Build GRCh38
rs894960127 Chr.1: 205062770-205062770 on Build GRCh38
rs376996860 Chr.1: 205062435-205062435 on Build GRCh38
rs372784277 Chr.1: 205062632-205062632 on Build GRCh38
rs375277010 Chr.1: 205062463-205062463 on Build GRCh38
rs538644037 Chr.1: 205062443-205062443 on Build GRCh38
rs200630793 Chr.1: 205062587-205062587 on Build GRCh38
rs2242001 Chr.1: 205062616-205062616 on Build GRCh38
rs368954831 Chr.1: 205062400-205062400 on Build GRCh38
rs372410577 Chr.1: 205062412-205062412 on Build GRCh38
rs568181579 Chr.1: 205062360-205062360 on Build GRCh38
rs754588590 Chr.1: 205062434-205062434 on Build GRCh38
rs774139246 Chr.1: 205062550-205062550 on Build GRCh38
rs372644641 Chr.1: 205062593-205062593 on Build GRCh38
rs772630695 Chr.1: 205062563-205062563 on Build GRCh38
rs545797566 Chr.1: 205062642-205062642 on Build GRCh38
rs955594502 Chr.1: 205062643-205062643 on Build GRCh38
rs1054922120 Chr.1: 205062466-205062466 on Build GRCh38
rs763225347 Chr.1: 205062389-205062389 on Build GRCh38
rs769920262 Chr.1: 205062464-205062464 on Build GRCh38
rs949334449 Chr.1: 205062573-205062573 on Build GRCh38
rs374199117 Chr.1: 205062553-205062553 on Build GRCh38
rs183284269 Chr.1: 205062558-205062558 on Build GRCh38
rs201580593 Chr.1: 205062526-205062526 on Build GRCh38

View Full Product Details