Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 206961133-206961635 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FCAMR
Entrez Gene ID
83953
Gene Name
Fc fragment of IgA and IgM receptor
Gene Aliases
CD351, FCA/MR, FKSG87
UniGene
Hs.145519
Gene Chromosome Location
Chr.1: 206957955-206970657 on Build GRCh38
Transcripts
NM_001122979.2 NM_001170631.1 NM_032029.4 XM_011510051.2 XM_011510052.2 XM_017002442.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4505078 c.683C>T p.P228L Chr.1: 206961193-206961193 on Build GRCh38
3803701 c.633G>C p.M211I Chr.1: 206961171-206961171 on Build GRCh38
3803700 c.705G>C p.M235I Chr.1: 206961171-206961171 on Build GRCh38
5426395 c.581-2A>G p.? Chr.1: 206961225-206961225 on Build GRCh38
7316728 c.627C>T p.L209L Chr.1: 206961177-206961177 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs959789209 Chr.1: 206961335-206961335 on Build GRCh38
rs751883557 Chr.1: 206961204-206961204 on Build GRCh38
rs188504174 Chr.1: 206961331-206961331 on Build GRCh38
rs970275554 Chr.1: 206961250-206961250 on Build GRCh38
rs1011563000 Chr.1: 206961536-206961536 on Build GRCh38
rs1022570023 Chr.1: 206961563-206961563 on Build GRCh38
rs1040935445 Chr.1: 206961515-206961515 on Build GRCh38
rs985283990 Chr.1: 206961443-206961443 on Build GRCh38
rs973163836 Chr.1: 206961370-206961370 on Build GRCh38
rs574707132 Chr.1: 206961352-206961352 on Build GRCh38
rs758876024 Chr.1: 206961602-206961602 on Build GRCh38
rs200011175 Chr.1: 206961169-206961169 on Build GRCh38
rs893542132 Chr.1: 206961587-206961587 on Build GRCh38
rs1032024961 Chr.1: 206961188-206961188 on Build GRCh38
rs893573373 Chr.1: 206961211-206961211 on Build GRCh38
rs768213670 Chr.1: 206961182-206961182 on Build GRCh38
rs193085555 Chr.1: 206961418-206961418 on Build GRCh38
rs763503910 Chr.1: 206961174-206961174 on Build GRCh38
rs939981945 Chr.1: 206961410-206961410 on Build GRCh38
rs911099135 Chr.1: 206961465-206961465 on Build GRCh38
rs901277909 Chr.1: 206961170-206961170 on Build GRCh38
rs376426554 Chr.1: 206961185-206961185 on Build GRCh38
rs765352860 Chr.1: 206961362-206961362 on Build GRCh38
rs915783797 Chr.1: 206961351-206961351 on Build GRCh38
rs998275131 Chr.1: 206961436-206961436 on Build GRCh38
rs909853053 Chr.1: 206961369-206961369 on Build GRCh38
rs117620733 Chr.1: 206961246-206961246 on Build GRCh38
rs954402599 Chr.1: 206961206-206961206 on Build GRCh38
rs757629990 Chr.1: 206961218-206961218 on Build GRCh38
rs889995707 Chr.1: 206961531-206961531 on Build GRCh38
rs992580226 Chr.1: 206961340-206961340 on Build GRCh38
rs552556338 Chr.1: 206961601-206961601 on Build GRCh38
rs201258954 Chr.1: 206961208-206961208 on Build GRCh38
rs752944927 Chr.1: 206961573-206961573 on Build GRCh38
rs1011984051 Chr.1: 206961589-206961589 on Build GRCh38
rs541351315 Chr.1: 206961273-206961273 on Build GRCh38
rs369380685 Chr.1: 206961157-206961157 on Build GRCh38
rs943873073 Chr.1: 206961468-206961468 on Build GRCh38
rs532496254 Chr.1: 206961527-206961527 on Build GRCh38
rs563633412 Chr.1: 206961419-206961419 on Build GRCh38
rs1053406814 Chr.1: 206961584-206961584 on Build GRCh38
rs1050346587 Chr.1: 206961526-206961526 on Build GRCh38
rs552971213 Chr.1: 206961219-206961219 on Build GRCh38
rs372580744 Chr.1: 206961203-206961203 on Build GRCh38
rs1038601734 Chr.1: 206961181-206961182 on Build GRCh38
rs977074432 Chr.1: 206961268-206961268 on Build GRCh38

View Full Product Details