Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 207113998-207114458 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C4BPA
Entrez Gene ID
722
Gene Name
complement component 4 binding protein alpha
Gene Aliases
C4BP, PRP
UniGene
Hs.1012
Gene Chromosome Location
Chr.1: 207104232-207144972 on Build GRCh38
Transcripts
NM_000715.3 XM_005273251.1 XM_005273252.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1928110 c.194C>T p.T65M Chr.1: 207114151-207114151 on Build GRCh38
6060847 c.293C>A p.S98Y Chr.1: 207114250-207114250 on Build GRCh38
1928113 c.321C>T p.F107F Chr.1: 207114278-207114278 on Build GRCh38
4772929 c.155C>A p.P52H Chr.1: 207114112-207114112 on Build GRCh38
5589920 c.202G>A p.E68K Chr.1: 207114159-207114159 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs774080937 Chr.1: 207114241-207114241 on Build GRCh38
rs17020956 Chr.1: 207114136-207114136 on Build GRCh38
rs749573985 Chr.1: 207114321-207114321 on Build GRCh38
rs747022283 Chr.1: 207114190-207114190 on Build GRCh38
rs756830651 Chr.1: 207114150-207114150 on Build GRCh38
rs754833771 Chr.1: 207114272-207114272 on Build GRCh38
rs765651659 Chr.1: 207114091-207114091 on Build GRCh38
rs1025506359 Chr.1: 207114415-207114415 on Build GRCh38
rs778061020 Chr.1: 207114184-207114184 on Build GRCh38
rs747282623 Chr.1: 207114282-207114282 on Build GRCh38
rs755403317 Chr.1: 207114077-207114077 on Build GRCh38
rs757197172 Chr.1: 207114050-207114050 on Build GRCh38
rs370737185 Chr.1: 207114235-207114235 on Build GRCh38
rs965160959 Chr.1: 207114378-207114378 on Build GRCh38
rs757329946 Chr.1: 207114128-207114140 on Build GRCh38
rs376241781 Chr.1: 207114213-207114213 on Build GRCh38
rs781316367 Chr.1: 207114299-207114299 on Build GRCh38
rs1054184973 Chr.1: 207114080-207114080 on Build GRCh38
rs765637075 Chr.1: 207114125-207114125 on Build GRCh38
rs761354473 Chr.1: 207114242-207114242 on Build GRCh38
rs552530711 Chr.1: 207114183-207114183 on Build GRCh38
rs369914937 Chr.1: 207114206-207114206 on Build GRCh38
rs781285524 Chr.1: 207114061-207114061 on Build GRCh38
rs562959071 Chr.1: 207114310-207114310 on Build GRCh38
rs772191838 Chr.1: 207114096-207114096 on Build GRCh38
rs148489140 Chr.1: 207114258-207114258 on Build GRCh38
rs369081234 Chr.1: 207114073-207114073 on Build GRCh38
rs771022113 Chr.1: 207114212-207114212 on Build GRCh38
rs772453050 Chr.1: 207114386-207114386 on Build GRCh38
rs989536462 Chr.1: 207114417-207114417 on Build GRCh38
rs149247641 Chr.1: 207114244-207114244 on Build GRCh38
rs750971991 Chr.1: 207114147-207114147 on Build GRCh38
rs748392732 Chr.1: 207114172-207114172 on Build GRCh38
rs753511286 Chr.1: 207114264-207114264 on Build GRCh38
rs777811856 Chr.1: 207114278-207114278 on Build GRCh38
rs915191799 Chr.1: 207114428-207114428 on Build GRCh38
rs745587081 Chr.1: 207114068-207114068 on Build GRCh38
rs952528698 Chr.1: 207114324-207114324 on Build GRCh38
rs769084248 Chr.1: 207114326-207114326 on Build GRCh38
rs746412562 Chr.1: 207114223-207114223 on Build GRCh38
rs147172618 Chr.1: 207114166-207114166 on Build GRCh38
rs1001512697 Chr.1: 207114064-207114064 on Build GRCh38
rs773226295 Chr.1: 207114101-207114101 on Build GRCh38
rs759844205 Chr.1: 207114120-207114120 on Build GRCh38
rs763385898 Chr.1: 207114236-207114236 on Build GRCh38
rs762579470 Chr.1: 207114139-207114139 on Build GRCh38
rs1003813666 Chr.1: 207114170-207114170 on Build GRCh38
rs533950846 Chr.1: 207114195-207114195 on Build GRCh38
rs777155856 Chr.1: 207114112-207114112 on Build GRCh38
rs770138949 Chr.1: 207114234-207114234 on Build GRCh38
rs201739512 Chr.1: 207114301-207114301 on Build GRCh38
rs921222194 Chr.1: 207114406-207114406 on Build GRCh38
rs893916409 Chr.1: 207114086-207114086 on Build GRCh38
rs753257530 Chr.1: 207114314-207114314 on Build GRCh38
rs767519496 Chr.1: 207114151-207114151 on Build GRCh38
rs750384904 Chr.1: 207114252-207114252 on Build GRCh38
rs756182911 Chr.1: 207114257-207114257 on Build GRCh38
rs757294011 Chr.1: 207114290-207114290 on Build GRCh38
rs142009017 Chr.1: 207114103-207114103 on Build GRCh38
rs535925890 Chr.1: 207114405-207114405 on Build GRCh38
rs779259212 Chr.1: 207114074-207114074 on Build GRCh38
rs750438201 Chr.1: 207114152-207114152 on Build GRCh38
rs749042273 Chr.1: 207114372-207114372 on Build GRCh38
rs779026666 Chr.1: 207114302-207114303 on Build GRCh38
rs373033326 Chr.1: 207114164-207114164 on Build GRCh38
rs548541174 Chr.1: 207114353-207114353 on Build GRCh38
rs56180080 Chr.1: 207114316-207114316 on Build GRCh38
rs796589446 Chr.1: 207114056-207114056 on Build GRCh38
rs1012607185 Chr.1: 207114293-207114293 on Build GRCh38
rs763643517 Chr.1: 207114140-207114140 on Build GRCh38
rs746359674 Chr.1: 207114311-207114311 on Build GRCh38
rs943966641 Chr.1: 207114045-207114045 on Build GRCh38
rs375229033 Chr.1: 207114289-207114289 on Build GRCh38
rs954082303 Chr.1: 207114413-207114413 on Build GRCh38
rs773848071 Chr.1: 207114328-207114328 on Build GRCh38

View Full Product Details