Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 207677022-207677569 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CR1L
Entrez Gene ID
1379
Gene Name
complement component 3b/4b receptor 1-like
Gene Aliases
-
UniGene
Hs.628568
Gene Chromosome Location
Chr.1: 207645113-207723691 on Build GRCh38
Transcripts
NM_175710.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5053041 c.300A>G p.K100K Chr.1: 207677567-207677567 on Build GRCh38
5617766 c.128T>C p.F43S Chr.1: 207677419-207677419 on Build GRCh38
3864340 c.190C>T p.P64S Chr.1: 207677457-207677457 on Build GRCh38
6635088 c.175A>G p.T59A Chr.1: 207677466-207677466 on Build GRCh38
4764492 c.197G>A p.R66H Chr.1: 207677488-207677488 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs11118328 Chr.1: 207677194-207677194 on Build GRCh38
rs67422482 Chr.1: 207677215-207677215 on Build GRCh38
rs376930215 Chr.1: 207677502-207677502 on Build GRCh38
rs779704034 Chr.1: 207677354-207677354 on Build GRCh38
rs192078528 Chr.1: 207677488-207677488 on Build GRCh38
rs1040482732 Chr.1: 207677195-207677195 on Build GRCh38
rs557417304 Chr.1: 207677362-207677362 on Build GRCh38
rs1004821823 Chr.1: 207677077-207677077 on Build GRCh38
rs979554643 Chr.1: 207677177-207677177 on Build GRCh38
rs34451130 Chr.1: 207677244-207677244 on Build GRCh38
rs969470114 Chr.1: 207677141-207677141 on Build GRCh38
rs969414245 Chr.1: 207677336-207677336 on Build GRCh38
rs1005174804 Chr.1: 207677245-207677245 on Build GRCh38
rs538431014 Chr.1: 207677262-207677262 on Build GRCh38
rs778860020 Chr.1: 207677447-207677447 on Build GRCh38
rs764512095 Chr.1: 207677479-207677479 on Build GRCh38
rs187761228 Chr.1: 207677179-207677179 on Build GRCh38
rs781203518 Chr.1: 207677436-207677439 on Build GRCh38
rs896392624 Chr.1: 207677506-207677506 on Build GRCh38
rs896434901 Chr.1: 207677258-207677258 on Build GRCh38
rs776437165 Chr.1: 207677436-207677436 on Build GRCh38
rs565216969 Chr.1: 207677178-207677178 on Build GRCh38
rs377664418 Chr.1: 207677394-207677394 on Build GRCh38
rs184089830 Chr.1: 207677501-207677501 on Build GRCh38
rs777667413 Chr.1: 207677539-207677539 on Build GRCh38
rs745380158 Chr.1: 207677342-207677342 on Build GRCh38
rs565635329 Chr.1: 207677350-207677350 on Build GRCh38
rs747900861 Chr.1: 207677448-207677448 on Build GRCh38
rs183894980 Chr.1: 207677398-207677398 on Build GRCh38
rs188536074 Chr.1: 207677449-207677449 on Build GRCh38
rs912855131 Chr.1: 207677185-207677185 on Build GRCh38
rs960748417 Chr.1: 207677092-207677092 on Build GRCh38
rs377607560 Chr.1: 207677461-207677461 on Build GRCh38
rs779759328 Chr.1: 207677435-207677435 on Build GRCh38
rs764601215 Chr.1: 207677481-207677481 on Build GRCh38
rs748175888 Chr.1: 207677533-207677533 on Build GRCh38
rs764163141 Chr.1: 207677226-207677226 on Build GRCh38
rs549700045 Chr.1: 207677526-207677526 on Build GRCh38
rs1023675097 Chr.1: 207677132-207677132 on Build GRCh38
rs887666056 Chr.1: 207677055-207677055 on Build GRCh38
rs764547935 Chr.1: 207677402-207677402 on Build GRCh38
rs574438364 Chr.1: 207677415-207677415 on Build GRCh38
rs751894755 Chr.1: 207677404-207677404 on Build GRCh38
rs115303620 Chr.1: 207677419-207677419 on Build GRCh38
rs74231794 Chr.1: 207677372-207677372 on Build GRCh38
rs762973767 Chr.1: 207677476-207677476 on Build GRCh38
rs1013572378 Chr.1: 207677288-207677288 on Build GRCh38
rs370187813 Chr.1: 207677365-207677365 on Build GRCh38
rs142146162 Chr.1: 207677140-207677140 on Build GRCh38
rs552278382 Chr.1: 207677263-207677263 on Build GRCh38
rs373799944 Chr.1: 207677446-207677446 on Build GRCh38
rs1036275976 Chr.1: 207677251-207677251 on Build GRCh38
rs757581505 Chr.1: 207677405-207677405 on Build GRCh38
rs202202158 Chr.1: 207677465-207677465 on Build GRCh38
rs191975411 Chr.1: 207677238-207677238 on Build GRCh38
rs771073527 Chr.1: 207677384-207677384 on Build GRCh38
rs561296768 Chr.1: 207677214-207677214 on Build GRCh38
rs776848671 Chr.1: 207677388-207677388 on Build GRCh38
rs774052674 Chr.1: 207677367-207677367 on Build GRCh38
rs755299963 Chr.1: 207677051-207677051 on Build GRCh38
rs572460849 Chr.1: 207677468-207677468 on Build GRCh38
rs1023622783 Chr.1: 207677335-207677335 on Build GRCh38
rs34509370 Chr.1: 207677534-207677534 on Build GRCh38
rs750728001 Chr.1: 207677509-207677509 on Build GRCh38
rs759674020 Chr.1: 207677392-207677392 on Build GRCh38
rs769359307 Chr.1: 207677352-207677352 on Build GRCh38
rs921661135 Chr.1: 207677201-207677201 on Build GRCh38
rs1014039443 Chr.1: 207677095-207677095 on Build GRCh38
rs778285001 Chr.1: 207677524-207677524 on Build GRCh38
rs780850710 Chr.1: 207677339-207677339 on Build GRCh38
rs755811490 Chr.1: 207677423-207677423 on Build GRCh38
rs529897154 Chr.1: 207677413-207677413 on Build GRCh38
rs748067438 Chr.1: 207677441-207677441 on Build GRCh38
rs530003963 Chr.1: 207677519-207677519 on Build GRCh38
rs1015301217 Chr.1: 207677090-207677090 on Build GRCh38
rs753420337 Chr.1: 207677510-207677510 on Build GRCh38
rs57151160 Chr.1: 207677332-207677337 on Build GRCh38
rs769902083 Chr.1: 207677508-207677508 on Build GRCh38
rs377244503 Chr.1: 207677460-207677460 on Build GRCh38

View Full Product Details