Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 209836469-209836977 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DIEXF
Entrez Gene ID
27042
Gene Name
digestive organ expansion factor homolog (zebrafish)
Gene Aliases
C1orf107, DEF, DJ434O14.5, UTP25
UniGene
Hs.194754
Gene Chromosome Location
Chr.1: 209827967-209857565 on Build GRCh38
Transcripts
NM_014388.6 XM_006711275.3 XM_011509418.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
40322 c.652-1G>A p.? Chr.1: 209836800-209836800 on Build GRCh38
4846440 c.700G>A p.E234K Chr.1: 209836849-209836849 on Build GRCh38
5934281 c.715C>T p.P239S Chr.1: 209836864-209836864 on Build GRCh38
6541674 c.782A>T p.N261I Chr.1: 209836931-209836931 on Build GRCh38
5542831 c.802C>T p.P268S Chr.1: 209836951-209836951 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs755721451 Chr.1: 209836766-209836766 on Build GRCh38
rs575622401 Chr.1: 209836774-209836774 on Build GRCh38
rs547446607 Chr.1: 209836649-209836649 on Build GRCh38
rs778845850 Chr.1: 209836779-209836779 on Build GRCh38
rs151234588 Chr.1: 209836621-209836621 on Build GRCh38
rs898832376 Chr.1: 209836725-209836725 on Build GRCh38
rs11119352 Chr.1: 209836775-209836775 on Build GRCh38
rs887764019 Chr.1: 209836651-209836651 on Build GRCh38
rs752661750 Chr.1: 209836909-209836909 on Build GRCh38
rs1037737102 Chr.1: 209836724-209836724 on Build GRCh38
rs76438611 Chr.1: 209836931-209836931 on Build GRCh38
rs1005146098 Chr.1: 209836669-209836669 on Build GRCh38
rs770017983 Chr.1: 209836946-209836946 on Build GRCh38
rs1032881967 Chr.1: 209836932-209836932 on Build GRCh38
rs746058692 Chr.1: 209836801-209836801 on Build GRCh38
rs777576505 Chr.1: 209836937-209836937 on Build GRCh38
rs749384173 Chr.1: 209836951-209836951 on Build GRCh38
rs767959214 Chr.1: 209836855-209836855 on Build GRCh38
rs533314685 Chr.1: 209836762-209836762 on Build GRCh38
rs926339392 Chr.1: 209836654-209836654 on Build GRCh38
rs766069202 Chr.1: 209836871-209836871 on Build GRCh38
rs778717225 Chr.1: 209836854-209836854 on Build GRCh38
rs559541454 Chr.1: 209836674-209836674 on Build GRCh38
rs754509432 Chr.1: 209836881-209836881 on Build GRCh38
rs912412304 Chr.1: 209836556-209836556 on Build GRCh38
rs537745859 Chr.1: 209836943-209836943 on Build GRCh38
rs775570004 Chr.1: 209836813-209836813 on Build GRCh38
rs368563474 Chr.1: 209836836-209836836 on Build GRCh38
rs142891522 Chr.1: 209836636-209836636 on Build GRCh38
rs979128555 Chr.1: 209836497-209836497 on Build GRCh38
rs778777754 Chr.1: 209836904-209836904 on Build GRCh38
rs767337332 Chr.1: 209836756-209836756 on Build GRCh38
rs377646967 Chr.1: 209836506-209836506 on Build GRCh38
rs913855920 Chr.1: 209836618-209836618 on Build GRCh38
rs771891452 Chr.1: 209836794-209836794 on Build GRCh38
rs988018031 Chr.1: 209836616-209836616 on Build GRCh38
rs1032353218 Chr.1: 209836580-209836580 on Build GRCh38
rs960908848 Chr.1: 209836771-209836771 on Build GRCh38
rs779739167 Chr.1: 209836772-209836772 on Build GRCh38
rs763021292 Chr.1: 209836821-209836821 on Build GRCh38
rs979158002 Chr.1: 209836845-209836845 on Build GRCh38
rs74427459 Chr.1: 209836543-209836543 on Build GRCh38
rs769966467 Chr.1: 209836805-209836805 on Build GRCh38
rs1052770002 Chr.1: 209836544-209836544 on Build GRCh38
rs943828825 Chr.1: 209836588-209836588 on Build GRCh38
rs548836130 Chr.1: 209836602-209836602 on Build GRCh38
rs141256304 Chr.1: 209836832-209836832 on Build GRCh38
rs981936883 Chr.1: 209836653-209836653 on Build GRCh38
rs144243504 Chr.1: 209836859-209836859 on Build GRCh38
rs146066445 Chr.1: 209836864-209836864 on Build GRCh38
rs143934578 Chr.1: 209836874-209836874 on Build GRCh38
rs549153041 Chr.1: 209836947-209836947 on Build GRCh38
rs1023803866 Chr.1: 209836830-209836830 on Build GRCh38
rs924938177 Chr.1: 209836503-209836503 on Build GRCh38
rs756858660 Chr.1: 209836741-209836741 on Build GRCh38
rs267598352 Chr.1: 209836903-209836903 on Build GRCh38
rs935259662 Chr.1: 209836537-209836537 on Build GRCh38
rs747964444 Chr.1: 209836788-209836788 on Build GRCh38
rs768757354 Chr.1: 209836952-209836952 on Build GRCh38
rs781443110 Chr.1: 209836795-209836795 on Build GRCh38

View Full Product Details