Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 212071970-212072515 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DTL
Entrez Gene ID
51514
Gene Name
denticleless E3 ubiquitin protein ligase homolog
Gene Aliases
CDT2, DCAF2, L2DTL, RAMP
UniGene
Hs.656473
Gene Chromosome Location
Chr.1: 212035553-212105006 on Build GRCh38
Transcripts
NM_001286229.1 NM_001286230.1 NM_016448.3 XM_011509614.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7326183 c.928A>G p.I310V Chr.1: 212072106-212072106 on Build GRCh38
4882841 c.1001G>T p.S334I Chr.1: 212072179-212072179 on Build GRCh38
6061391 c.959A>C p.Y320S Chr.1: 212072137-212072137 on Build GRCh38
5788154 c.938G>A p.G313E Chr.1: 212072116-212072116 on Build GRCh38
7400860 c.985G>A p.D329N Chr.1: 212072163-212072163 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs754445116 Chr.1: 212072175-212072175 on Build GRCh38
rs932537250 Chr.1: 212072296-212072296 on Build GRCh38
rs554559180 Chr.1: 212072436-212072436 on Build GRCh38
rs895602761 Chr.1: 212072093-212072093 on Build GRCh38
rs779173861 Chr.1: 212072090-212072090 on Build GRCh38
rs878887714 Chr.1: 212072087-212072087 on Build GRCh38
rs374261828 Chr.1: 212072058-212072058 on Build GRCh38
rs745838994 Chr.1: 212072231-212072231 on Build GRCh38
rs764496974 Chr.1: 212072353-212072353 on Build GRCh38
rs754196340 Chr.1: 212072408-212072408 on Build GRCh38
rs375041545 Chr.1: 212072227-212072227 on Build GRCh38
rs200632812 Chr.1: 212072083-212072083 on Build GRCh38
rs536400371 Chr.1: 212072320-212072320 on Build GRCh38
rs757095816 Chr.1: 212072223-212072223 on Build GRCh38
rs763877768 Chr.1: 212072210-212072210 on Build GRCh38
rs753964130 Chr.1: 212072070-212072070 on Build GRCh38
rs142217683 Chr.1: 212072135-212072135 on Build GRCh38
rs775692209 Chr.1: 212072298-212072298 on Build GRCh38
rs762537119 Chr.1: 212072127-212072127 on Build GRCh38
rs768189545 Chr.1: 212072265-212072265 on Build GRCh38
rs574557127 Chr.1: 212072276-212072276 on Build GRCh38
rs752900381 Chr.1: 212072091-212072091 on Build GRCh38
rs990651260 Chr.1: 212072235-212072235 on Build GRCh38
rs540487470 Chr.1: 212072488-212072488 on Build GRCh38
rs1048317077 Chr.1: 212072301-212072301 on Build GRCh38
rs567322508 Chr.1: 212071995-212071995 on Build GRCh38
rs974848136 Chr.1: 212072455-212072455 on Build GRCh38
rs779445163 Chr.1: 212072254-212072254 on Build GRCh38
rs766500536 Chr.1: 212072061-212072061 on Build GRCh38
rs759654934 Chr.1: 212072179-212072179 on Build GRCh38
rs921956722 Chr.1: 212072295-212072295 on Build GRCh38
rs752912904 Chr.1: 212072200-212072200 on Build GRCh38
rs1026707089 Chr.1: 212072068-212072068 on Build GRCh38
rs766978008 Chr.1: 212072158-212072158 on Build GRCh38
rs781170147 Chr.1: 212072114-212072114 on Build GRCh38
rs576157934 Chr.1: 212072457-212072457 on Build GRCh38
rs144876166 Chr.1: 212072113-212072113 on Build GRCh38
rs751347042 Chr.1: 212072213-212072213 on Build GRCh38
rs1048622600 Chr.1: 212072383-212072383 on Build GRCh38
rs1036732767 Chr.1: 212072006-212072006 on Build GRCh38
rs928780455 Chr.1: 212072280-212072280 on Build GRCh38
rs918270753 Chr.1: 212072346-212072346 on Build GRCh38
rs745364873 Chr.1: 212072120-212072120 on Build GRCh38
rs2251398 Chr.1: 212072450-212072450 on Build GRCh38
rs17018435 Chr.1: 212072306-212072306 on Build GRCh38
rs755604151 Chr.1: 212072248-212072248 on Build GRCh38
rs765262770 Chr.1: 212072187-212072187 on Build GRCh38
rs772425233 Chr.1: 212072129-212072129 on Build GRCh38
rs748905951 Chr.1: 212072261-212072261 on Build GRCh38
rs1014336795 Chr.1: 212072106-212072106 on Build GRCh38
rs1019145148 Chr.1: 212072438-212072438 on Build GRCh38
rs558601077 Chr.1: 212072490-212072490 on Build GRCh38
rs367760119 Chr.1: 212072252-212072252 on Build GRCh38
rs140344470 Chr.1: 212072205-212072205 on Build GRCh38
rs12145020 Chr.1: 212072002-212072002 on Build GRCh38
rs1040888501 Chr.1: 212072451-212072451 on Build GRCh38
rs999833506 Chr.1: 212072398-212072398 on Build GRCh38
rs1037263071 Chr.1: 212072317-212072317 on Build GRCh38
rs888377377 Chr.1: 212072302-212072302 on Build GRCh38
rs202197897 Chr.1: 212072123-212072123 on Build GRCh38
rs889584580 Chr.1: 212072416-212072416 on Build GRCh38
rs761188839 Chr.1: 212072147-212072147 on Build GRCh38
rs896807247 Chr.1: 212072010-212072010 on Build GRCh38
rs1022570811 Chr.1: 212072380-212072380 on Build GRCh38
rs1033245493 Chr.1: 212072167-212072167 on Build GRCh38
rs758170384 Chr.1: 212072099-212072099 on Build GRCh38
rs370526029 Chr.1: 212072111-212072111 on Build GRCh38
rs1031654053 Chr.1: 212072226-212072226 on Build GRCh38
rs546990270 Chr.1: 212072051-212072051 on Build GRCh38
rs545072692 Chr.1: 212072486-212072486 on Build GRCh38

View Full Product Details