Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 214603100-214603589 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CENPF
Entrez Gene ID
1063
Gene Name
centromere protein F
Gene Aliases
CENF, CILD31, PRO1779, STROMS, hcp-1
UniGene
Hs.497741
Gene Chromosome Location
Chr.1: 214603181-214664574 on Build GRCh38
Transcripts
NM_016343.3 XM_011509082.2

SNP Detail

SNP ID Chromosome Location
rs936966784 Chr.1: 214603263-214603263 on Build GRCh38
rs936916983 Chr.1: 214603435-214603435 on Build GRCh38
rs758311312 Chr.1: 214603204-214603204 on Build GRCh38
rs1009442446 Chr.1: 214603279-214603279 on Build GRCh38
rs900742154 Chr.1: 214603422-214603422 on Build GRCh38
rs937777642 Chr.1: 214603156-214603156 on Build GRCh38
rs529834560 Chr.1: 214603534-214603534 on Build GRCh38
rs1028343399 Chr.1: 214603426-214603426 on Build GRCh38
rs970814807 Chr.1: 214603423-214603423 on Build GRCh38
rs372480170 Chr.1: 214603165-214603165 on Build GRCh38
rs3748691 Chr.1: 214603264-214603264 on Build GRCh38
rs574947473 Chr.1: 214603135-214603135 on Build GRCh38
rs1025951949 Chr.1: 214603307-214603307 on Build GRCh38
rs768351710 Chr.1: 214603379-214603379 on Build GRCh38
rs1467083 Chr.1: 214603303-214603303 on Build GRCh38
rs745667841 Chr.1: 214603149-214603149 on Build GRCh38
rs943106449 Chr.1: 214603287-214603287 on Build GRCh38
rs983122106 Chr.1: 214603561-214603561 on Build GRCh38
rs959268634 Chr.1: 214603324-214603324 on Build GRCh38
rs967966370 Chr.1: 214603292-214603292 on Build GRCh38
rs368278693 Chr.1: 214603483-214603483 on Build GRCh38
rs900479462 Chr.1: 214603254-214603254 on Build GRCh38
rs544668185 Chr.1: 214603270-214603270 on Build GRCh38
rs990652422 Chr.1: 214603349-214603349 on Build GRCh38
rs377078645 Chr.1: 214603303-214603303 on Build GRCh38
rs1006418683 Chr.1: 214603250-214603250 on Build GRCh38
rs140934727 Chr.1: 214603118-214603118 on Build GRCh38
rs2809588 Chr.1: 214603537-214603537 on Build GRCh38
rs924159237 Chr.1: 214603263-214603263 on Build GRCh38
rs888066659 Chr.1: 214603239-214603239 on Build GRCh38
rs989783363 Chr.1: 214603442-214603442 on Build GRCh38
rs1000802835 Chr.1: 214603488-214603488 on Build GRCh38
rs929052541 Chr.1: 214603362-214603362 on Build GRCh38
rs933101633 Chr.1: 214603546-214603546 on Build GRCh38
rs926325925 Chr.1: 214603311-214603311 on Build GRCh38
rs959300576 Chr.1: 214603532-214603532 on Build GRCh38
rs1467084 Chr.1: 214603306-214603306 on Build GRCh38
rs917688968 Chr.1: 214603350-214603350 on Build GRCh38
rs929504509 Chr.1: 214603218-214603218 on Build GRCh38
rs1020803949 Chr.1: 214603474-214603474 on Build GRCh38
rs1007334617 Chr.1: 214603215-214603215 on Build GRCh38
rs780855797 Chr.1: 214603485-214603485 on Build GRCh38
rs901604387 Chr.1: 214603305-214603305 on Build GRCh38
rs41277186 Chr.1: 214603487-214603487 on Build GRCh38
rs918105197 Chr.1: 214603180-214603180 on Build GRCh38
rs956092032 Chr.1: 214603276-214603276 on Build GRCh38
rs570925404 Chr.1: 214603146-214603146 on Build GRCh38
rs1006175468 Chr.1: 214603329-214603329 on Build GRCh38
rs1016129033 Chr.1: 214603243-214603243 on Build GRCh38
rs188564247 Chr.1: 214603445-214603445 on Build GRCh38
rs1020792589 Chr.1: 214603291-214603291 on Build GRCh38
rs1016500984 Chr.1: 214603330-214603330 on Build GRCh38
rs144701714 Chr.1: 214603377-214603377 on Build GRCh38
rs949323491 Chr.1: 214603252-214603252 on Build GRCh38
rs1039041183 Chr.1: 214603253-214603253 on Build GRCh38
rs950477504 Chr.1: 214603541-214603541 on Build GRCh38
rs1034042264 Chr.1: 214603493-214603493 on Build GRCh38
rs1056591210 Chr.1: 214603161-214603161 on Build GRCh38
rs923988567 Chr.1: 214603428-214603428 on Build GRCh38
rs933343026 Chr.1: 214603427-214603427 on Build GRCh38
rs926249248 Chr.1: 214603147-214603147 on Build GRCh38
rs1011973753 Chr.1: 214603414-214603414 on Build GRCh38
rs911471336 Chr.1: 214603480-214603480 on Build GRCh38
rs913844023 Chr.1: 214603126-214603126 on Build GRCh38
rs983616042 Chr.1: 214603368-214603368 on Build GRCh38
rs1010292220 Chr.1: 214603473-214603473 on Build GRCh38
rs891920194 Chr.1: 214603452-214603452 on Build GRCh38
rs551280620 Chr.1: 214603142-214603142 on Build GRCh38
rs991824809 Chr.1: 214603343-214603343 on Build GRCh38
rs1026151945 Chr.1: 214603229-214603229 on Build GRCh38
rs768638817 Chr.1: 214603519-214603519 on Build GRCh38
rs918012614 Chr.1: 214603251-214603251 on Build GRCh38
rs950684172 Chr.1: 214603233-214603233 on Build GRCh38
rs554146658 Chr.1: 214603152-214603152 on Build GRCh38
rs41277188 Chr.1: 214603562-214603562 on Build GRCh38
rs959717251 Chr.1: 214603244-214603244 on Build GRCh38
rs1047865021 Chr.1: 214603230-214603230 on Build GRCh38
rs942318282 Chr.1: 214603382-214603382 on Build GRCh38
rs1017221988 Chr.1: 214603216-214603216 on Build GRCh38

View Full Product Details