Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 97866012-97866509 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CRTAC1
Entrez Gene ID
55118
Gene Name
cartilage acidic protein 1
Gene Aliases
ASPIC, ASPIC1, CEP-68
UniGene
Hs.500736
Gene Chromosome Location
Chr.10: 97854711-98030828 on Build GRCh38
Transcripts
NM_018058.6 XM_011539917.1 XM_017016367.1 NM_001010917.2

Gene Symbol
GOLGA7B
Entrez Gene ID
401647
Gene Name
golgin A7 family member B
Gene Aliases
C10orf132, C10orf133, bA451M19.3, bA459F3.4
UniGene
Hs.567524
Gene Chromosome Location
Chr.10: 97850238-97871578 on Build GRCh38
Transcripts
NM_018058.6 XM_011539917.1 XM_017016367.1 NM_001010917.2

SNP Detail

SNP ID Chromosome Location
rs760870381 Chr.10: 97866438-97866438 on Build GRCh38
rs1010206813 Chr.10: 97866069-97866069 on Build GRCh38
rs1017502987 Chr.10: 97866093-97866093 on Build GRCh38
rs912592229 Chr.10: 97866202-97866202 on Build GRCh38
rs1054260037 Chr.10: 97866472-97866472 on Build GRCh38
rs1000986988 Chr.10: 97866045-97866046 on Build GRCh38
rs527673482 Chr.10: 97866275-97866275 on Build GRCh38
rs549927337 Chr.10: 97866439-97866439 on Build GRCh38
rs545377570 Chr.10: 97866343-97866343 on Build GRCh38
rs967425159 Chr.10: 97866483-97866483 on Build GRCh38
rs535883681 Chr.10: 97866033-97866033 on Build GRCh38
rs1027781990 Chr.10: 97866242-97866242 on Build GRCh38
rs562988245 Chr.10: 97866192-97866192 on Build GRCh38
rs965985806 Chr.10: 97866291-97866291 on Build GRCh38
rs145921870 Chr.10: 97866188-97866188 on Build GRCh38
rs1029669622 Chr.10: 97866324-97866324 on Build GRCh38
rs936145224 Chr.10: 97866145-97866145 on Build GRCh38
rs997452223 Chr.10: 97866473-97866473 on Build GRCh38
rs1053972781 Chr.10: 97866251-97866251 on Build GRCh38
rs918937512 Chr.10: 97866392-97866392 on Build GRCh38
rs1027527989 Chr.10: 97866046-97866046 on Build GRCh38
rs370632894 Chr.10: 97866199-97866199 on Build GRCh38
rs1039803359 Chr.10: 97866224-97866224 on Build GRCh38
rs762610534 Chr.10: 97866299-97866299 on Build GRCh38
rs1046365977 Chr.10: 97866391-97866391 on Build GRCh38
rs773358758 Chr.10: 97866338-97866338 on Build GRCh38
rs1026838145 Chr.10: 97866384-97866385 on Build GRCh38
rs977548440 Chr.10: 97866330-97866330 on Build GRCh38
rs554579150 Chr.10: 97866053-97866053 on Build GRCh38
rs1046668104 Chr.10: 97866288-97866288 on Build GRCh38
rs531796350 Chr.10: 97866386-97866386 on Build GRCh38
rs552495352 Chr.10: 97866320-97866320 on Build GRCh38
rs748332459 Chr.10: 97866138-97866138 on Build GRCh38
rs114750996 Chr.10: 97866191-97866191 on Build GRCh38
rs536419504 Chr.10: 97866140-97866140 on Build GRCh38
rs946194458 Chr.10: 97866139-97866139 on Build GRCh38
rs976808279 Chr.10: 97866135-97866135 on Build GRCh38
rs374297457 Chr.10: 97866331-97866331 on Build GRCh38
rs200435847 Chr.10: 97866119-97866119 on Build GRCh38
rs958566691 Chr.10: 97866159-97866160 on Build GRCh38
rs1019965306 Chr.10: 97866077-97866077 on Build GRCh38
rs967751245 Chr.10: 97866327-97866327 on Build GRCh38
rs1007307899 Chr.10: 97866455-97866455 on Build GRCh38
rs944035309 Chr.10: 97866206-97866206 on Build GRCh38
rs540561500 Chr.10: 97866168-97866168 on Build GRCh38
rs190687005 Chr.10: 97866247-97866247 on Build GRCh38
rs993263079 Chr.10: 97866240-97866240 on Build GRCh38
rs1034091094 Chr.10: 97866126-97866126 on Build GRCh38
rs560566628 Chr.10: 97866272-97866272 on Build GRCh38
rs916069763 Chr.10: 97866363-97866363 on Build GRCh38
rs541934506 Chr.10: 97866267-97866267 on Build GRCh38
rs564392223 Chr.10: 97866346-97866346 on Build GRCh38
rs573440787 Chr.10: 97866239-97866239 on Build GRCh38
rs965935180 Chr.10: 97866109-97866109 on Build GRCh38
rs112863923 Chr.10: 97866182-97866182 on Build GRCh38
rs574850325 Chr.10: 97866230-97866230 on Build GRCh38
rs901640766 Chr.10: 97866467-97866467 on Build GRCh38
rs892607063 Chr.10: 97866266-97866266 on Build GRCh38
rs76918243 Chr.10: 97866481-97866481 on Build GRCh38
rs536382263 Chr.10: 97866459-97866459 on Build GRCh38
rs981215317 Chr.10: 97866107-97866107 on Build GRCh38
rs371331042 Chr.10: 97866144-97866144 on Build GRCh38
rs971030795 Chr.10: 97866091-97866091 on Build GRCh38
rs887556601 Chr.10: 97866440-97866440 on Build GRCh38
rs993642802 Chr.10: 97866044-97866044 on Build GRCh38
rs965229352 Chr.10: 97866397-97866397 on Build GRCh38
rs11189413 Chr.10: 97866061-97866061 on Build GRCh38
rs951277253 Chr.10: 97866385-97866385 on Build GRCh38
rs1007423850 Chr.10: 97866260-97866260 on Build GRCh38
rs568540594 Chr.10: 97866448-97866448 on Build GRCh38

View Full Product Details