Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 99799102-99799604 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ABCC2
Entrez Gene ID
1244
Gene Name
ATP binding cassette subfamily C member 2
Gene Aliases
ABC30, CMOAT, DJS, MRP2, cMRP
UniGene
Hs.368243
Gene Chromosome Location
Chr.10: 99782598-99853741 on Build GRCh38
Transcripts
NM_000392.4 XM_006717630.3 XM_006717631.3 XM_011539291.2 XM_017015675.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
913797 c.1002C>A p.I334I Chr.10: 99799341-99799341 on Build GRCh38
6128252 c.913G>C p.V305L Chr.10: 99799252-99799252 on Build GRCh38
5043971 c.1002C>G p.I334M Chr.10: 99799341-99799341 on Build GRCh38
6590338 c.889_891delAAG p.K298delK Chr.10: 99799228-99799230 on Build GRCh38
5836068 c.904delA p.D304fs*7 Chr.10: 99799243-99799243 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs769945419 Chr.10: 99799310-99799310 on Build GRCh38
rs373391779 Chr.10: 99799410-99799410 on Build GRCh38
rs544849962 Chr.10: 99799531-99799531 on Build GRCh38
rs757061562 Chr.10: 99799397-99799397 on Build GRCh38
rs750931555 Chr.10: 99799208-99799208 on Build GRCh38
rs879391920 Chr.10: 99799405-99799405 on Build GRCh38
rs745760752 Chr.10: 99799418-99799418 on Build GRCh38
rs985000794 Chr.10: 99799571-99799571 on Build GRCh38
rs754424951 Chr.10: 99799220-99799220 on Build GRCh38
rs746097328 Chr.10: 99799558-99799558 on Build GRCh38
rs1049949075 Chr.10: 99799295-99799295 on Build GRCh38
rs751924435 Chr.10: 99799186-99799186 on Build GRCh38
rs111312853 Chr.10: 99799401-99799401 on Build GRCh38
rs931458251 Chr.10: 99799253-99799253 on Build GRCh38
rs199528736 Chr.10: 99799322-99799322 on Build GRCh38
rs765412802 Chr.10: 99799180-99799180 on Build GRCh38
rs766835577 Chr.10: 99799355-99799355 on Build GRCh38
rs371866713 Chr.10: 99799313-99799313 on Build GRCh38
rs959463409 Chr.10: 99799140-99799140 on Build GRCh38
rs1034163605 Chr.10: 99799138-99799138 on Build GRCh38
rs200342237 Chr.10: 99799247-99799247 on Build GRCh38
rs17222674 Chr.10: 99799337-99799337 on Build GRCh38
rs765708935 Chr.10: 99799129-99799129 on Build GRCh38
rs779887351 Chr.10: 99799338-99799338 on Build GRCh38
rs770919031 Chr.10: 99799281-99799281 on Build GRCh38
rs978166050 Chr.10: 99799214-99799214 on Build GRCh38
rs1021504487 Chr.10: 99799438-99799438 on Build GRCh38
rs764701480 Chr.10: 99799392-99799392 on Build GRCh38
rs530962267 Chr.10: 99799554-99799554 on Build GRCh38
rs149854486 Chr.10: 99799346-99799346 on Build GRCh38
rs376851610 Chr.10: 99799332-99799332 on Build GRCh38
rs972755202 Chr.10: 99799525-99799525 on Build GRCh38
rs200435012 Chr.10: 99799184-99799184 on Build GRCh38
rs751178244 Chr.10: 99799384-99799384 on Build GRCh38
rs559521134 Chr.10: 99799473-99799473 on Build GRCh38
rs761197359 Chr.10: 99799347-99799347 on Build GRCh38
rs1031069856 Chr.10: 99799530-99799530 on Build GRCh38
rs778413437 Chr.10: 99799228-99799228 on Build GRCh38
rs373384181 Chr.10: 99799374-99799374 on Build GRCh38
rs565840553 Chr.10: 99799276-99799276 on Build GRCh38
rs990418911 Chr.10: 99799136-99799136 on Build GRCh38
rs992595423 Chr.10: 99799533-99799533 on Build GRCh38
rs992258579 Chr.10: 99799176-99799176 on Build GRCh38
rs768001720 Chr.10: 99799191-99799191 on Build GRCh38
rs539847766 Chr.10: 99799402-99799402 on Build GRCh38
rs550451127 Chr.10: 99799428-99799428 on Build GRCh38
rs768904287 Chr.10: 99799331-99799331 on Build GRCh38
rs775517724 Chr.10: 99799182-99799182 on Build GRCh38
rs959203054 Chr.10: 99799431-99799431 on Build GRCh38
rs940478596 Chr.10: 99799383-99799383 on Build GRCh38
rs553674996 Chr.10: 99799268-99799268 on Build GRCh38
rs763258395 Chr.10: 99799183-99799183 on Build GRCh38
rs1029284698 Chr.10: 99799124-99799124 on Build GRCh38
rs777475470 Chr.10: 99799378-99799378 on Build GRCh38
rs575848842 Chr.10: 99799239-99799239 on Build GRCh38
rs375385616 Chr.10: 99799297-99799297 on Build GRCh38
rs144523808 Chr.10: 99799538-99799538 on Build GRCh38
rs776765875 Chr.10: 99799168-99799168 on Build GRCh38
rs751569886 Chr.10: 99799422-99799422 on Build GRCh38
rs765779011 Chr.10: 99799244-99799256 on Build GRCh38
rs547515246 Chr.10: 99799429-99799429 on Build GRCh38
rs968164660 Chr.10: 99799185-99799185 on Build GRCh38
rs1013634305 Chr.10: 99799440-99799440 on Build GRCh38
rs864309675 Chr.10: 99799352-99799353 on Build GRCh38
rs186268497 Chr.10: 99799508-99799508 on Build GRCh38
rs777159132 Chr.10: 99799163-99799166 on Build GRCh38
rs368723727 Chr.10: 99799404-99799404 on Build GRCh38
rs752550310 Chr.10: 99799428-99799428 on Build GRCh38
rs759685557 Chr.10: 99799178-99799178 on Build GRCh38
rs181542662 Chr.10: 99799323-99799323 on Build GRCh38
rs145841924 Chr.10: 99799353-99799353 on Build GRCh38
rs757613968 Chr.10: 99799190-99799190 on Build GRCh38
rs759781877 Chr.10: 99799221-99799222 on Build GRCh38
rs948342422 Chr.10: 99799570-99799570 on Build GRCh38
rs1025101362 Chr.10: 99799448-99799448 on Build GRCh38
rs939311117 Chr.10: 99799289-99799289 on Build GRCh38

View Full Product Details