Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 100286008-100286503 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BLOC1S2
Entrez Gene ID
282991
Gene Name
biogenesis of lysosomal organelles complex 1 subunit 2
Gene Aliases
BLOS2, BORCS2, CEAP, CEAP11
UniGene
Hs.34906
Gene Chromosome Location
Chr.10: 100273278-100286712 on Build GRCh38
Transcripts
NM_001001342.2 NM_001282436.1 NM_001282437.1 NM_001282438.1 NM_001282439.1 NM_173809.4 XM_017016102.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7164350 c.79G>A p.E27K Chr.10: 100286190-100286190 on Build GRCh38
3952757 c.63C>G p.A21A Chr.10: 100286206-100286206 on Build GRCh38
7143814 c.131T>A p.M44K Chr.10: 100286138-100286138 on Build GRCh38
3806272 c.124C>T p.R42W Chr.10: 100286145-100286145 on Build GRCh38
3790398 c.111C>T p.I37I Chr.10: 100286158-100286158 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs776795501 Chr.10: 100286156-100286156 on Build GRCh38
rs942697857 Chr.10: 100286422-100286422 on Build GRCh38
rs746881044 Chr.10: 100286301-100286301 on Build GRCh38
rs546907090 Chr.10: 100286455-100286455 on Build GRCh38
rs775898626 Chr.10: 100286203-100286203 on Build GRCh38
rs964961850 Chr.10: 100286407-100286407 on Build GRCh38
rs771973021 Chr.10: 100286141-100286141 on Build GRCh38
rs925571215 Chr.10: 100286433-100286433 on Build GRCh38
rs929974088 Chr.10: 100286063-100286063 on Build GRCh38
rs528723294 Chr.10: 100286418-100286418 on Build GRCh38
rs115065418 Chr.10: 100286330-100286330 on Build GRCh38
rs373415529 Chr.10: 100286050-100286050 on Build GRCh38
rs753522565 Chr.10: 100286238-100286238 on Build GRCh38
rs759837705 Chr.10: 100286204-100286204 on Build GRCh38
rs918872492 Chr.10: 100286031-100286031 on Build GRCh38
rs1026142150 Chr.10: 100286283-100286283 on Build GRCh38
rs777897324 Chr.10: 100286142-100286142 on Build GRCh38
rs930253926 Chr.10: 100286380-100286380 on Build GRCh38
rs765622528 Chr.10: 100286060-100286060 on Build GRCh38
rs767047746 Chr.10: 100286093-100286093 on Build GRCh38
rs770489244 Chr.10: 100286358-100286358 on Build GRCh38
rs935707187 Chr.10: 100286123-100286123 on Build GRCh38
rs916320774 Chr.10: 100286298-100286298 on Build GRCh38
rs906925409 Chr.10: 100286275-100286291 on Build GRCh38
rs1049134150 Chr.10: 100286384-100286384 on Build GRCh38
rs945645244 Chr.10: 100286170-100286170 on Build GRCh38
rs747005756 Chr.10: 100286146-100286146 on Build GRCh38
rs911579282 Chr.10: 100286065-100286065 on Build GRCh38
rs796927630 Chr.10: 100286266-100286267 on Build GRCh38
rs184234759 Chr.10: 100286216-100286216 on Build GRCh38
rs74900080 Chr.10: 100286267-100286267 on Build GRCh38
rs141949554 Chr.10: 100286463-100286464 on Build GRCh38
rs990563607 Chr.10: 100286295-100286295 on Build GRCh38
rs749957540 Chr.10: 100286098-100286098 on Build GRCh38
rs771047631 Chr.10: 100286147-100286147 on Build GRCh38
rs113283197 Chr.10: 100286481-100286481 on Build GRCh38
rs760060239 Chr.10: 100286234-100286234 on Build GRCh38
rs368757508 Chr.10: 100286149-100286149 on Build GRCh38
rs78762965 Chr.10: 100286266-100286266 on Build GRCh38
rs558294871 Chr.10: 100286104-100286104 on Build GRCh38
rs765881700 Chr.10: 100286068-100286069 on Build GRCh38
rs764447514 Chr.10: 100286219-100286219 on Build GRCh38
rs778850175 Chr.10: 100286249-100286249 on Build GRCh38
rs578038639 Chr.10: 100286471-100286471 on Build GRCh38
rs753179347 Chr.10: 100286067-100286067 on Build GRCh38
rs1030246289 Chr.10: 100286227-100286227 on Build GRCh38
rs763409177 Chr.10: 100286077-100286077 on Build GRCh38
rs1009258512 Chr.10: 100286349-100286349 on Build GRCh38
rs778905454 Chr.10: 100286121-100286121 on Build GRCh38
rs758227485 Chr.10: 100286257-100286257 on Build GRCh38
rs775938487 Chr.10: 100286476-100286476 on Build GRCh38
rs752495569 Chr.10: 100286250-100286250 on Build GRCh38
rs559742801 Chr.10: 100286086-100286086 on Build GRCh38
rs1018843690 Chr.10: 100286370-100286370 on Build GRCh38
rs770112434 Chr.10: 100286173-100286173 on Build GRCh38
rs376650396 Chr.10: 100286073-100286073 on Build GRCh38
rs752109622 Chr.10: 100286224-100286224 on Build GRCh38
rs994840944 Chr.10: 100286282-100286282 on Build GRCh38
rs766006769 Chr.10: 100286237-100286237 on Build GRCh38
rs748069503 Chr.10: 100286134-100286134 on Build GRCh38
rs200618064 Chr.10: 100286247-100286247 on Build GRCh38
rs144731392 Chr.10: 100286116-100286116 on Build GRCh38
rs777507289 Chr.10: 100286262-100286262 on Build GRCh38
rs759803168 Chr.10: 100286158-100286158 on Build GRCh38

View Full Product Details