Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 116340271-116340791 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CCDC172
Entrez Gene ID
374355
Gene Name
coiled-coil domain containing 172
Gene Aliases
C10orf96
UniGene
Hs.233407
Gene Chromosome Location
Chr.10: 116324097-116380029 on Build GRCh38
Transcripts
NM_198515.2 XM_006717820.3 XM_011539771.2 XM_017016206.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5521548 c.203T>G p.L68R Chr.10: 116340771-116340771 on Build GRCh38
5969672 c.211G>T p.A71S Chr.10: 116340779-116340779 on Build GRCh38
7299471 c.189C>T p.S63S Chr.10: 116340757-116340757 on Build GRCh38
1728509 c.180T>A p.F60L Chr.10: 116340748-116340748 on Build GRCh38
1474381 c.207G>C p.L69F Chr.10: 116340775-116340775 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs955042404 Chr.10: 116340393-116340393 on Build GRCh38
rs937701722 Chr.10: 116340758-116340758 on Build GRCh38
rs773084922 Chr.10: 116340716-116340716 on Build GRCh38
rs981898413 Chr.10: 116340709-116340709 on Build GRCh38
rs964729254 Chr.10: 116340291-116340291 on Build GRCh38
rs938477765 Chr.10: 116340573-116340573 on Build GRCh38
rs74891130 Chr.10: 116340396-116340396 on Build GRCh38
rs746631943 Chr.10: 116340764-116340764 on Build GRCh38
rs908083922 Chr.10: 116340431-116340431 on Build GRCh38
rs774412345 Chr.10: 116340688-116340688 on Build GRCh38
rs753826983 Chr.10: 116340721-116340721 on Build GRCh38
rs117513116 Chr.10: 116340756-116340756 on Build GRCh38
rs1019337617 Chr.10: 116340503-116340503 on Build GRCh38
rs964718172 Chr.10: 116340513-116340513 on Build GRCh38
rs760576448 Chr.10: 116340700-116340700 on Build GRCh38
rs1008984712 Chr.10: 116340497-116340497 on Build GRCh38
rs754900913 Chr.10: 116340723-116340723 on Build GRCh38
rs764859297 Chr.10: 116340736-116340736 on Build GRCh38
rs1026389390 Chr.10: 116340435-116340435 on Build GRCh38
rs1419870 Chr.10: 116340292-116340292 on Build GRCh38
rs886488975 Chr.10: 116340461-116340461 on Build GRCh38
rs188265199 Chr.10: 116340402-116340402 on Build GRCh38
rs928444221 Chr.10: 116340568-116340568 on Build GRCh38
rs560388003 Chr.10: 116340670-116340670 on Build GRCh38
rs1030789797 Chr.10: 116340755-116340755 on Build GRCh38
rs779626879 Chr.10: 116340743-116340743 on Build GRCh38
rs957851400 Chr.10: 116340563-116340563 on Build GRCh38
rs1028285545 Chr.10: 116340534-116340534 on Build GRCh38
rs137998964 Chr.10: 116340746-116340746 on Build GRCh38
rs545383672 Chr.10: 116340650-116340650 on Build GRCh38
rs752211066 Chr.10: 116340740-116340740 on Build GRCh38
rs996749892 Chr.10: 116340518-116340518 on Build GRCh38
rs373300112 Chr.10: 116340731-116340731 on Build GRCh38
rs532094148 Chr.10: 116340482-116340482 on Build GRCh38
rs919575660 Chr.10: 116340442-116340442 on Build GRCh38
rs766082125 Chr.10: 116340719-116340719 on Build GRCh38
rs74654779 Chr.10: 116340434-116340434 on Build GRCh38
rs748942710 Chr.10: 116340757-116340759 on Build GRCh38
rs1018903190 Chr.10: 116340734-116340734 on Build GRCh38
rs758641687 Chr.10: 116340309-116340309 on Build GRCh38
rs1055333621 Chr.10: 116340347-116340347 on Build GRCh38
rs772141410 Chr.10: 116340432-116340432 on Build GRCh38
rs1005057232 Chr.10: 116340484-116340484 on Build GRCh38
rs866153692 Chr.10: 116340646-116340646 on Build GRCh38
rs1052967440 Chr.10: 116340600-116340600 on Build GRCh38
rs989687022 Chr.10: 116340596-116340596 on Build GRCh38
rs180904496 Chr.10: 116340648-116340648 on Build GRCh38
rs1011518972 Chr.10: 116340658-116340660 on Build GRCh38
rs966840296 Chr.10: 116340678-116340678 on Build GRCh38

View Full Product Details