Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 119207571-119208109 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GRK5
Entrez Gene ID
2869
Gene Name
G protein-coupled receptor kinase 5
Gene Aliases
GPRK5
UniGene
Hs.524625
Gene Chromosome Location
Chr.10: 119207685-119455619 on Build GRCh38
Transcripts
NM_005308.2 XM_005269707.1 XM_005269708.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4012075 c.1A>G p.M1V Chr.10: 119207918-119207918 on Build GRCh38
6575772 c.52+5G>C p.? Chr.10: 119207974-119207974 on Build GRCh38
3985450 c.1A>T p.M1L Chr.10: 119207918-119207918 on Build GRCh38
4798692 c.22G>A p.A8T Chr.10: 119207939-119207939 on Build GRCh38
6575771 c.52+5G>C p.? Chr.10: 119207974-119207974 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs112389603 Chr.10: 119207893-119207893 on Build GRCh38
rs868848878 Chr.10: 119207873-119207873 on Build GRCh38
rs1017557633 Chr.10: 119207890-119207890 on Build GRCh38
rs553243953 Chr.10: 119207816-119207816 on Build GRCh38
rs949436788 Chr.10: 119207600-119207600 on Build GRCh38
rs1022698368 Chr.10: 119207613-119207613 on Build GRCh38
rs865934691 Chr.10: 119207646-119207646 on Build GRCh38
rs373449275 Chr.10: 119207991-119207991 on Build GRCh38
rs539094837 Chr.10: 119207811-119207811 on Build GRCh38
rs759503865 Chr.10: 119207979-119207979 on Build GRCh38
rs1033984801 Chr.10: 119208079-119208079 on Build GRCh38
rs910141340 Chr.10: 119207717-119207717 on Build GRCh38
rs200950380 Chr.10: 119208002-119208002 on Build GRCh38
rs961963890 Chr.10: 119207727-119207727 on Build GRCh38
rs369987575 Chr.10: 119207984-119207984 on Build GRCh38
rs756847219 Chr.10: 119207870-119207870 on Build GRCh38
rs763787681 Chr.10: 119207996-119207996 on Build GRCh38
rs2230347 Chr.10: 119207864-119207864 on Build GRCh38
rs749632512 Chr.10: 119208021-119208021 on Build GRCh38
rs869145481 Chr.10: 119207718-119207723 on Build GRCh38
rs781095348 Chr.10: 119207883-119207884 on Build GRCh38
rs765343773 Chr.10: 119207606-119207606 on Build GRCh38
rs376205003 Chr.10: 119207910-119207910 on Build GRCh38
rs932982926 Chr.10: 119207831-119207848 on Build GRCh38
rs994042276 Chr.10: 119208049-119208049 on Build GRCh38
rs761833081 Chr.10: 119207941-119207941 on Build GRCh38
rs113937734 Chr.10: 119207690-119207690 on Build GRCh38
rs985394764 Chr.10: 119207967-119207967 on Build GRCh38
rs984655667 Chr.10: 119207631-119207631 on Build GRCh38
rs780467929 Chr.10: 119208019-119208019 on Build GRCh38
rs933967702 Chr.10: 119207753-119207753 on Build GRCh38
rs781507167 Chr.10: 119207871-119207871 on Build GRCh38
rs1043316652 Chr.10: 119207810-119207810 on Build GRCh38
rs993912483 Chr.10: 119207812-119207812 on Build GRCh38
rs1053029416 Chr.10: 119207847-119207847 on Build GRCh38
rs869097446 Chr.10: 119207714-119207719 on Build GRCh38
rs889679733 Chr.10: 119207721-119207721 on Build GRCh38
rs536534805 Chr.10: 119207706-119207711 on Build GRCh38
rs776373760 Chr.10: 119207954-119207954 on Build GRCh38
rs895496985 Chr.10: 119207848-119207848 on Build GRCh38
rs888178611 Chr.10: 119208069-119208069 on Build GRCh38
rs1013904505 Chr.10: 119207851-119207851 on Build GRCh38
rs112475529 Chr.10: 119207597-119207597 on Build GRCh38
rs535634278 Chr.10: 119207857-119207857 on Build GRCh38
rs869232126 Chr.10: 119207721-119207726 on Build GRCh38
rs540763293 Chr.10: 119207857-119207857 on Build GRCh38
rs961546492 Chr.10: 119207891-119207891 on Build GRCh38
rs923156519 Chr.10: 119207681-119207681 on Build GRCh38
rs369362370 Chr.10: 119207879-119207879 on Build GRCh38
rs746493368 Chr.10: 119207867-119207867 on Build GRCh38
rs952467654 Chr.10: 119207817-119207817 on Build GRCh38
rs775913595 Chr.10: 119207892-119207892 on Build GRCh38
rs943215400 Chr.10: 119207807-119207807 on Build GRCh38
rs28399015 Chr.10: 119207708-119207708 on Build GRCh38
rs76294185 Chr.10: 119208057-119208057 on Build GRCh38
rs748884473 Chr.10: 119207706-119207708 on Build GRCh38
rs1006831663 Chr.10: 119207730-119207730 on Build GRCh38
rs936709811 Chr.10: 119207676-119207676 on Build GRCh38
rs573237429 Chr.10: 119207827-119207827 on Build GRCh38
rs955379106 Chr.10: 119207950-119207950 on Build GRCh38
rs1034448133 Chr.10: 119207839-119207850 on Build GRCh38
rs377433200 Chr.10: 119208003-119208003 on Build GRCh38
rs774606571 Chr.10: 119207918-119207918 on Build GRCh38
rs869084250 Chr.10: 119207720-119207725 on Build GRCh38
rs949707027 Chr.10: 119208050-119208050 on Build GRCh38
rs869192691 Chr.10: 119207717-119207722 on Build GRCh38
rs1054099940 Chr.10: 119207697-119207697 on Build GRCh38
rs575765965 Chr.10: 119207868-119207868 on Build GRCh38
rs1049820929 Chr.10: 119207761-119207761 on Build GRCh38
rs767615142 Chr.10: 119207947-119207947 on Build GRCh38
rs895696766 Chr.10: 119208035-119208035 on Build GRCh38
rs540663749 Chr.10: 119208018-119208018 on Build GRCh38
rs924078874 Chr.10: 119207989-119207989 on Build GRCh38
rs973004588 Chr.10: 119207735-119207735 on Build GRCh38
rs769999727 Chr.10: 119207883-119207883 on Build GRCh38
rs866808768 Chr.10: 119207596-119207596 on Build GRCh38
rs869214320 Chr.10: 119207715-119207720 on Build GRCh38
rs922614989 Chr.10: 119207739-119207739 on Build GRCh38
rs905522759 Chr.10: 119208075-119208075 on Build GRCh38
rs911230484 Chr.10: 119207992-119207992 on Build GRCh38
rs76422636 Chr.10: 119207729-119207734 on Build GRCh38
rs970232127 Chr.10: 119207616-119207616 on Build GRCh38
rs749963451 Chr.10: 119208010-119208010 on Build GRCh38
rs911268563 Chr.10: 119207805-119207805 on Build GRCh38

View Full Product Details