Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 123768425-123768923 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CPXM2
Entrez Gene ID
119587
Gene Name
carboxypeptidase X (M14 family), member 2
Gene Aliases
CPX2, UNQ676
UniGene
Hs.656887
Gene Chromosome Location
Chr.10: 123745636-123944094 on Build GRCh38
Transcripts
NM_198148.2 XM_005269528.3 XM_011539283.2 XM_011539285.2 XM_011539286.1 XM_017015673.1 XM_017015674.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1346541 c.1243C>T p.R415W Chr.10: 123768582-123768582 on Build GRCh38
2021096 c.1147G>C p.V383L Chr.10: 123768678-123768678 on Build GRCh38
4674517 c.1137C>T p.H379H Chr.10: 123768688-123768688 on Build GRCh38
340855 c.1147G>T p.V383L Chr.10: 123768678-123768678 on Build GRCh38
5359913 c.1204G>A p.A402T Chr.10: 123768621-123768621 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1047879737 Chr.10: 123768804-123768804 on Build GRCh38
rs758678722 Chr.10: 123768518-123768518 on Build GRCh38
rs996169764 Chr.10: 123768896-123768899 on Build GRCh38
rs900515442 Chr.10: 123768881-123768881 on Build GRCh38
rs144412439 Chr.10: 123768669-123768669 on Build GRCh38
rs745538812 Chr.10: 123768715-123768715 on Build GRCh38
rs570702833 Chr.10: 123768639-123768639 on Build GRCh38
rs772000176 Chr.10: 123768603-123768603 on Build GRCh38
rs370958461 Chr.10: 123768583-123768583 on Build GRCh38
rs745908668 Chr.10: 123768531-123768531 on Build GRCh38
rs550032711 Chr.10: 123768697-123768697 on Build GRCh38
rs762517969 Chr.10: 123768740-123768740 on Build GRCh38
rs756306654 Chr.10: 123768765-123768765 on Build GRCh38
rs532384672 Chr.10: 123768452-123768452 on Build GRCh38
rs539111465 Chr.10: 123768620-123768620 on Build GRCh38
rs746015077 Chr.10: 123768585-123768585 on Build GRCh38
rs762306663 Chr.10: 123768756-123768756 on Build GRCh38
rs778945977 Chr.10: 123768592-123768592 on Build GRCh38
rs774140430 Chr.10: 123768536-123768536 on Build GRCh38
rs140548347 Chr.10: 123768618-123768618 on Build GRCh38
rs965150946 Chr.10: 123768467-123768467 on Build GRCh38
rs150240288 Chr.10: 123768617-123768617 on Build GRCh38
rs752035323 Chr.10: 123768640-123768640 on Build GRCh38
rs569785003 Chr.10: 123768779-123768779 on Build GRCh38
rs966833897 Chr.10: 123768579-123768579 on Build GRCh38
rs774973935 Chr.10: 123768748-123768748 on Build GRCh38
rs753122950 Chr.10: 123768574-123768574 on Build GRCh38
rs868029784 Chr.10: 123768687-123768687 on Build GRCh38
rs771729647 Chr.10: 123768544-123768544 on Build GRCh38
rs376056113 Chr.10: 123768478-123768478 on Build GRCh38
rs199923458 Chr.10: 123768698-123768698 on Build GRCh38
rs755656874 Chr.10: 123768481-123768481 on Build GRCh38
rs866598999 Chr.10: 123768666-123768666 on Build GRCh38
rs1219727 Chr.10: 123768448-123768448 on Build GRCh38
rs200536913 Chr.10: 123768678-123768678 on Build GRCh38
rs1219724 Chr.10: 123768781-123768781 on Build GRCh38
rs1018900130 Chr.10: 123768649-123768657 on Build GRCh38
rs1038443099 Chr.10: 123768723-123768723 on Build GRCh38
rs757837701 Chr.10: 123768649-123768651 on Build GRCh38
rs143761278 Chr.10: 123768613-123768613 on Build GRCh38
rs780253314 Chr.10: 123768763-123768764 on Build GRCh38
rs572240858 Chr.10: 123768837-123768838 on Build GRCh38
rs771228059 Chr.10: 123768526-123768526 on Build GRCh38
rs966385686 Chr.10: 123768802-123768802 on Build GRCh38
rs568542152 Chr.10: 123768699-123768699 on Build GRCh38
rs151184826 Chr.10: 123768700-123768700 on Build GRCh38
rs199611205 Chr.10: 123768573-123768573 on Build GRCh38
rs576708220 Chr.10: 123768821-123768821 on Build GRCh38
rs201470381 Chr.10: 123768668-123768668 on Build GRCh38
rs748064476 Chr.10: 123768598-123768598 on Build GRCh38
rs1004306318 Chr.10: 123768457-123768457 on Build GRCh38
rs539700603 Chr.10: 123768792-123768792 on Build GRCh38
rs781558861 Chr.10: 123768584-123768584 on Build GRCh38
rs767194600 Chr.10: 123768633-123768633 on Build GRCh38
rs200730488 Chr.10: 123768483-123768483 on Build GRCh38
rs760398201 Chr.10: 123768609-123768609 on Build GRCh38
rs867376681 Chr.10: 123768673-123768673 on Build GRCh38
rs367761553 Chr.10: 123768552-123768552 on Build GRCh38
rs112510825 Chr.10: 123768831-123768831 on Build GRCh38
rs1219726 Chr.10: 123768491-123768491 on Build GRCh38
rs115623953 Chr.10: 123768778-123768778 on Build GRCh38
rs139425659 Chr.10: 123768581-123768581 on Build GRCh38
rs778302866 Chr.10: 123768582-123768582 on Build GRCh38
rs1032082687 Chr.10: 123768844-123768844 on Build GRCh38
rs764754854 Chr.10: 123768743-123768743 on Build GRCh38
rs770692756 Chr.10: 123768709-123768709 on Build GRCh38
rs149261159 Chr.10: 123768535-123768535 on Build GRCh38
rs1017148733 Chr.10: 123768474-123768474 on Build GRCh38
rs112482384 Chr.10: 123768458-123768458 on Build GRCh38
rs562235853 Chr.10: 123768525-123768525 on Build GRCh38
rs753174527 Chr.10: 123768512-123768512 on Build GRCh38
rs773241060 Chr.10: 123768820-123768820 on Build GRCh38
rs758966246 Chr.10: 123768561-123768561 on Build GRCh38
rs1219725 Chr.10: 123768532-123768532 on Build GRCh38
rs375547613 Chr.10: 123768630-123768630 on Build GRCh38
rs376782848 Chr.10: 123768757-123768757 on Build GRCh38
rs370103790 Chr.10: 123768484-123768484 on Build GRCh38
rs959149434 Chr.10: 123768877-123768877 on Build GRCh38
rs202065646 Chr.10: 123768683-123768683 on Build GRCh38
rs779224061 Chr.10: 123768492-123768492 on Build GRCh38
rs778190214 Chr.10: 123768521-123768521 on Build GRCh38
rs752230154 Chr.10: 123768657-123768658 on Build GRCh38
rs116463741 Chr.10: 123768793-123768793 on Build GRCh38
rs769737772 Chr.10: 123768591-123768591 on Build GRCh38
rs761713000 Chr.10: 123768543-123768543 on Build GRCh38
rs773008676 Chr.10: 123768604-123768604 on Build GRCh38
rs368011035 Chr.10: 123768736-123768736 on Build GRCh38
rs780023934 Chr.10: 123768693-123768693 on Build GRCh38
rs746866224 Chr.10: 123768707-123768707 on Build GRCh38
rs756316209 Chr.10: 123768688-123768688 on Build GRCh38
rs752234963 Chr.10: 123768538-123768540 on Build GRCh38
rs772955437 Chr.10: 123768546-123768546 on Build GRCh38
rs750747933 Chr.10: 123768758-123768758 on Build GRCh38
rs978689560 Chr.10: 123768499-123768499 on Build GRCh38
rs17106006 Chr.10: 123768553-123768553 on Build GRCh38
rs776387177 Chr.10: 123768714-123768714 on Build GRCh38
rs368099662 Chr.10: 123768767-123768767 on Build GRCh38
rs756569879 Chr.10: 123768693-123768694 on Build GRCh38

View Full Product Details