Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 125997277-125997784 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FANK1
Entrez Gene ID
92565
Gene Name
fibronectin type III and ankyrin repeat domains 1
Gene Aliases
HSD13
UniGene
Hs.352591
Gene Chromosome Location
Chr.10: 125896539-126009599 on Build GRCh38
Transcripts
NM_145235.3 XM_005270280.3 XM_006718058.3 XM_006718064.3 XM_011540348.2 XM_011540349.2 XM_011540350.2 XM_011540351.2 XM_011540356.2 XM_011540357.2 XM_011540358.2 XM_011540360.2 XM_011540361.2 XM_011540362.2 XM_011540363.2 XM_017016916.1 XM_017016917.1 XM_017016918.1 XM_017016919.1 XM_017016920.1 XM_017016921.1 XM_017016922.1 XM_017016923.1 XM_017016924.1 XM_017016925.1 XM_017016926.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3375418 c.507C>T p.D169D Chr.10: 125997453-125997453 on Build GRCh38
5623766 c.494C>G p.S165C Chr.10: 125997440-125997440 on Build GRCh38
3435486 c.495T>C p.S165S Chr.10: 125997441-125997441 on Build GRCh38
684107 c.474G>A p.R158R Chr.10: 125997420-125997420 on Build GRCh38
5787249 c.491T>C p.V164A Chr.10: 125997437-125997437 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs889325752 Chr.10: 125997476-125997476 on Build GRCh38
rs144982939 Chr.10: 125997748-125997748 on Build GRCh38
rs1048845310 Chr.10: 125997656-125997657 on Build GRCh38
rs1036386382 Chr.10: 125997431-125997431 on Build GRCh38
rs755121753 Chr.10: 125997673-125997673 on Build GRCh38
rs776774927 Chr.10: 125997536-125997536 on Build GRCh38
rs751561229 Chr.10: 125997417-125997417 on Build GRCh38
rs185380267 Chr.10: 125997689-125997689 on Build GRCh38
rs140634850 Chr.10: 125997418-125997418 on Build GRCh38
rs375041809 Chr.10: 125997412-125997412 on Build GRCh38
rs545600195 Chr.10: 125997750-125997750 on Build GRCh38
rs371045332 Chr.10: 125997400-125997400 on Build GRCh38
rs765311231 Chr.10: 125997708-125997708 on Build GRCh38
rs1040194180 Chr.10: 125997693-125997693 on Build GRCh38
rs746980836 Chr.10: 125997456-125997457 on Build GRCh38
rs957420512 Chr.10: 125997361-125997361 on Build GRCh38
rs542737827 Chr.10: 125997454-125997454 on Build GRCh38
rs766493086 Chr.10: 125997420-125997420 on Build GRCh38
rs368797806 Chr.10: 125997362-125997362 on Build GRCh38
rs770711488 Chr.10: 125997489-125997489 on Build GRCh38
rs747568682 Chr.10: 125997474-125997474 on Build GRCh38
rs771851803 Chr.10: 125997386-125997387 on Build GRCh38
rs61863577 Chr.10: 125997307-125997307 on Build GRCh38
rs930364904 Chr.10: 125997386-125997386 on Build GRCh38
rs202212822 Chr.10: 125997491-125997491 on Build GRCh38
rs368033192 Chr.10: 125997512-125997512 on Build GRCh38
rs374729861 Chr.10: 125997422-125997422 on Build GRCh38
rs948281787 Chr.10: 125997557-125997557 on Build GRCh38
rs776404787 Chr.10: 125997390-125997390 on Build GRCh38
rs578198216 Chr.10: 125997342-125997342 on Build GRCh38
rs371515160 Chr.10: 125997534-125997534 on Build GRCh38
rs138551736 Chr.10: 125997432-125997432 on Build GRCh38
rs754087885 Chr.10: 125997612-125997612 on Build GRCh38
rs943543763 Chr.10: 125997752-125997752 on Build GRCh38
rs1031091861 Chr.10: 125997751-125997751 on Build GRCh38
rs1031831485 Chr.10: 125997535-125997535 on Build GRCh38
rs990351772 Chr.10: 125997580-125997580 on Build GRCh38
rs368490061 Chr.10: 125997453-125997453 on Build GRCh38
rs777746099 Chr.10: 125997403-125997404 on Build GRCh38
rs758758335 Chr.10: 125997462-125997462 on Build GRCh38
rs756782038 Chr.10: 125997484-125997484 on Build GRCh38
rs752844178 Chr.10: 125997749-125997749 on Build GRCh38
rs990035193 Chr.10: 125997365-125997365 on Build GRCh38
rs770606485 Chr.10: 125997373-125997373 on Build GRCh38
rs778394380 Chr.10: 125997468-125997468 on Build GRCh38
rs374080105 Chr.10: 125997497-125997497 on Build GRCh38
rs957184315 Chr.10: 125997573-125997573 on Build GRCh38
rs903879764 Chr.10: 125997743-125997743 on Build GRCh38
rs927981554 Chr.10: 125997414-125997414 on Build GRCh38
rs956965356 Chr.10: 125997760-125997760 on Build GRCh38
rs188419621 Chr.10: 125997427-125997427 on Build GRCh38
rs759260456 Chr.10: 125997391-125997391 on Build GRCh38
rs745447545 Chr.10: 125997494-125997494 on Build GRCh38
rs774249318 Chr.10: 125997527-125997527 on Build GRCh38
rs769438389 Chr.10: 125997396-125997396 on Build GRCh38
rs761761458 Chr.10: 125997532-125997532 on Build GRCh38
rs949728277 Chr.10: 125997346-125997346 on Build GRCh38
rs928579257 Chr.10: 125997618-125997618 on Build GRCh38
rs116419360 Chr.10: 125997655-125997655 on Build GRCh38
rs972072292 Chr.10: 125997677-125997677 on Build GRCh38
rs73372506 Chr.10: 125997324-125997324 on Build GRCh38
rs746513886 Chr.10: 125997371-125997371 on Build GRCh38
rs1040638276 Chr.10: 125997759-125997759 on Build GRCh38
rs996067156 Chr.10: 125997713-125997713 on Build GRCh38
rs371125128 Chr.10: 125997653-125997653 on Build GRCh38
rs981342307 Chr.10: 125997615-125997615 on Build GRCh38
rs1007820926 Chr.10: 125997486-125997486 on Build GRCh38
rs768476080 Chr.10: 125997525-125997525 on Build GRCh38
rs919332478 Chr.10: 125997690-125997690 on Build GRCh38
rs764003777 Chr.10: 125997413-125997413 on Build GRCh38
rs1043998965 Chr.10: 125997359-125997359 on Build GRCh38
rs1022805698 Chr.10: 125997585-125997585 on Build GRCh38
rs915812157 Chr.10: 125997382-125997382 on Build GRCh38
rs886300217 Chr.10: 125997399-125997399 on Build GRCh38
rs12411612 Chr.10: 125997444-125997444 on Build GRCh38
rs930745486 Chr.10: 125997703-125997703 on Build GRCh38
rs1050320977 Chr.10: 125997740-125997740 on Build GRCh38
rs749102875 Chr.10: 125997523-125997523 on Build GRCh38

View Full Product Details