Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 617355-617880 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CDHR5
Entrez Gene ID
53841
Gene Name
cadherin related family member 5
Gene Aliases
MLPCDH, MU-PCDH, MUCDHL, MUPCDH
UniGene
Hs.165619
Gene Chromosome Location
Chr.11: 616565-625067 on Build GRCh38
Transcripts
NM_001171968.1 NM_021924.4 NM_031264.3 XM_006718253.3 XM_011520188.2 XM_011520189.2 XM_011520190.2 XM_011520191.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1200528 c.2345G>A p.R782Q Chr.11: 617544-617544 on Build GRCh38
4536222 c.2278G>A p.E760K Chr.11: 617611-617611 on Build GRCh38
6625274 c.2397G>A p.T799T Chr.11: 617492-617492 on Build GRCh38
6396938 c.2356G>T p.G786C Chr.11: 617533-617533 on Build GRCh38
1247904 c.2316G>A p.T772T Chr.11: 617573-617573 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs368300364 Chr.11: 617573-617573 on Build GRCh38
rs755486369 Chr.11: 617631-617631 on Build GRCh38
rs747852490 Chr.11: 617596-617596 on Build GRCh38
rs764911557 Chr.11: 617432-617432 on Build GRCh38
rs905116432 Chr.11: 617845-617845 on Build GRCh38
rs370576784 Chr.11: 617584-617584 on Build GRCh38
rs748198900 Chr.11: 617417-617417 on Build GRCh38
rs745861045 Chr.11: 617663-617663 on Build GRCh38
rs751683517 Chr.11: 617543-617543 on Build GRCh38
rs137998609 Chr.11: 617593-617593 on Build GRCh38
rs781732884 Chr.11: 617517-617517 on Build GRCh38
rs143800672 Chr.11: 617383-617383 on Build GRCh38
rs750236957 Chr.11: 617492-617492 on Build GRCh38
rs183406300 Chr.11: 617708-617708 on Build GRCh38
rs758111098 Chr.11: 617639-617639 on Build GRCh38
rs756124038 Chr.11: 617771-617771 on Build GRCh38
rs1055838699 Chr.11: 617810-617810 on Build GRCh38
rs747080771 Chr.11: 617508-617508 on Build GRCh38
rs977802678 Chr.11: 617659-617659 on Build GRCh38
rs2740380 Chr.11: 617537-617537 on Build GRCh38
rs780506088 Chr.11: 617527-617527 on Build GRCh38
rs758449664 Chr.11: 617408-617408 on Build GRCh38
rs372636570 Chr.11: 617791-617791 on Build GRCh38
rs957715577 Chr.11: 617770-617770 on Build GRCh38
rs752923592 Chr.11: 617504-617504 on Build GRCh38
rs34889521 Chr.11: 617519-617519 on Build GRCh38
rs550217560 Chr.11: 617507-617507 on Build GRCh38
rs761007488 Chr.11: 617401-617401 on Build GRCh38
rs764278187 Chr.11: 617542-617542 on Build GRCh38
rs12385783 Chr.11: 617780-617780 on Build GRCh38
rs371766448 Chr.11: 617556-617556 on Build GRCh38
rs776997889 Chr.11: 617425-617425 on Build GRCh38
rs745736724 Chr.11: 617603-617603 on Build GRCh38
rs759177014 Chr.11: 617431-617431 on Build GRCh38
rs768476530 Chr.11: 617551-617551 on Build GRCh38
rs186854698 Chr.11: 617375-617375 on Build GRCh38
rs755383441 Chr.11: 617612-617613 on Build GRCh38
rs764668042 Chr.11: 617406-617406 on Build GRCh38
rs780603503 Chr.11: 617571-617571 on Build GRCh38
rs772540298 Chr.11: 617567-617567 on Build GRCh38
rs199786415 Chr.11: 617704-617704 on Build GRCh38
rs773235186 Chr.11: 617534-617534 on Build GRCh38
rs909901636 Chr.11: 617688-617688 on Build GRCh38
rs139083373 Chr.11: 617465-617465 on Build GRCh38
rs369213863 Chr.11: 617785-617785 on Build GRCh38
rs369218232 Chr.11: 617483-617483 on Build GRCh38
rs758506352 Chr.11: 617466-617466 on Build GRCh38
rs754404388 Chr.11: 617628-617628 on Build GRCh38
rs770002127 Chr.11: 617481-617481 on Build GRCh38
rs753083717 Chr.11: 617491-617491 on Build GRCh38
rs773980723 Chr.11: 617489-617489 on Build GRCh38
rs779084236 Chr.11: 617595-617595 on Build GRCh38
rs753466360 Chr.11: 617403-617403 on Build GRCh38
rs896111006 Chr.11: 617396-617396 on Build GRCh38
rs762160066 Chr.11: 617613-617613 on Build GRCh38
rs571665990 Chr.11: 617784-617784 on Build GRCh38
rs756616027 Chr.11: 617471-617473 on Build GRCh38
rs150617987 Chr.11: 617456-617456 on Build GRCh38
rs756764471 Chr.11: 617656-617656 on Build GRCh38
rs375729796 Chr.11: 617414-617414 on Build GRCh38
rs757031773 Chr.11: 617410-617410 on Build GRCh38
rs149934780 Chr.11: 617493-617493 on Build GRCh38
rs142038877 Chr.11: 617538-617538 on Build GRCh38
rs146893811 Chr.11: 617406-617407 on Build GRCh38
rs774047706 Chr.11: 617530-617530 on Build GRCh38
rs147490212 Chr.11: 617468-617468 on Build GRCh38
rs199885960 Chr.11: 617740-617740 on Build GRCh38
rs746133763 Chr.11: 617687-617687 on Build GRCh38
rs139645120 Chr.11: 617477-617477 on Build GRCh38
rs749483684 Chr.11: 617379-617379 on Build GRCh38
rs564520780 Chr.11: 617585-617585 on Build GRCh38
rs777311849 Chr.11: 617647-617647 on Build GRCh38
rs199739008 Chr.11: 617452-617452 on Build GRCh38
rs922755312 Chr.11: 617685-617685 on Build GRCh38
rs1030685973 Chr.11: 617761-617761 on Build GRCh38
rs774427670 Chr.11: 617384-617384 on Build GRCh38
rs1052145423 Chr.11: 617859-617859 on Build GRCh38
rs146458539 Chr.11: 617574-617574 on Build GRCh38
rs749635188 Chr.11: 617549-617549 on Build GRCh38
rs1050571021 Chr.11: 617786-617786 on Build GRCh38
rs761367679 Chr.11: 617531-617531 on Build GRCh38
rs763654741 Chr.11: 617443-617443 on Build GRCh38
rs575687624 Chr.11: 617673-617673 on Build GRCh38
rs776458803 Chr.11: 617572-617572 on Build GRCh38
rs771274750 Chr.11: 617818-617818 on Build GRCh38
rs773602374 Chr.11: 617675-617675 on Build GRCh38
rs771617655 Chr.11: 617559-617559 on Build GRCh38
rs549263574 Chr.11: 617721-617721 on Build GRCh38
rs761727654 Chr.11: 617490-617490 on Build GRCh38
rs773610074 Chr.11: 617400-617400 on Build GRCh38
rs775392594 Chr.11: 617672-617672 on Build GRCh38
rs746221782 Chr.11: 617479-617479 on Build GRCh38
rs930073262 Chr.11: 617686-617686 on Build GRCh38
rs146662733 Chr.11: 617541-617541 on Build GRCh38
rs982524956 Chr.11: 617710-617710 on Build GRCh38
rs578096953 Chr.11: 617520-617520 on Build GRCh38
rs766177413 Chr.11: 617539-617539 on Build GRCh38
rs767600057 Chr.11: 617620-617620 on Build GRCh38
rs765572781 Chr.11: 617453-617453 on Build GRCh38
rs186765155 Chr.11: 617796-617796 on Build GRCh38
rs757156059 Chr.11: 617446-617446 on Build GRCh38
rs757975661 Chr.11: 617597-617597 on Build GRCh38
rs909932733 Chr.11: 617802-617802 on Build GRCh38
rs780628525 Chr.11: 617548-617548 on Build GRCh38
rs552855718 Chr.11: 617855-617855 on Build GRCh38
rs746623032 Chr.11: 617648-617648 on Build GRCh38
rs752250033 Chr.11: 617407-617407 on Build GRCh38
rs141531457 Chr.11: 617444-617444 on Build GRCh38
rs775131015 Chr.11: 617608-617608 on Build GRCh38
rs111246420 Chr.11: 617854-617854 on Build GRCh38
rs760663910 Chr.11: 617569-617569 on Build GRCh38
rs1057304926 Chr.11: 617390-617390 on Build GRCh38
rs759162239 Chr.11: 617405-617405 on Build GRCh38
rs771456343 Chr.11: 617476-617476 on Build GRCh38
rs143109433 Chr.11: 617467-617467 on Build GRCh38
rs773732726 Chr.11: 617421-617421 on Build GRCh38
rs757387099 Chr.11: 617544-617544 on Build GRCh38
rs777409245 Chr.11: 617598-617598 on Build GRCh38
rs144034443 Chr.11: 617571-617571 on Build GRCh38
rs866839898 Chr.11: 617723-617723 on Build GRCh38
rs568251906 Chr.11: 617607-617607 on Build GRCh38
rs768415400 Chr.11: 617529-617529 on Build GRCh38
rs909042304 Chr.11: 617399-617399 on Build GRCh38
rs374607923 Chr.11: 617590-617590 on Build GRCh38
rs1046282436 Chr.11: 617853-617853 on Build GRCh38
rs373271397 Chr.11: 617413-617413 on Build GRCh38
rs573809635 Chr.11: 617844-617844 on Build GRCh38
rs982877206 Chr.11: 617797-617797 on Build GRCh38
rs747718637 Chr.11: 617557-617557 on Build GRCh38
rs749638785 Chr.11: 617583-617584 on Build GRCh38
rs374458706 Chr.11: 617423-617423 on Build GRCh38
rs781324004 Chr.11: 617638-617638 on Build GRCh38
rs778930946 Chr.11: 617415-617415 on Build GRCh38
rs150870939 Chr.11: 617570-617570 on Build GRCh38
rs779161504 Chr.11: 617639-617639 on Build GRCh38
rs775311797 Chr.11: 617482-617482 on Build GRCh38
rs201360349 Chr.11: 617738-617738 on Build GRCh38
rs534793497 Chr.11: 617741-617741 on Build GRCh38
rs760918157 Chr.11: 617690-617690 on Build GRCh38
rs768452448 Chr.11: 617674-617674 on Build GRCh38
rs771437191 Chr.11: 617533-617533 on Build GRCh38
rs370991978 Chr.11: 617540-617540 on Build GRCh38
rs750886772 Chr.11: 617431-617432 on Build GRCh38
rs371913447 Chr.11: 617506-617506 on Build GRCh38
rs754352880 Chr.11: 617592-617592 on Build GRCh38
rs922787873 Chr.11: 617790-617790 on Build GRCh38
rs757752942 Chr.11: 617514-617514 on Build GRCh38
rs908269069 Chr.11: 617727-617727 on Build GRCh38
rs749003240 Chr.11: 617528-617528 on Build GRCh38
rs200173856 Chr.11: 617437-617437 on Build GRCh38
rs780622120 Chr.11: 617657-617657 on Build GRCh38
rs770378107 Chr.11: 617602-617602 on Build GRCh38
rs763708001 Chr.11: 617485-617485 on Build GRCh38
rs769610439 Chr.11: 617664-617664 on Build GRCh38
rs61877857 Chr.11: 617420-617420 on Build GRCh38
rs375605005 Chr.11: 617709-617709 on Build GRCh38
rs758719553 Chr.11: 617715-617715 on Build GRCh38
rs779874989 Chr.11: 617778-617778 on Build GRCh38
rs756723845 Chr.11: 617589-617589 on Build GRCh38
rs768342805 Chr.11: 617779-617779 on Build GRCh38
rs111307862 Chr.11: 617833-617833 on Build GRCh38
rs976909646 Chr.11: 617676-617676 on Build GRCh38
rs138995984 Chr.11: 617388-617388 on Build GRCh38
rs990159228 Chr.11: 617669-617669 on Build GRCh38
rs767270922 Chr.11: 617448-617448 on Build GRCh38
rs766123915 Chr.11: 617498-617498 on Build GRCh38
rs899903005 Chr.11: 617801-617801 on Build GRCh38
rs1053920660 Chr.11: 617422-617422 on Build GRCh38
rs765691308 Chr.11: 617634-617634 on Build GRCh38
rs374367073 Chr.11: 617545-617545 on Build GRCh38
rs570930849 Chr.11: 617441-617441 on Build GRCh38
rs114976644 Chr.11: 617705-617705 on Build GRCh38
rs201483927 Chr.11: 617627-617627 on Build GRCh38
rs941360953 Chr.11: 617808-617808 on Build GRCh38
rs773471613 Chr.11: 617621-617621 on Build GRCh38
rs941009228 Chr.11: 617601-617601 on Build GRCh38
rs766718914 Chr.11: 617402-617402 on Build GRCh38
rs984582048 Chr.11: 617398-617398 on Build GRCh38
rs77535085 Chr.11: 617409-617409 on Build GRCh38
rs756889133 Chr.11: 617739-617739 on Build GRCh38
rs560704453 Chr.11: 617691-617691 on Build GRCh38
rs1038030628 Chr.11: 617696-617696 on Build GRCh38
rs756673532 Chr.11: 617546-617546 on Build GRCh38
rs528202606 Chr.11: 617411-617411 on Build GRCh38
rs746492354 Chr.11: 617600-617600 on Build GRCh38
rs61877856 Chr.11: 617378-617378 on Build GRCh38

View Full Product Details