Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 6234466-6234972 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CNGA4
Entrez Gene ID
1262
Gene Name
cyclic nucleotide gated channel alpha 4
Gene Aliases
CNCA2, CNG-4, CNG4, CNG5, CNGB2, OCNC2, OCNCBETA, OCNCb
UniGene
Hs.434618
Gene Chromosome Location
Chr.11: 6234774-6245924 on Build GRCh38
Transcripts
XM_017017218.1 XM_017017219.1 NM_001098794.1 NM_032127.3 XM_006718343.2 XM_011520398.2 XM_011520399.2

Gene Symbol
FAM160A2
Entrez Gene ID
84067
Gene Name
family with sequence similarity 160 member A2
Gene Aliases
C11orf56
UniGene
Hs.501793
Gene Chromosome Location
Chr.11: 6211334-6234711 on Build GRCh38
Transcripts
XM_017017218.1 XM_017017219.1 NM_001098794.1 NM_032127.3 XM_006718343.2 XM_011520398.2 XM_011520399.2

SNP Detail

SNP ID Chromosome Location
rs987368379 Chr.11: 6234666-6234666 on Build GRCh38
rs1022425486 Chr.11: 6234699-6234699 on Build GRCh38
rs1008866340 Chr.11: 6234650-6234665 on Build GRCh38
rs1009602725 Chr.11: 6234862-6234862 on Build GRCh38
rs189888044 Chr.11: 6234810-6234810 on Build GRCh38
rs575394913 Chr.11: 6234730-6234730 on Build GRCh38
rs946433960 Chr.11: 6234704-6234704 on Build GRCh38
rs545582336 Chr.11: 6234753-6234753 on Build GRCh38
rs535018965 Chr.11: 6234593-6234593 on Build GRCh38
rs988785432 Chr.11: 6234634-6234634 on Build GRCh38
rs770524229 Chr.11: 6234675-6234675 on Build GRCh38
rs757999882 Chr.11: 6234566-6234566 on Build GRCh38
rs535418062 Chr.11: 6234693-6234693 on Build GRCh38
rs761129413 Chr.11: 6234631-6234631 on Build GRCh38
rs973612299 Chr.11: 6234563-6234563 on Build GRCh38
rs368101082 Chr.11: 6234697-6234697 on Build GRCh38
rs561495222 Chr.11: 6234781-6234781 on Build GRCh38
rs1034959016 Chr.11: 6234533-6234533 on Build GRCh38
rs943040013 Chr.11: 6234580-6234580 on Build GRCh38
rs1008769749 Chr.11: 6234865-6234865 on Build GRCh38
rs968612193 Chr.11: 6234702-6234702 on Build GRCh38
rs1017783828 Chr.11: 6234925-6234925 on Build GRCh38
rs905148655 Chr.11: 6234656-6234656 on Build GRCh38
rs1046856155 Chr.11: 6234639-6234639 on Build GRCh38
rs929184808 Chr.11: 6234569-6234569 on Build GRCh38
rs976357945 Chr.11: 6234716-6234716 on Build GRCh38
rs943040937 Chr.11: 6234691-6234691 on Build GRCh38
rs893580472 Chr.11: 6234591-6234591 on Build GRCh38
rs956749552 Chr.11: 6234686-6234686 on Build GRCh38
rs1003721028 Chr.11: 6234616-6234616 on Build GRCh38
rs371350555 Chr.11: 6234673-6234673 on Build GRCh38
rs895130194 Chr.11: 6234625-6234625 on Build GRCh38
rs1012698403 Chr.11: 6234627-6234627 on Build GRCh38
rs570189523 Chr.11: 6234511-6234511 on Build GRCh38
rs80264592 Chr.11: 6234708-6234708 on Build GRCh38
rs144353621 Chr.11: 6234574-6234574 on Build GRCh38
rs1047259454 Chr.11: 6234756-6234756 on Build GRCh38
rs1056636958 Chr.11: 6234751-6234751 on Build GRCh38
rs905098701 Chr.11: 6234814-6234814 on Build GRCh38
rs532360390 Chr.11: 6234680-6234680 on Build GRCh38
rs888279502 Chr.11: 6234644-6234644 on Build GRCh38
rs34007206 Chr.11: 6234565-6234566 on Build GRCh38
rs974779954 Chr.11: 6234695-6234695 on Build GRCh38
rs1056457382 Chr.11: 6234816-6234816 on Build GRCh38
rs572558232 Chr.11: 6234763-6234763 on Build GRCh38
rs548255379 Chr.11: 6234517-6234517 on Build GRCh38
rs959377678 Chr.11: 6234536-6234536 on Build GRCh38
rs325700 Chr.11: 6234584-6234584 on Build GRCh38
rs554908041 Chr.11: 6234615-6234615 on Build GRCh38
rs987399731 Chr.11: 6234572-6234572 on Build GRCh38
rs966293810 Chr.11: 6234709-6234709 on Build GRCh38
rs1026123538 Chr.11: 6234653-6234653 on Build GRCh38
rs950560656 Chr.11: 6234661-6234661 on Build GRCh38
rs12274456 Chr.11: 6234754-6234754 on Build GRCh38
rs531169879 Chr.11: 6234849-6234849 on Build GRCh38
rs939543310 Chr.11: 6234740-6234740 on Build GRCh38
rs963238261 Chr.11: 6234562-6234562 on Build GRCh38
rs181396922 Chr.11: 6234840-6234840 on Build GRCh38
rs766796807 Chr.11: 6234632-6234632 on Build GRCh38
rs186154323 Chr.11: 6234848-6234848 on Build GRCh38
rs990904099 Chr.11: 6234540-6234540 on Build GRCh38
rs1016946324 Chr.11: 6234633-6234633 on Build GRCh38
rs976305736 Chr.11: 6234946-6234946 on Build GRCh38
rs1003476555 Chr.11: 6234815-6234815 on Build GRCh38
rs915411001 Chr.11: 6234579-6234579 on Build GRCh38
rs966242812 Chr.11: 6234943-6234943 on Build GRCh38
rs139785949 Chr.11: 6234491-6234491 on Build GRCh38
rs895370163 Chr.11: 6234752-6234752 on Build GRCh38
rs563701870 Chr.11: 6234619-6234619 on Build GRCh38
rs1042910770 Chr.11: 6234598-6234598 on Build GRCh38
rs544148784 Chr.11: 6234604-6234604 on Build GRCh38
rs1023966670 Chr.11: 6234655-6234655 on Build GRCh38
rs968181996 Chr.11: 6234907-6234907 on Build GRCh38
rs902706763 Chr.11: 6234612-6234612 on Build GRCh38
rs941189236 Chr.11: 6234798-6234798 on Build GRCh38
rs531111549 Chr.11: 6234555-6234555 on Build GRCh38
rs553373245 Chr.11: 6234620-6234620 on Build GRCh38
rs754240554 Chr.11: 6234539-6234539 on Build GRCh38
rs568359605 Chr.11: 6234668-6234668 on Build GRCh38
rs972861863 Chr.11: 6234643-6234643 on Build GRCh38
rs1003752364 Chr.11: 6234499-6234499 on Build GRCh38
rs950528451 Chr.11: 6234861-6234861 on Build GRCh38

View Full Product Details