Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 34051576-34052069 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CAPRIN1
Entrez Gene ID
4076
Gene Name
cell cycle associated protein 1
Gene Aliases
GPIAP1, GPIP137, GRIP137, M11S1, RNG105, p137GPI
UniGene
Hs.471818
Gene Chromosome Location
Chr.11: 34051617-34102610 on Build GRCh38
Transcripts
NM_005898.4 NM_203364.2

SNP Detail

SNP ID Chromosome Location
rs1011628736 Chr.11: 34051961-34051961 on Build GRCh38
rs978753326 Chr.11: 34051904-34051904 on Build GRCh38
rs925976871 Chr.11: 34051908-34051908 on Build GRCh38
rs965910447 Chr.11: 34051655-34051655 on Build GRCh38
rs865827144 Chr.11: 34051681-34051681 on Build GRCh38
rs894911375 Chr.11: 34051737-34051737 on Build GRCh38
rs981456624 Chr.11: 34052013-34052013 on Build GRCh38
rs967340455 Chr.11: 34051973-34051973 on Build GRCh38
rs1020200455 Chr.11: 34051848-34051848 on Build GRCh38
rs912020477 Chr.11: 34051959-34051959 on Build GRCh38
rs992714263 Chr.11: 34051846-34051846 on Build GRCh38
rs946271929 Chr.11: 34051692-34051692 on Build GRCh38
rs930933744 Chr.11: 34051978-34051978 on Build GRCh38
rs913232442 Chr.11: 34051814-34051814 on Build GRCh38
rs983368824 Chr.11: 34051642-34051642 on Build GRCh38
rs888124208 Chr.11: 34051983-34051983 on Build GRCh38
rs898686439 Chr.11: 34051859-34051859 on Build GRCh38
rs890698386 Chr.11: 34051640-34051640 on Build GRCh38
rs886147434 Chr.11: 34051765-34051765 on Build GRCh38
rs960195817 Chr.11: 34051688-34051688 on Build GRCh38
rs764008256 Chr.11: 34051808-34051808 on Build GRCh38
rs1048800982 Chr.11: 34051878-34051878 on Build GRCh38
rs1036587355 Chr.11: 34051793-34051793 on Build GRCh38
rs1048018417 Chr.11: 34051979-34051979 on Build GRCh38
rs1034559008 Chr.11: 34051833-34051833 on Build GRCh38
rs946032959 Chr.11: 34051815-34051815 on Build GRCh38
rs552069117 Chr.11: 34051851-34051851 on Build GRCh38
rs1020388435 Chr.11: 34051964-34051964 on Build GRCh38
rs565892244 Chr.11: 34051902-34051902 on Build GRCh38
rs1020166349 Chr.11: 34052033-34052033 on Build GRCh38
rs907325925 Chr.11: 34051708-34051708 on Build GRCh38
rs1026860571 Chr.11: 34052005-34052005 on Build GRCh38
rs148293074 Chr.11: 34051673-34051673 on Build GRCh38
rs930394411 Chr.11: 34051877-34051877 on Build GRCh38
rs568022324 Chr.11: 34052029-34052029 on Build GRCh38
rs950616204 Chr.11: 34051637-34051637 on Build GRCh38
rs996526755 Chr.11: 34051937-34051937 on Build GRCh38
rs967279725 Chr.11: 34052046-34052046 on Build GRCh38
rs561712473 Chr.11: 34051679-34051679 on Build GRCh38
rs957470975 Chr.11: 34051942-34051942 on Build GRCh38
rs932122876 Chr.11: 34051773-34051773 on Build GRCh38
rs974622233 Chr.11: 34051785-34051785 on Build GRCh38
rs1027271076 Chr.11: 34051758-34051758 on Build GRCh38
rs995017487 Chr.11: 34051738-34051738 on Build GRCh38
rs1034115169 Chr.11: 34051999-34051999 on Build GRCh38
rs994523596 Chr.11: 34051604-34051604 on Build GRCh38
rs542539224 Chr.11: 34051615-34051615 on Build GRCh38
rs996734848 Chr.11: 34051599-34051599 on Build GRCh38
rs966243193 Chr.11: 34051784-34051784 on Build GRCh38
rs993851499 Chr.11: 34051668-34051668 on Build GRCh38
rs530823205 Chr.11: 34051690-34051690 on Build GRCh38
rs528344302 Chr.11: 34051919-34051919 on Build GRCh38
rs961495217 Chr.11: 34051984-34051984 on Build GRCh38
rs928913850 Chr.11: 34051819-34051819 on Build GRCh38
rs959262088 Chr.11: 34051602-34051602 on Build GRCh38
rs879565140 Chr.11: 34051778-34051778 on Build GRCh38
rs143034568 Chr.11: 34051718-34051718 on Build GRCh38
rs975201478 Chr.11: 34051669-34051669 on Build GRCh38
rs1055252029 Chr.11: 34051736-34051736 on Build GRCh38
rs374389217 Chr.11: 34051685-34051685 on Build GRCh38
rs753759502 Chr.11: 34051600-34051601 on Build GRCh38
rs760387329 Chr.11: 34051756-34051756 on Build GRCh38
rs961048301 Chr.11: 34051667-34051667 on Build GRCh38
rs922052701 Chr.11: 34051671-34051671 on Build GRCh38
rs1026650379 Chr.11: 34051809-34051809 on Build GRCh38
rs1001669457 Chr.11: 34051990-34051990 on Build GRCh38
rs564236166 Chr.11: 34051781-34051781 on Build GRCh38
rs1035558663 Chr.11: 34051974-34051974 on Build GRCh38
rs895582029 Chr.11: 34052021-34052021 on Build GRCh38
rs1028819459 Chr.11: 34051941-34051941 on Build GRCh38
rs981656110 Chr.11: 34051817-34051817 on Build GRCh38
rs937480341 Chr.11: 34051835-34051835 on Build GRCh38
rs990132541 Chr.11: 34051683-34051683 on Build GRCh38
rs1046052708 Chr.11: 34051701-34051701 on Build GRCh38
rs1019254374 Chr.11: 34051921-34051921 on Build GRCh38
rs1055947988 Chr.11: 34051836-34051836 on Build GRCh38
rs531558916 Chr.11: 34051888-34051888 on Build GRCh38
rs7950888 Chr.11: 34051813-34051813 on Build GRCh38
rs994085532 Chr.11: 34051989-34051989 on Build GRCh38
rs1018732692 Chr.11: 34051649-34051649 on Build GRCh38
rs926008071 Chr.11: 34051756-34051756 on Build GRCh38
rs7951306 Chr.11: 34051925-34051925 on Build GRCh38
rs946042826 Chr.11: 34051732-34051732 on Build GRCh38
rs1029266729 Chr.11: 34051807-34051807 on Build GRCh38

View Full Product Details