Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 44048264-44048764 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ACCSL
Entrez Gene ID
390110
Gene Name
1-aminocyclopropane-1-carboxylate synthase homolog (inactive) like
Gene Aliases
-
UniGene
Hs.558851
Gene Chromosome Location
Chr.11: 43920987-44060667 on Build GRCh38
Transcripts
NM_001031854.2 XM_017017714.1 XM_017017715.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6132861 c.314T>C p.V105A Chr.11: 44048350-44048350 on Build GRCh38
5177283 c.504+5G>T p.? Chr.11: 44048545-44048545 on Build GRCh38
1604541 c.500C>A p.T167N Chr.11: 44048536-44048536 on Build GRCh38
688156 c.398G>C p.R133P Chr.11: 44048434-44048434 on Build GRCh38
378632 c.415G>T p.G139W Chr.11: 44048451-44048451 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs766259014 Chr.11: 44048303-44048303 on Build GRCh38
rs372992418 Chr.11: 44048410-44048410 on Build GRCh38
rs770394976 Chr.11: 44048557-44048557 on Build GRCh38
rs60488147 Chr.11: 44048708-44048708 on Build GRCh38
rs745567296 Chr.11: 44048561-44048561 on Build GRCh38
rs541794514 Chr.11: 44048645-44048645 on Build GRCh38
rs907569857 Chr.11: 44048738-44048738 on Build GRCh38
rs781502418 Chr.11: 44048435-44048435 on Build GRCh38
rs376261378 Chr.11: 44048433-44048433 on Build GRCh38
rs540474011 Chr.11: 44048717-44048717 on Build GRCh38
rs955849340 Chr.11: 44048704-44048704 on Build GRCh38
rs760702457 Chr.11: 44048489-44048489 on Build GRCh38
rs943101711 Chr.11: 44048365-44048365 on Build GRCh38
rs760509714 Chr.11: 44048563-44048563 on Build GRCh38
rs751767223 Chr.11: 44048520-44048520 on Build GRCh38
rs980200096 Chr.11: 44048734-44048734 on Build GRCh38
rs566938729 Chr.11: 44048320-44048320 on Build GRCh38
rs78719262 Chr.11: 44048554-44048554 on Build GRCh38
rs548419052 Chr.11: 44048307-44048307 on Build GRCh38
rs776956310 Chr.11: 44048344-44048344 on Build GRCh38
rs771831713 Chr.11: 44048480-44048480 on Build GRCh38
rs774199441 Chr.11: 44048381-44048381 on Build GRCh38
rs771412568 Chr.11: 44048559-44048562 on Build GRCh38
rs766463284 Chr.11: 44048504-44048504 on Build GRCh38
rs759197314 Chr.11: 44048479-44048479 on Build GRCh38
rs748718263 Chr.11: 44048436-44048436 on Build GRCh38
rs769239667 Chr.11: 44048342-44048342 on Build GRCh38
rs769330956 Chr.11: 44048357-44048357 on Build GRCh38
rs186883674 Chr.11: 44048296-44048296 on Build GRCh38
rs948663061 Chr.11: 44048715-44048715 on Build GRCh38
rs770267749 Chr.11: 44048444-44048444 on Build GRCh38
rs773825632 Chr.11: 44048474-44048474 on Build GRCh38
rs778336343 Chr.11: 44048357-44048377 on Build GRCh38
rs201522650 Chr.11: 44048289-44048289 on Build GRCh38
rs750566975 Chr.11: 44048494-44048494 on Build GRCh38
rs77451436 Chr.11: 44048333-44048333 on Build GRCh38
rs370597673 Chr.11: 44048568-44048568 on Build GRCh38
rs755221260 Chr.11: 44048525-44048525 on Build GRCh38
rs752724012 Chr.11: 44048403-44048403 on Build GRCh38
rs978132695 Chr.11: 44048648-44048648 on Build GRCh38
rs1038753403 Chr.11: 44048395-44048395 on Build GRCh38
rs920324163 Chr.11: 44048706-44048706 on Build GRCh38
rs867581126 Chr.11: 44048710-44048710 on Build GRCh38
rs765889809 Chr.11: 44048376-44048376 on Build GRCh38
rs369930593 Chr.11: 44048392-44048392 on Build GRCh38
rs1047956030 Chr.11: 44048517-44048517 on Build GRCh38
rs369721305 Chr.11: 44048318-44048318 on Build GRCh38
rs968538350 Chr.11: 44048613-44048613 on Build GRCh38
rs1031478828 Chr.11: 44048652-44048652 on Build GRCh38
rs574456946 Chr.11: 44048577-44048577 on Build GRCh38
rs900506293 Chr.11: 44048600-44048600 on Build GRCh38
rs748630136 Chr.11: 44048355-44048355 on Build GRCh38
rs772701076 Chr.11: 44048367-44048367 on Build GRCh38
rs766433400 Chr.11: 44048566-44048566 on Build GRCh38
rs376209867 Chr.11: 44048449-44048449 on Build GRCh38
rs770646737 Chr.11: 44048461-44048461 on Build GRCh38
rs775412043 Chr.11: 44048481-44048481 on Build GRCh38
rs548571995 Chr.11: 44048325-44048325 on Build GRCh38
rs965049904 Chr.11: 44048310-44048310 on Build GRCh38
rs767655979 Chr.11: 44048578-44048578 on Build GRCh38
rs550573443 Chr.11: 44048294-44048294 on Build GRCh38
rs1031040299 Chr.11: 44048629-44048629 on Build GRCh38
rs372956008 Chr.11: 44048448-44048448 on Build GRCh38
rs747538398 Chr.11: 44048556-44048556 on Build GRCh38
rs780102897 Chr.11: 44048418-44048418 on Build GRCh38
rs1003789922 Chr.11: 44048514-44048514 on Build GRCh38
rs201604433 Chr.11: 44048334-44048334 on Build GRCh38
rs762559172 Chr.11: 44048297-44048297 on Build GRCh38
rs756617656 Chr.11: 44048543-44048543 on Build GRCh38
rs974985045 Chr.11: 44048690-44048690 on Build GRCh38
rs761302315 Chr.11: 44048287-44048287 on Build GRCh38
rs1044060577 Chr.11: 44048475-44048475 on Build GRCh38
rs781590490 Chr.11: 44048535-44048535 on Build GRCh38
rs762648064 Chr.11: 44048370-44048370 on Build GRCh38
rs750473048 Chr.11: 44048405-44048405 on Build GRCh38
rs762895022 Chr.11: 44048502-44048502 on Build GRCh38
rs371000393 Chr.11: 44048295-44048295 on Build GRCh38
rs964555191 Chr.11: 44048660-44048660 on Build GRCh38
rs911827369 Chr.11: 44048655-44048655 on Build GRCh38
rs777713778 Chr.11: 44048343-44048343 on Build GRCh38
rs776761709 Chr.11: 44048565-44048565 on Build GRCh38
rs747424625 Chr.11: 44048432-44048432 on Build GRCh38
rs999496403 Chr.11: 44048621-44048621 on Build GRCh38
rs1016887613 Chr.11: 44048599-44048599 on Build GRCh38
rs775430363 Chr.11: 44048562-44048562 on Build GRCh38
rs556208891 Chr.11: 44048552-44048552 on Build GRCh38
rs767511611 Chr.11: 44048402-44048402 on Build GRCh38
rs911753915 Chr.11: 44048364-44048364 on Build GRCh38
rs768740007 Chr.11: 44048564-44048564 on Build GRCh38
rs190169018 Chr.11: 44048434-44048434 on Build GRCh38
rs1047382968 Chr.11: 44048713-44048713 on Build GRCh38
rs199731719 Chr.11: 44048312-44048312 on Build GRCh38
rs955924413 Chr.11: 44048636-44048636 on Build GRCh38
rs764060093 Chr.11: 44048490-44048490 on Build GRCh38
rs758655681 Chr.11: 44048413-44048413 on Build GRCh38
rs752997629 Chr.11: 44048541-44048541 on Build GRCh38
rs765472708 Chr.11: 44048404-44048404 on Build GRCh38
rs560032745 Chr.11: 44048654-44048654 on Build GRCh38
rs577211880 Chr.11: 44048358-44048358 on Build GRCh38
rs776213936 Chr.11: 44048293-44048293 on Build GRCh38

View Full Product Details