Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 57742025-57742520 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C11orf31
Entrez Gene ID
280636
Gene Name
chromosome 11 open reading frame 31
Gene Aliases
C17orf10, SELH
UniGene
Hs.655373
Gene Chromosome Location
Chr.11: 57741250-57743554 on Build GRCh38
Transcripts
NM_001321335.1 NM_170746.3 NR_037646.1

Gene Symbol
TMX2-CTNND1
Entrez Gene ID
100528016
Gene Name
TMX2-CTNND1 readthrough (NMD candidate)
Gene Aliases
-
UniGene
Hs.166011
Gene Chromosome Location
Chr.11: 57712523-57819180 on Build GRCh38
Transcripts
NM_001321335.1 NM_170746.3 NR_037646.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7133241 c.351G>C p.L117F Chr.11: 57742199-57742199 on Build GRCh38
1317295 c.279C>T p.L93L Chr.11: 57742127-57742127 on Build GRCh38
7134406 c.315C>G p.L105L Chr.11: 57742163-57742163 on Build GRCh38
1741243 c.323C>G p.P108R Chr.11: 57742171-57742171 on Build GRCh38
6313917 c.269-1G>T p.? Chr.11: 57742116-57742116 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs549289169 Chr.11: 57742469-57742469 on Build GRCh38
rs374990252 Chr.11: 57742165-57742165 on Build GRCh38
rs916136296 Chr.11: 57742059-57742059 on Build GRCh38
rs763474985 Chr.11: 57742078-57742078 on Build GRCh38
rs779825709 Chr.11: 57742257-57742257 on Build GRCh38
rs771021142 Chr.11: 57742097-57742097 on Build GRCh38
rs549110995 Chr.11: 57742457-57742457 on Build GRCh38
rs760021571 Chr.11: 57742345-57742345 on Build GRCh38
rs201621399 Chr.11: 57742127-57742127 on Build GRCh38
rs370024227 Chr.11: 57742213-57742213 on Build GRCh38
rs767638486 Chr.11: 57742148-57742148 on Build GRCh38
rs746484242 Chr.11: 57742184-57742184 on Build GRCh38
rs371704385 Chr.11: 57742161-57742161 on Build GRCh38
rs996356482 Chr.11: 57742406-57742406 on Build GRCh38
rs752659016 Chr.11: 57742149-57742149 on Build GRCh38
rs751630667 Chr.11: 57742080-57742080 on Build GRCh38
rs768337262 Chr.11: 57742185-57742185 on Build GRCh38
rs756336275 Chr.11: 57742084-57742084 on Build GRCh38
rs956209527 Chr.11: 57742331-57742331 on Build GRCh38
rs752958130 Chr.11: 57742083-57742083 on Build GRCh38
rs981833644 Chr.11: 57742456-57742456 on Build GRCh38
rs200643822 Chr.11: 57742179-57742179 on Build GRCh38
rs971674268 Chr.11: 57742319-57742319 on Build GRCh38
rs1041824471 Chr.11: 57742291-57742291 on Build GRCh38
rs971396111 Chr.11: 57742060-57742060 on Build GRCh38
rs749040104 Chr.11: 57742092-57742092 on Build GRCh38
rs902091918 Chr.11: 57742355-57742355 on Build GRCh38
rs750138667 Chr.11: 57742245-57742245 on Build GRCh38
rs752093324 Chr.11: 57742132-57742132 on Build GRCh38
rs1040653789 Chr.11: 57742347-57742347 on Build GRCh38
rs1029136558 Chr.11: 57742413-57742419 on Build GRCh38
rs754813448 Chr.11: 57742069-57742069 on Build GRCh38
rs763391302 Chr.11: 57742463-57742463 on Build GRCh38
rs1041399191 Chr.11: 57742051-57742051 on Build GRCh38
rs757490141 Chr.11: 57742237-57742237 on Build GRCh38
rs753722142 Chr.11: 57742155-57742155 on Build GRCh38
rs760801933 Chr.11: 57742220-57742220 on Build GRCh38
rs750730948 Chr.11: 57742074-57742074 on Build GRCh38
rs947121501 Chr.11: 57742286-57742286 on Build GRCh38
rs958628773 Chr.11: 57742481-57742481 on Build GRCh38
rs771609631 Chr.11: 57742117-57742117 on Build GRCh38
rs1013055950 Chr.11: 57742307-57742307 on Build GRCh38
rs1023225382 Chr.11: 57742316-57742316 on Build GRCh38
rs981677238 Chr.11: 57742323-57742323 on Build GRCh38
rs762154867 Chr.11: 57742071-57742071 on Build GRCh38
rs146047458 Chr.11: 57742314-57742314 on Build GRCh38
rs778734578 Chr.11: 57742072-57742072 on Build GRCh38
rs754112113 Chr.11: 57742224-57742224 on Build GRCh38
rs756424319 Chr.11: 57742153-57742153 on Build GRCh38
rs778097988 Chr.11: 57742154-57742154 on Build GRCh38
rs563062355 Chr.11: 57742126-57742126 on Build GRCh38
rs562038698 Chr.11: 57742055-57742055 on Build GRCh38
rs544488180 Chr.11: 57742107-57742107 on Build GRCh38
rs1047422188 Chr.11: 57742216-57742216 on Build GRCh38
rs902882291 Chr.11: 57742294-57742294 on Build GRCh38
rs780607862 Chr.11: 57742064-57742064 on Build GRCh38
rs747931318 Chr.11: 57742120-57742120 on Build GRCh38
rs1048089326 Chr.11: 57742492-57742492 on Build GRCh38
rs1006314838 Chr.11: 57742439-57742439 on Build GRCh38
rs962923647 Chr.11: 57742285-57742285 on Build GRCh38
rs943700320 Chr.11: 57742302-57742302 on Build GRCh38
rs923102127 Chr.11: 57742484-57742484 on Build GRCh38
rs190189307 Chr.11: 57742202-57742202 on Build GRCh38
rs887908126 Chr.11: 57742432-57742432 on Build GRCh38
rs985258442 Chr.11: 57742163-57742163 on Build GRCh38
rs766920051 Chr.11: 57742130-57742130 on Build GRCh38
rs771676224 Chr.11: 57742067-57742067 on Build GRCh38
rs1016402741 Chr.11: 57742282-57742282 on Build GRCh38
rs778890993 Chr.11: 57742105-57742105 on Build GRCh38
rs745825356 Chr.11: 57742163-57742163 on Build GRCh38
rs766832316 Chr.11: 57742079-57742079 on Build GRCh38
rs761998886 Chr.11: 57742190-57742190 on Build GRCh38
rs757108374 Chr.11: 57742159-57742159 on Build GRCh38
rs17152073 Chr.11: 57742218-57742218 on Build GRCh38
rs765837860 Chr.11: 57742428-57742428 on Build GRCh38
rs760390496 Chr.11: 57742121-57742121 on Build GRCh38
rs1031694405 Chr.11: 57742330-57742330 on Build GRCh38
rs376111197 Chr.11: 57742206-57742206 on Build GRCh38
rs529341659 Chr.11: 57742073-57742073 on Build GRCh38
rs962110547 Chr.11: 57742453-57742453 on Build GRCh38
rs991757354 Chr.11: 57742056-57742056 on Build GRCh38
rs991363993 Chr.11: 57742491-57742491 on Build GRCh38
rs373869913 Chr.11: 57742214-57742214 on Build GRCh38
rs933270161 Chr.11: 57742487-57742487 on Build GRCh38
rs755063730 Chr.11: 57742081-57742081 on Build GRCh38
rs767623062 Chr.11: 57742199-57742201 on Build GRCh38
rs777208663 Chr.11: 57742276-57742276 on Build GRCh38
rs746572071 Chr.11: 57742264-57742264 on Build GRCh38
rs758393721 Chr.11: 57742248-57742248 on Build GRCh38
rs759598701 Chr.11: 57742145-57742145 on Build GRCh38
rs776039549 Chr.11: 57742444-57742444 on Build GRCh38

View Full Product Details