Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 63818710-63819205 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C11orf84
Entrez Gene ID
144097
Gene Name
chromosome 11 open reading frame 84
Gene Aliases
-
UniGene
Hs.502793
Gene Chromosome Location
Chr.11: 63813374-63827718 on Build GRCh38
Transcripts
NM_138471.2 XM_005273782.2 XM_005273783.2 XM_006718437.1 XM_011544770.1 XM_011544771.1 XM_011544772.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
467184 c.919G>A p.E307K Chr.11: 63818987-63818987 on Build GRCh38
7298114 c.841C>T p.H281Y Chr.11: 63818909-63818909 on Build GRCh38
5373168 c.822C>T p.F274F Chr.11: 63818890-63818890 on Build GRCh38
7300914 c.800A>G p.D267G Chr.11: 63818868-63818868 on Build GRCh38
4645786 c.818G>A p.G273D Chr.11: 63818886-63818886 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs544912861 Chr.11: 63818746-63818746 on Build GRCh38
rs774543419 Chr.11: 63818837-63818837 on Build GRCh38
rs770861041 Chr.11: 63819021-63819021 on Build GRCh38
rs762037545 Chr.11: 63818847-63818847 on Build GRCh38
rs370050900 Chr.11: 63818884-63818884 on Build GRCh38
rs867324895 Chr.11: 63818978-63818979 on Build GRCh38
rs553290378 Chr.11: 63818964-63818964 on Build GRCh38
rs767745888 Chr.11: 63818932-63818932 on Build GRCh38
rs766769553 Chr.11: 63818860-63818860 on Build GRCh38
rs1052867312 Chr.11: 63818750-63818750 on Build GRCh38
rs982518411 Chr.11: 63819103-63819103 on Build GRCh38
rs925016019 Chr.11: 63819099-63819116 on Build GRCh38
rs780255272 Chr.11: 63818778-63818779 on Build GRCh38
rs907245171 Chr.11: 63819095-63819095 on Build GRCh38
rs201086880 Chr.11: 63818827-63818827 on Build GRCh38
rs769395077 Chr.11: 63818891-63818891 on Build GRCh38
rs777776749 Chr.11: 63818945-63818945 on Build GRCh38
rs899354258 Chr.11: 63819121-63819121 on Build GRCh38
rs761019870 Chr.11: 63819019-63819019 on Build GRCh38
rs1046269069 Chr.11: 63819061-63819061 on Build GRCh38
rs774669515 Chr.11: 63818988-63818989 on Build GRCh38
rs929458850 Chr.11: 63819087-63819087 on Build GRCh38
rs564187812 Chr.11: 63819066-63819067 on Build GRCh38
rs199846422 Chr.11: 63818984-63818984 on Build GRCh38
rs972224361 Chr.11: 63818939-63818939 on Build GRCh38
rs557608335 Chr.11: 63818849-63818849 on Build GRCh38
rs755297783 Chr.11: 63818870-63818870 on Build GRCh38
rs768380262 Chr.11: 63818921-63818921 on Build GRCh38
rs769118722 Chr.11: 63818807-63818807 on Build GRCh38
rs372129782 Chr.11: 63818912-63818912 on Build GRCh38
rs779481076 Chr.11: 63818881-63818881 on Build GRCh38
rs932965803 Chr.11: 63818794-63818794 on Build GRCh38
rs1023363206 Chr.11: 63819149-63819149 on Build GRCh38
rs989435424 Chr.11: 63819013-63819013 on Build GRCh38
rs756592176 Chr.11: 63818856-63818856 on Build GRCh38
rs749561211 Chr.11: 63818815-63818816 on Build GRCh38
rs781308402 Chr.11: 63818788-63818788 on Build GRCh38
rs753145068 Chr.11: 63818968-63818968 on Build GRCh38
rs770549168 Chr.11: 63818820-63818820 on Build GRCh38
rs368084206 Chr.11: 63818786-63818786 on Build GRCh38
rs750874077 Chr.11: 63818789-63818789 on Build GRCh38
rs532939537 Chr.11: 63818811-63818811 on Build GRCh38
rs943602944 Chr.11: 63819064-63819064 on Build GRCh38
rs756586473 Chr.11: 63818791-63818791 on Build GRCh38
rs371799487 Chr.11: 63818960-63818960 on Build GRCh38
rs768609496 Chr.11: 63818983-63818983 on Build GRCh38
rs1056193389 Chr.11: 63819139-63819139 on Build GRCh38
rs981374257 Chr.11: 63818921-63818922 on Build GRCh38
rs748255165 Chr.11: 63819036-63819062 on Build GRCh38
rs758498190 Chr.11: 63818892-63818892 on Build GRCh38
rs189841854 Chr.11: 63819000-63819000 on Build GRCh38
rs146412650 Chr.11: 63818822-63818822 on Build GRCh38
rs376766268 Chr.11: 63819039-63819039 on Build GRCh38
rs779389588 Chr.11: 63819049-63819049 on Build GRCh38
rs568698696 Chr.11: 63818836-63818836 on Build GRCh38
rs780810675 Chr.11: 63818890-63818890 on Build GRCh38
rs574860842 Chr.11: 63818946-63818946 on Build GRCh38
rs780429410 Chr.11: 63818793-63818793 on Build GRCh38
rs760636626 Chr.11: 63818950-63818950 on Build GRCh38
rs757325870 Chr.11: 63818787-63818787 on Build GRCh38
rs533857629 Chr.11: 63818937-63818937 on Build GRCh38
rs78113693 Chr.11: 63818795-63818795 on Build GRCh38
rs537681016 Chr.11: 63818848-63818848 on Build GRCh38
rs894507126 Chr.11: 63819154-63819154 on Build GRCh38
rs144000790 Chr.11: 63818821-63818821 on Build GRCh38
rs902132991 Chr.11: 63819083-63819083 on Build GRCh38
rs754291128 Chr.11: 63818956-63818956 on Build GRCh38
rs142087991 Chr.11: 63818992-63818992 on Build GRCh38
rs1022972421 Chr.11: 63818882-63818882 on Build GRCh38
rs780970208 Chr.11: 63818973-63818973 on Build GRCh38
rs908292595 Chr.11: 63819107-63819107 on Build GRCh38
rs149061077 Chr.11: 63818917-63818917 on Build GRCh38
rs753306617 Chr.11: 63819036-63819036 on Build GRCh38
rs1016449614 Chr.11: 63819176-63819176 on Build GRCh38
rs544850529 Chr.11: 63819030-63819030 on Build GRCh38
rs376557647 Chr.11: 63818986-63818986 on Build GRCh38
rs533643543 Chr.11: 63819155-63819155 on Build GRCh38
rs938817133 Chr.11: 63819113-63819113 on Build GRCh38
rs375494902 Chr.11: 63818985-63818985 on Build GRCh38
rs544816090 Chr.11: 63818775-63818775 on Build GRCh38
rs866084492 Chr.11: 63819112-63819112 on Build GRCh38
rs755794215 Chr.11: 63819048-63819048 on Build GRCh38
rs763194319 Chr.11: 63818767-63818767 on Build GRCh38
rs370894205 Chr.11: 63818982-63818982 on Build GRCh38
rs756976251 Chr.11: 63818885-63818885 on Build GRCh38
rs776779023 Chr.11: 63819024-63819024 on Build GRCh38
rs202015322 Chr.11: 63818981-63818981 on Build GRCh38
rs529071042 Chr.11: 63818740-63818740 on Build GRCh38
rs745574265 Chr.11: 63818978-63818978 on Build GRCh38
rs756922742 Chr.11: 63818969-63818969 on Build GRCh38
rs201645866 Chr.11: 63818812-63818812 on Build GRCh38
rs769035932 Chr.11: 63818909-63818910 on Build GRCh38
rs766551295 Chr.11: 63818952-63818952 on Build GRCh38
rs887825187 Chr.11: 63819099-63819107 on Build GRCh38
rs1035585565 Chr.11: 63819178-63819178 on Build GRCh38
rs761980411 Chr.11: 63818925-63818925 on Build GRCh38
rs754413369 Chr.11: 63818869-63818869 on Build GRCh38
rs1034984261 Chr.11: 63818739-63818739 on Build GRCh38
rs762633038 Chr.11: 63818830-63818830 on Build GRCh38
rs73492262 Chr.11: 63819040-63819040 on Build GRCh38
rs955979300 Chr.11: 63819179-63819179 on Build GRCh38
rs945160607 Chr.11: 63819052-63819052 on Build GRCh38
rs769649340 Chr.11: 63818826-63818826 on Build GRCh38
rs1038262062 Chr.11: 63819116-63819116 on Build GRCh38

View Full Product Details