Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 26421113-26421521 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DRC1
Entrez Gene ID
92749
Gene Name
dynein regulatory complex subunit 1
Gene Aliases
C2orf39, CCDC164, CILD21
UniGene
Hs.393714
Gene Chromosome Location
Chr.2: 26401912-26456713 on Build GRCh38
Transcripts
NM_145038.4 XM_005264637.3 XM_005264638.3 XM_017005271.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
2912386 c.352C>T p.Q118* Chr.2: 26421396-26421396 on Build GRCh38
5509791 c.274G>A p.E92K Chr.2: 26421318-26421318 on Build GRCh38
4093330 c.347A>C p.K116T Chr.2: 26421391-26421391 on Build GRCh38
2912383 c.266G>T p.C89F Chr.2: 26421310-26421310 on Build GRCh38
3391652 c.324G>A p.R108R Chr.2: 26421368-26421368 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs988805295 Chr.2: 26421232-26421232 on Build GRCh38
rs763436176 Chr.2: 26421341-26421341 on Build GRCh38
rs927441613 Chr.2: 26421296-26421296 on Build GRCh38
rs752358195 Chr.2: 26421362-26421362 on Build GRCh38
rs200567817 Chr.2: 26421394-26421394 on Build GRCh38
rs765588210 Chr.2: 26421405-26421405 on Build GRCh38
rs769795298 Chr.2: 26421319-26421319 on Build GRCh38
rs369587363 Chr.2: 26421317-26421317 on Build GRCh38
rs753766730 Chr.2: 26421254-26421254 on Build GRCh38
rs747358703 Chr.2: 26421387-26421388 on Build GRCh38
rs111807831 Chr.2: 26421485-26421485 on Build GRCh38
rs745718864 Chr.2: 26421312-26421312 on Build GRCh38
rs777457305 Chr.2: 26421252-26421252 on Build GRCh38
rs755637691 Chr.2: 26421251-26421251 on Build GRCh38
rs1036668603 Chr.2: 26421392-26421392 on Build GRCh38
rs556219697 Chr.2: 26421166-26421166 on Build GRCh38
rs878889754 Chr.2: 26421455-26421455 on Build GRCh38
rs142371860 Chr.2: 26421396-26421396 on Build GRCh38
rs758085776 Chr.2: 26421393-26421393 on Build GRCh38
rs968925020 Chr.2: 26421238-26421238 on Build GRCh38
rs767947875 Chr.2: 26421358-26421358 on Build GRCh38
rs374892627 Chr.2: 26421257-26421257 on Build GRCh38
rs750393829 Chr.2: 26421387-26421387 on Build GRCh38
rs577266124 Chr.2: 26421203-26421203 on Build GRCh38
rs371616390 Chr.2: 26421375-26421375 on Build GRCh38
rs748397280 Chr.2: 26421318-26421318 on Build GRCh38
rs780462851 Chr.2: 26421354-26421355 on Build GRCh38
rs753459451 Chr.2: 26421371-26421371 on Build GRCh38
rs1019641286 Chr.2: 26421221-26421221 on Build GRCh38
rs746418475 Chr.2: 26421445-26421445 on Build GRCh38
rs537895608 Chr.2: 26421139-26421139 on Build GRCh38
rs750845149 Chr.2: 26421491-26421491 on Build GRCh38
rs774673149 Chr.2: 26421444-26421444 on Build GRCh38
rs1028107492 Chr.2: 26421175-26421175 on Build GRCh38
rs889603879 Chr.2: 26421480-26421480 on Build GRCh38
rs993039506 Chr.2: 26421463-26421463 on Build GRCh38
rs763644509 Chr.2: 26421370-26421370 on Build GRCh38
rs774744171 Chr.2: 26421352-26421352 on Build GRCh38
rs760409243 Chr.2: 26421363-26421363 on Build GRCh38
rs138488150 Chr.2: 26421366-26421366 on Build GRCh38
rs538825721 Chr.2: 26421174-26421174 on Build GRCh38
rs146060843 Chr.2: 26421374-26421374 on Build GRCh38
rs199583175 Chr.2: 26421449-26421449 on Build GRCh38
rs913520496 Chr.2: 26421477-26421477 on Build GRCh38
rs759868480 Chr.2: 26421357-26421357 on Build GRCh38
rs914810750 Chr.2: 26421235-26421235 on Build GRCh38
rs771406695 Chr.2: 26421345-26421345 on Build GRCh38
rs773292881 Chr.2: 26421320-26421320 on Build GRCh38
rs775188209 Chr.2: 26421205-26421205 on Build GRCh38
rs980299015 Chr.2: 26421290-26421290 on Build GRCh38
rs373304971 Chr.2: 26421448-26421448 on Build GRCh38
rs572607941 Chr.2: 26421314-26421314 on Build GRCh38
rs554368476 Chr.2: 26421266-26421266 on Build GRCh38
rs561605921 Chr.2: 26421400-26421400 on Build GRCh38
rs749365742 Chr.2: 26421408-26421408 on Build GRCh38
rs578075025 Chr.2: 26421169-26421169 on Build GRCh38
rs956248765 Chr.2: 26421226-26421226 on Build GRCh38

View Full Product Details