Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 112257468-112258035 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AP1AR
Entrez Gene ID
55435
Gene Name
adaptor related protein complex 1 associated regulatory protein
Gene Aliases
2C18, C4orf16, GBAR, PRO0971, gamma-BAR
UniGene
Hs.435991
Gene Chromosome Location
Chr.4: 112231727-112273110 on Build GRCh38
Transcripts
NM_001128426.2 NM_018569.5 NR_136622.1 XM_017008378.1 XM_017008379.1 XM_017008380.1

SNP Detail

SNP ID Chromosome Location
rs895083743 Chr.4: 112257531-112257531 on Build GRCh38
rs369515291 Chr.4: 112257810-112257811 on Build GRCh38
rs895037089 Chr.4: 112257965-112257965 on Build GRCh38
rs79605872 Chr.4: 112257955-112257957 on Build GRCh38
rs745680647 Chr.4: 112257820-112257823 on Build GRCh38
rs930346672 Chr.4: 112257894-112257894 on Build GRCh38
rs147158310 Chr.4: 112257739-112257743 on Build GRCh38
rs550017583 Chr.4: 112257881-112257881 on Build GRCh38
rs1013066772 Chr.4: 112257590-112257590 on Build GRCh38
rs934286049 Chr.4: 112257574-112257574 on Build GRCh38
rs752665184 Chr.4: 112257770-112257770 on Build GRCh38
rs767806247 Chr.4: 112257761-112257761 on Build GRCh38
rs548289317 Chr.4: 112257648-112257648 on Build GRCh38
rs529605015 Chr.4: 112257855-112257855 on Build GRCh38
rs1018517589 Chr.4: 112257829-112257829 on Build GRCh38
rs550067920 Chr.4: 112257692-112257695 on Build GRCh38
rs934545432 Chr.4: 112257956-112257956 on Build GRCh38
rs939016712 Chr.4: 112257945-112257945 on Build GRCh38
rs796239937 Chr.4: 112257938-112257938 on Build GRCh38
rs530938165 Chr.4: 112257822-112257822 on Build GRCh38
rs747667130 Chr.4: 112257774-112257774 on Build GRCh38
rs777771068 Chr.4: 112257811-112257811 on Build GRCh38
rs1035203509 Chr.4: 112257710-112257710 on Build GRCh38
rs775472543 Chr.4: 112257831-112257831 on Build GRCh38
rs760804449 Chr.4: 112257838-112257838 on Build GRCh38
rs33918954 Chr.4: 112257742-112257746 on Build GRCh38
rs989858759 Chr.4: 112257993-112257993 on Build GRCh38
rs547174670 Chr.4: 112257823-112257823 on Build GRCh38
rs768235858 Chr.4: 112257736-112257736 on Build GRCh38
rs903916940 Chr.4: 112257704-112257704 on Build GRCh38
rs530190896 Chr.4: 112257520-112257520 on Build GRCh38
rs779574970 Chr.4: 112257732-112257732 on Build GRCh38
rs756224592 Chr.4: 112257808-112257809 on Build GRCh38
rs892908929 Chr.4: 112257587-112257588 on Build GRCh38
rs76129513 Chr.4: 112257953-112257953 on Build GRCh38
rs1040312751 Chr.4: 112257526-112257526 on Build GRCh38
rs759416887 Chr.4: 112257828-112257828 on Build GRCh38
rs905364863 Chr.4: 112257670-112257670 on Build GRCh38
rs762754162 Chr.4: 112257820-112257820 on Build GRCh38
rs986006540 Chr.4: 112257505-112257505 on Build GRCh38
rs947976985 Chr.4: 112257982-112257982 on Build GRCh38
rs4834248 Chr.4: 112257924-112257924 on Build GRCh38
rs771566036 Chr.4: 112257823-112257823 on Build GRCh38
rs563279589 Chr.4: 112257609-112257609 on Build GRCh38
rs770643579 Chr.4: 112257821-112257821 on Build GRCh38
rs1017477278 Chr.4: 112257900-112257900 on Build GRCh38
rs749259269 Chr.4: 112257738-112257739 on Build GRCh38
rs910365968 Chr.4: 112257505-112257505 on Build GRCh38
rs767464657 Chr.4: 112257830-112257830 on Build GRCh38
rs530550336 Chr.4: 112257631-112257631 on Build GRCh38
rs141048838 Chr.4: 112257712-112257712 on Build GRCh38
rs528654692 Chr.4: 112257818-112257818 on Build GRCh38
rs4834247 Chr.4: 112257921-112257921 on Build GRCh38
rs568629765 Chr.4: 112257842-112257842 on Build GRCh38
rs769403725 Chr.4: 112257795-112257795 on Build GRCh38
rs746339877 Chr.4: 112257733-112257733 on Build GRCh38
rs778807265 Chr.4: 112257811-112257812 on Build GRCh38
rs951992134 Chr.4: 112257504-112257505 on Build GRCh38
rs541428125 Chr.4: 112257577-112257577 on Build GRCh38

View Full Product Details