Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 123607413-123607854 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DDX55
Entrez Gene ID
57696
Gene Name
DEAD-box helicase 55
Gene Aliases
-
UniGene
Hs.286173
Gene Chromosome Location
Chr.12: 123602077-123620943 on Build GRCh38
Transcripts
NM_020936.2 NR_135104.1 NR_135105.1 XM_011538608.2 XM_017019710.1 XM_017019711.1 XM_017019712.1 XM_017019713.1 XM_017019714.1 XM_017019715.1 XM_017019716.1 XM_017019717.1 XM_017019718.1 XM_017019719.1 XM_017019720.1 XM_017019721.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
2225940 c.384G>A p.E128E Chr.12: 123607645-123607645 on Build GRCh38
6452819 c.312T>C p.H104H Chr.12: 123607497-123607497 on Build GRCh38
6641981 c.317C>T p.T106M Chr.12: 123607502-123607502 on Build GRCh38
3739485 c.347T>G p.L116R Chr.12: 123607608-123607608 on Build GRCh38
6136037 c.247-2A>T p.? Chr.12: 123607430-123607430 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs778479840 Chr.12: 123607553-123607553 on Build GRCh38
rs763233912 Chr.12: 123607528-123607528 on Build GRCh38
rs762282042 Chr.12: 123607640-123607640 on Build GRCh38
rs577087138 Chr.12: 123607709-123607709 on Build GRCh38
rs773049141 Chr.12: 123607507-123607507 on Build GRCh38
rs757879112 Chr.12: 123607538-123607538 on Build GRCh38
rs770668191 Chr.12: 123607576-123607576 on Build GRCh38
rs776566785 Chr.12: 123607581-123607581 on Build GRCh38
rs767924924 Chr.12: 123607657-123607657 on Build GRCh38
rs778130500 Chr.12: 123607476-123607476 on Build GRCh38
rs770167069 Chr.12: 123607738-123607741 on Build GRCh38
rs562895054 Chr.12: 123607729-123607729 on Build GRCh38
rs1011391510 Chr.12: 123607769-123607769 on Build GRCh38
rs374130222 Chr.12: 123607545-123607545 on Build GRCh38
rs901104924 Chr.12: 123607829-123607829 on Build GRCh38
rs61756271 Chr.12: 123607518-123607518 on Build GRCh38
rs371949103 Chr.12: 123607455-123607455 on Build GRCh38
rs764700195 Chr.12: 123607678-123607678 on Build GRCh38
rs752178019 Chr.12: 123607481-123607481 on Build GRCh38
rs1043819043 Chr.12: 123607741-123607741 on Build GRCh38
rs140354761 Chr.12: 123607503-123607503 on Build GRCh38
rs778444738 Chr.12: 123607502-123607502 on Build GRCh38
rs752912823 Chr.12: 123607468-123607468 on Build GRCh38
rs752217898 Chr.12: 123607685-123607685 on Build GRCh38
rs999531037 Chr.12: 123607802-123607802 on Build GRCh38
rs112887831 Chr.12: 123607728-123607728 on Build GRCh38
rs190027168 Chr.12: 123607482-123607482 on Build GRCh38
rs373488429 Chr.12: 123607493-123607493 on Build GRCh38
rs768164303 Chr.12: 123607494-123607494 on Build GRCh38
rs35941774 Chr.12: 123607778-123607778 on Build GRCh38
rs376680229 Chr.12: 123607533-123607533 on Build GRCh38
rs80215100 Chr.12: 123607457-123607457 on Build GRCh38
rs200182547 Chr.12: 123607515-123607515 on Build GRCh38
rs370673271 Chr.12: 123607568-123607568 on Build GRCh38
rs764425062 Chr.12: 123607530-123607530 on Build GRCh38
rs201842459 Chr.12: 123607478-123607478 on Build GRCh38
rs919467243 Chr.12: 123607582-123607582 on Build GRCh38
rs544514370 Chr.12: 123607724-123607724 on Build GRCh38
rs1018925635 Chr.12: 123607824-123607824 on Build GRCh38
rs181627413 Chr.12: 123607539-123607539 on Build GRCh38
rs766485430 Chr.12: 123607682-123607683 on Build GRCh38
rs759637997 Chr.12: 123607516-123607516 on Build GRCh38
rs775627647 Chr.12: 123607597-123607597 on Build GRCh38
rs765405824 Chr.12: 123607465-123607465 on Build GRCh38
rs752869604 Chr.12: 123607522-123607522 on Build GRCh38
rs17881657 Chr.12: 123607486-123607486 on Build GRCh38
rs771730185 Chr.12: 123607555-123607555 on Build GRCh38
rs774015411 Chr.12: 123607444-123607444 on Build GRCh38
rs751035732 Chr.12: 123607675-123607675 on Build GRCh38
rs374969518 Chr.12: 123607473-123607473 on Build GRCh38
rs373251266 Chr.12: 123607615-123607615 on Build GRCh38
rs763134643 Chr.12: 123607607-123607607 on Build GRCh38
rs763184304 Chr.12: 123607591-123607593 on Build GRCh38
rs753896710 Chr.12: 123607460-123607460 on Build GRCh38
rs376236187 Chr.12: 123607626-123607626 on Build GRCh38
rs769833920 Chr.12: 123607594-123607594 on Build GRCh38
rs922525255 Chr.12: 123607722-123607722 on Build GRCh38
rs146713519 Chr.12: 123607567-123607567 on Build GRCh38
rs74807783 Chr.12: 123607434-123607434 on Build GRCh38
rs754448851 Chr.12: 123607552-123607552 on Build GRCh38
rs902557333 Chr.12: 123607761-123607761 on Build GRCh38
rs983909521 Chr.12: 123607658-123607658 on Build GRCh38
rs909514060 Chr.12: 123607670-123607670 on Build GRCh38
rs759454773 Chr.12: 123607593-123607593 on Build GRCh38
rs370322499 Chr.12: 123607708-123607708 on Build GRCh38
rs770827926 Chr.12: 123607509-123607509 on Build GRCh38
rs766507170 Chr.12: 123607459-123607459 on Build GRCh38
rs1055310603 Chr.12: 123607590-123607590 on Build GRCh38
rs751954031 Chr.12: 123607669-123607669 on Build GRCh38
rs774667124 Chr.12: 123607630-123607630 on Build GRCh38
rs942393539 Chr.12: 123607686-123607686 on Build GRCh38
rs200057596 Chr.12: 123607616-123607616 on Build GRCh38
rs892329010 Chr.12: 123607732-123607732 on Build GRCh38
rs554684591 Chr.12: 123607735-123607735 on Build GRCh38
rs778710492 Chr.12: 123607529-123607529 on Build GRCh38
rs953839451 Chr.12: 123607517-123607517 on Build GRCh38
rs773030147 Chr.12: 123607456-123607456 on Build GRCh38
rs771831779 Chr.12: 123607505-123607505 on Build GRCh38
rs761535916 Chr.12: 123607451-123607451 on Build GRCh38
rs747810717 Chr.12: 123607554-123607554 on Build GRCh38

View Full Product Details