Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 80404218-80404746 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ARNT2
Entrez Gene ID
9915
Gene Name
aryl hydrocarbon receptor nuclear translocator 2
Gene Aliases
WEDAS, bHLHe1
UniGene
Hs.459070
Gene Chromosome Location
Chr.15: 80404350-80597937 on Build GRCh38
Transcripts
NM_014862.3

SNP Detail

SNP ID Chromosome Location
rs760114135 Chr.15: 80404590-80404590 on Build GRCh38
rs920430629 Chr.15: 80404686-80404686 on Build GRCh38
rs572066339 Chr.15: 80404554-80404554 on Build GRCh38
rs926129322 Chr.15: 80404377-80404377 on Build GRCh38
rs748498057 Chr.15: 80404465-80404465 on Build GRCh38
rs910494245 Chr.15: 80404433-80404433 on Build GRCh38
rs538143446 Chr.15: 80404628-80404628 on Build GRCh38
rs534422248 Chr.15: 80404654-80404654 on Build GRCh38
rs955441940 Chr.15: 80404651-80404651 on Build GRCh38
rs959653087 Chr.15: 80404350-80404350 on Build GRCh38
rs993251577 Chr.15: 80404319-80404319 on Build GRCh38
rs922613638 Chr.15: 80404408-80404409 on Build GRCh38
rs769959071 Chr.15: 80404509-80404509 on Build GRCh38
rs906136541 Chr.15: 80404582-80404582 on Build GRCh38
rs941961601 Chr.15: 80404343-80404343 on Build GRCh38
rs186548199 Chr.15: 80404427-80404427 on Build GRCh38
rs988873977 Chr.15: 80404402-80404402 on Build GRCh38
rs919071491 Chr.15: 80404538-80404538 on Build GRCh38
rs898941474 Chr.15: 80404273-80404273 on Build GRCh38
rs776919640 Chr.15: 80404500-80404500 on Build GRCh38
rs911216019 Chr.15: 80404673-80404673 on Build GRCh38
rs746092532 Chr.15: 80404503-80404503 on Build GRCh38
rs1033599084 Chr.15: 80404618-80404618 on Build GRCh38
rs558281643 Chr.15: 80404475-80404475 on Build GRCh38
rs1000450855 Chr.15: 80404614-80404614 on Build GRCh38
rs867169052 Chr.15: 80404674-80404674 on Build GRCh38
rs761547949 Chr.15: 80404543-80404543 on Build GRCh38
rs1022618236 Chr.15: 80404556-80404556 on Build GRCh38
rs774166346 Chr.15: 80404527-80404527 on Build GRCh38
rs979361661 Chr.15: 80404371-80404371 on Build GRCh38
rs988869574 Chr.15: 80404635-80404635 on Build GRCh38
rs865783188 Chr.15: 80404293-80404293 on Build GRCh38
rs1014520776 Chr.15: 80404361-80404361 on Build GRCh38
rs758897467 Chr.15: 80404470-80404470 on Build GRCh38
rs771137910 Chr.15: 80404496-80404496 on Build GRCh38
rs929123822 Chr.15: 80404713-80404713 on Build GRCh38
rs1029680533 Chr.15: 80404637-80404637 on Build GRCh38
rs943289296 Chr.15: 80404453-80404453 on Build GRCh38
rs767398217 Chr.15: 80404574-80404574 on Build GRCh38
rs938525487 Chr.15: 80404563-80404563 on Build GRCh38
rs933964746 Chr.15: 80404413-80404413 on Build GRCh38
rs777576398 Chr.15: 80404680-80404680 on Build GRCh38
rs985456828 Chr.15: 80404663-80404663 on Build GRCh38
rs1026023821 Chr.15: 80404344-80404344 on Build GRCh38
rs868095569 Chr.15: 80404367-80404367 on Build GRCh38
rs773149547 Chr.15: 80404575-80404575 on Build GRCh38
rs747026664 Chr.15: 80404490-80404490 on Build GRCh38
rs1037971559 Chr.15: 80404474-80404474 on Build GRCh38
rs956359322 Chr.15: 80404382-80404382 on Build GRCh38

View Full Product Details