Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 81196793-81197288 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
IL16
Entrez Gene ID
3603
Gene Name
interleukin 16
Gene Aliases
LCF, NIL16, PRIL16, prIL-16
UniGene
Hs.459095
Gene Chromosome Location
Chr.15: 81181844-81312763 on Build GRCh38
Transcripts
NM_001172128.1 XM_005254342.3 XM_011521520.2 XM_017022141.1 XM_017022142.1 XM_017022143.1 XM_017022144.1 XM_017022145.1

SNP Detail

SNP ID Chromosome Location
rs920982285 Chr.15: 81197128-81197128 on Build GRCh38
rs564616028 Chr.15: 81196855-81196855 on Build GRCh38
rs181295603 Chr.15: 81197194-81197194 on Build GRCh38
rs143647835 Chr.15: 81197185-81197185 on Build GRCh38
rs939460600 Chr.15: 81197137-81197137 on Build GRCh38
rs1056703392 Chr.15: 81196985-81196985 on Build GRCh38
rs958708825 Chr.15: 81197046-81197046 on Build GRCh38
rs975947469 Chr.15: 81196820-81196820 on Build GRCh38
rs114073584 Chr.15: 81197077-81197077 on Build GRCh38
rs8037800 Chr.15: 81196983-81196983 on Build GRCh38
rs1005389090 Chr.15: 81197187-81197187 on Build GRCh38
rs74030008 Chr.15: 81197162-81197162 on Build GRCh38
rs1056772289 Chr.15: 81197141-81197141 on Build GRCh38
rs1031901978 Chr.15: 81197209-81197209 on Build GRCh38
rs1049511410 Chr.15: 81197169-81197169 on Build GRCh38
rs759807670 Chr.15: 81197132-81197132 on Build GRCh38
rs921615304 Chr.15: 81196836-81196836 on Build GRCh38
rs939924901 Chr.15: 81197146-81197146 on Build GRCh38
rs201484678 Chr.15: 81197055-81197055 on Build GRCh38
rs1011265310 Chr.15: 81197212-81197212 on Build GRCh38
rs148743629 Chr.15: 81197133-81197133 on Build GRCh38
rs547050915 Chr.15: 81196975-81196975 on Build GRCh38
rs997303486 Chr.15: 81197198-81197198 on Build GRCh38
rs556631048 Chr.15: 81197118-81197118 on Build GRCh38
rs965821440 Chr.15: 81197052-81197052 on Build GRCh38
rs879233824 Chr.15: 81197256-81197256 on Build GRCh38
rs1035786285 Chr.15: 81197181-81197181 on Build GRCh38
rs965529655 Chr.15: 81197225-81197225 on Build GRCh38
rs924427069 Chr.15: 81196938-81196938 on Build GRCh38
rs932324337 Chr.15: 81197129-81197129 on Build GRCh38
rs953017127 Chr.15: 81196854-81196854 on Build GRCh38
rs967514154 Chr.15: 81197123-81197123 on Build GRCh38
rs965913007 Chr.15: 81196819-81196819 on Build GRCh38
rs1049179517 Chr.15: 81197003-81197003 on Build GRCh38
rs200767560 Chr.15: 81197051-81197051 on Build GRCh38
rs947588000 Chr.15: 81196907-81196907 on Build GRCh38
rs888324723 Chr.15: 81197018-81197018 on Build GRCh38
rs190196640 Chr.15: 81197034-81197034 on Build GRCh38
rs773516652 Chr.15: 81196950-81196950 on Build GRCh38
rs765318997 Chr.15: 81197162-81197165 on Build GRCh38
rs1028902693 Chr.15: 81197199-81197199 on Build GRCh38
rs914799713 Chr.15: 81196896-81196896 on Build GRCh38
rs901688116 Chr.15: 81197195-81197195 on Build GRCh38
rs772593948 Chr.15: 81196949-81196949 on Build GRCh38
rs149293610 Chr.15: 81196934-81196934 on Build GRCh38
rs773305433 Chr.15: 81197066-81197066 on Build GRCh38
rs536700095 Chr.15: 81197117-81197117 on Build GRCh38
rs775732245 Chr.15: 81196966-81196966 on Build GRCh38
rs1004741505 Chr.15: 81197036-81197036 on Build GRCh38
rs573448886 Chr.15: 81197201-81197201 on Build GRCh38
rs1028829073 Chr.15: 81197081-81197081 on Build GRCh38
rs761920758 Chr.15: 81197124-81197124 on Build GRCh38
rs757178178 Chr.15: 81197165-81197165 on Build GRCh38
rs1042938482 Chr.15: 81196974-81196974 on Build GRCh38
rs989862415 Chr.15: 81196895-81196895 on Build GRCh38
rs1035755546 Chr.15: 81197045-81197045 on Build GRCh38
rs916826040 Chr.15: 81197155-81197155 on Build GRCh38
rs1036894579 Chr.15: 81197153-81197153 on Build GRCh38
rs953089258 Chr.15: 81197111-81197111 on Build GRCh38
rs185808952 Chr.15: 81197217-81197217 on Build GRCh38
rs4346163 Chr.15: 81196998-81196998 on Build GRCh38
rs1015188163 Chr.15: 81197049-81197049 on Build GRCh38
rs144571346 Chr.15: 81196997-81196997 on Build GRCh38
rs542169753 Chr.15: 81197204-81197204 on Build GRCh38
rs991760273 Chr.15: 81196844-81196844 on Build GRCh38
rs924762465 Chr.15: 81197121-81197121 on Build GRCh38

View Full Product Details