Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 11875681-11876194 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GSPT1
Entrez Gene ID
2935
Gene Name
G1 to S phase transition 1
Gene Aliases
551G9.2, ETF3A, GST1, eRF3a
UniGene
Hs.528780
Gene Chromosome Location
Chr.16: 11868128-11916662 on Build GRCh38
Transcripts
NM_001130006.1 NM_001130007.1 NM_002094.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1194207 c.1249C>T p.H417Y Chr.16: 11876115-11876115 on Build GRCh38
5778426 c.1236G>C p.A412A Chr.16: 11876128-11876128 on Build GRCh38
5788545 c.1236G>A p.A412A Chr.16: 11876128-11876128 on Build GRCh38
6230775 c.1748G>A p.R583H Chr.16: 11875874-11875874 on Build GRCh38
4403299 c.1718A>C p.K573T Chr.16: 11875904-11875904 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs758194319 Chr.16: 11875959-11875959 on Build GRCh38
rs769364812 Chr.16: 11875882-11875882 on Build GRCh38
rs969134385 Chr.16: 11875793-11875793 on Build GRCh38
rs754881584 Chr.16: 11876038-11876038 on Build GRCh38
rs753362977 Chr.16: 11876041-11876051 on Build GRCh38
rs979467815 Chr.16: 11875848-11875848 on Build GRCh38
rs527304051 Chr.16: 11876041-11876041 on Build GRCh38
rs764237016 Chr.16: 11875948-11875948 on Build GRCh38
rs896170810 Chr.16: 11876016-11876016 on Build GRCh38
rs989103655 Chr.16: 11875993-11875993 on Build GRCh38
rs945460140 Chr.16: 11875785-11875785 on Build GRCh38
rs376930550 Chr.16: 11876075-11876075 on Build GRCh38
rs372857360 Chr.16: 11875740-11875740 on Build GRCh38
rs775055818 Chr.16: 11875900-11875900 on Build GRCh38
rs936241518 Chr.16: 11875973-11875973 on Build GRCh38
rs763175923 Chr.16: 11875978-11875978 on Build GRCh38
rs779890578 Chr.16: 11875977-11875977 on Build GRCh38
rs771249200 Chr.16: 11876113-11876113 on Build GRCh38
rs766757624 Chr.16: 11875889-11875889 on Build GRCh38
rs779864647 Chr.16: 11875774-11875774 on Build GRCh38
rs371597035 Chr.16: 11875943-11875943 on Build GRCh38
rs33657 Chr.16: 11875867-11875867 on Build GRCh38
rs564805529 Chr.16: 11876122-11876122 on Build GRCh38
rs752854616 Chr.16: 11875945-11875945 on Build GRCh38
rs752036346 Chr.16: 11875715-11875715 on Build GRCh38
rs772860100 Chr.16: 11875885-11875885 on Build GRCh38
rs768157958 Chr.16: 11876166-11876166 on Build GRCh38
rs942828628 Chr.16: 11876089-11876089 on Build GRCh38
rs781403566 Chr.16: 11875717-11875717 on Build GRCh38
rs753014078 Chr.16: 11875720-11875720 on Build GRCh38
rs1023360701 Chr.16: 11875767-11875767 on Build GRCh38
rs371628512 Chr.16: 11876101-11876101 on Build GRCh38
rs746948412 Chr.16: 11875808-11875808 on Build GRCh38
rs771846442 Chr.16: 11875749-11875749 on Build GRCh38
rs181080797 Chr.16: 11876059-11876059 on Build GRCh38
rs369462446 Chr.16: 11876128-11876128 on Build GRCh38
rs934134787 Chr.16: 11876026-11876026 on Build GRCh38
rs767744351 Chr.16: 11875944-11875944 on Build GRCh38
rs768214275 Chr.16: 11875852-11875852 on Build GRCh38
rs1051215546 Chr.16: 11876033-11876033 on Build GRCh38
rs995261513 Chr.16: 11876035-11876035 on Build GRCh38
rs761495945 Chr.16: 11875880-11875880 on Build GRCh38
rs541376565 Chr.16: 11876021-11876021 on Build GRCh38
rs201111838 Chr.16: 11875794-11875794 on Build GRCh38
rs200192283 Chr.16: 11875870-11875870 on Build GRCh38
rs745675621 Chr.16: 11875744-11875744 on Build GRCh38
rs992011239 Chr.16: 11875735-11875735 on Build GRCh38
rs913949538 Chr.16: 11875732-11875732 on Build GRCh38
rs142230716 Chr.16: 11875716-11875716 on Build GRCh38
rs756370927 Chr.16: 11875733-11875733 on Build GRCh38
rs1044311075 Chr.16: 11875830-11875830 on Build GRCh38
rs190312938 Chr.16: 11875772-11875772 on Build GRCh38
rs765293225 Chr.16: 11875845-11875845 on Build GRCh38
rs754026821 Chr.16: 11875949-11875949 on Build GRCh38
rs772803567 Chr.16: 11876067-11876067 on Build GRCh38

View Full Product Details