Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 19884638-19885129 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GPRC5B
Entrez Gene ID
51704
Gene Name
G protein-coupled receptor class C group 5 member B
Gene Aliases
RAIG-2, RAIG2
UniGene
Hs.148685
Gene Chromosome Location
Chr.16: 19858971-19886045 on Build GRCh38
Transcripts
NM_001304771.1 NM_016235.2 XM_006721052.2

SNP Detail

SNP ID Chromosome Location
rs921711461 Chr.16: 19884876-19884876 on Build GRCh38
rs1027544561 Chr.16: 19885075-19885075 on Build GRCh38
rs1020365486 Chr.16: 19885001-19885001 on Build GRCh38
rs188412083 Chr.16: 19885023-19885023 on Build GRCh38
rs1034472024 Chr.16: 19884767-19884767 on Build GRCh38
rs907073858 Chr.16: 19884742-19884742 on Build GRCh38
rs549506014 Chr.16: 19884893-19884893 on Build GRCh38
rs762178006 Chr.16: 19885032-19885032 on Build GRCh38
rs987077286 Chr.16: 19884820-19884820 on Build GRCh38
rs781518435 Chr.16: 19885037-19885037 on Build GRCh38
rs961720369 Chr.16: 19885108-19885108 on Build GRCh38
rs951843402 Chr.16: 19885080-19885080 on Build GRCh38
rs564636861 Chr.16: 19884940-19884940 on Build GRCh38
rs185723562 Chr.16: 19885104-19885104 on Build GRCh38
rs1008231631 Chr.16: 19885020-19885020 on Build GRCh38
rs1012045401 Chr.16: 19884982-19884982 on Build GRCh38
rs866945265 Chr.16: 19884896-19884896 on Build GRCh38
rs867083482 Chr.16: 19884661-19884661 on Build GRCh38
rs886361931 Chr.16: 19884701-19884701 on Build GRCh38
rs531985689 Chr.16: 19885033-19885033 on Build GRCh38
rs984492667 Chr.16: 19884889-19884889 on Build GRCh38
rs967634060 Chr.16: 19884789-19884789 on Build GRCh38
rs111852063 Chr.16: 19884990-19884990 on Build GRCh38
rs997029545 Chr.16: 19885065-19885065 on Build GRCh38
rs1045554302 Chr.16: 19884736-19884736 on Build GRCh38
rs531797431 Chr.16: 19884888-19884888 on Build GRCh38
rs184812233 Chr.16: 19884870-19884870 on Build GRCh38
rs1011578894 Chr.16: 19884775-19884775 on Build GRCh38
rs903206551 Chr.16: 19885018-19885022 on Build GRCh38
rs984533897 Chr.16: 19885102-19885102 on Build GRCh38
rs893176117 Chr.16: 19884769-19884769 on Build GRCh38
rs907121171 Chr.16: 19884911-19884911 on Build GRCh38
rs930067820 Chr.16: 19885109-19885109 on Build GRCh38
rs1049173221 Chr.16: 19884674-19884674 on Build GRCh38
rs1020272034 Chr.16: 19884777-19884777 on Build GRCh38
rs1019858545 Chr.16: 19884825-19884825 on Build GRCh38
rs964285133 Chr.16: 19884853-19884853 on Build GRCh38
rs918560822 Chr.16: 19884670-19884670 on Build GRCh38
rs925100921 Chr.16: 19884927-19884942 on Build GRCh38
rs546949916 Chr.16: 19885054-19885054 on Build GRCh38
rs907520279 Chr.16: 19884890-19884890 on Build GRCh38
rs893237543 Chr.16: 19884979-19884979 on Build GRCh38
rs180838340 Chr.16: 19885092-19885092 on Build GRCh38
rs975981238 Chr.16: 19884872-19884872 on Build GRCh38
rs940589846 Chr.16: 19884713-19884713 on Build GRCh38
rs867617674 Chr.16: 19884880-19884880 on Build GRCh38
rs376882270 Chr.16: 19885000-19885000 on Build GRCh38
rs1019914425 Chr.16: 19885024-19885024 on Build GRCh38

View Full Product Details