Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 68228625-68229170 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ESRP2
Entrez Gene ID
80004
Gene Name
epithelial splicing regulatory protein 2
Gene Aliases
RBM35B
UniGene
Hs.436585
Gene Chromosome Location
Chr.16: 68228547-68236233 on Build GRCh38
Transcripts
NM_024939.2 XM_006721273.3 XM_011523338.2 NM_004555.3 NM_173163.2 NM_173165.2

Gene Symbol
NFATC3
Entrez Gene ID
4775
Gene Name
nuclear factor of activated T-cells 3
Gene Aliases
NFAT4, NFATX
UniGene
Hs.436585
Gene Chromosome Location
Chr.16: 68085366-68229259 on Build GRCh38
Transcripts
NM_024939.2 XM_006721273.3 XM_011523338.2 NM_004555.3 NM_173163.2 NM_173165.2

SNP Detail

SNP ID Chromosome Location
rs1035104056 Chr.16: 68228728-68228728 on Build GRCh38
rs148162013 Chr.16: 68228897-68228897 on Build GRCh38
rs995154539 Chr.16: 68229119-68229119 on Build GRCh38
rs955115446 Chr.16: 68228678-68228679 on Build GRCh38
rs951275582 Chr.16: 68228950-68228950 on Build GRCh38
rs764534876 Chr.16: 68229041-68229041 on Build GRCh38
rs1012688720 Chr.16: 68228779-68228779 on Build GRCh38
rs551701291 Chr.16: 68229104-68229104 on Build GRCh38
rs1027611108 Chr.16: 68229136-68229136 on Build GRCh38
rs960674742 Chr.16: 68228778-68228778 on Build GRCh38
rs532698163 Chr.16: 68228837-68228837 on Build GRCh38
rs974827471 Chr.16: 68228915-68228915 on Build GRCh38
rs976882321 Chr.16: 68228938-68228938 on Build GRCh38
rs948160064 Chr.16: 68228753-68228753 on Build GRCh38
rs1024059468 Chr.16: 68229102-68229102 on Build GRCh38
rs759200993 Chr.16: 68229016-68229016 on Build GRCh38
rs796440943 Chr.16: 68228865-68228865 on Build GRCh38
rs969448972 Chr.16: 68229093-68229093 on Build GRCh38
rs902683922 Chr.16: 68228977-68228977 on Build GRCh38
rs142270392 Chr.16: 68228675-68228675 on Build GRCh38
rs1057067164 Chr.16: 68228792-68228792 on Build GRCh38
rs1022849863 Chr.16: 68228784-68228784 on Build GRCh38
rs902569880 Chr.16: 68228690-68228690 on Build GRCh38
rs565213530 Chr.16: 68228808-68228808 on Build GRCh38
rs911668809 Chr.16: 68228879-68228879 on Build GRCh38
rs1029047369 Chr.16: 68228935-68228935 on Build GRCh38
rs987751799 Chr.16: 68228693-68228693 on Build GRCh38
rs3833060 Chr.16: 68228665-68228665 on Build GRCh38
rs143832712 Chr.16: 68228695-68228695 on Build GRCh38
rs995968239 Chr.16: 68228903-68228903 on Build GRCh38
rs1015182937 Chr.16: 68228995-68228995 on Build GRCh38
rs1012223631 Chr.16: 68229065-68229065 on Build GRCh38
rs867879471 Chr.16: 68228791-68228791 on Build GRCh38
rs11043 Chr.16: 68229055-68229055 on Build GRCh38
rs7195949 Chr.16: 68228974-68228974 on Build GRCh38
rs562450958 Chr.16: 68228871-68228871 on Build GRCh38
rs746491441 Chr.16: 68228848-68228848 on Build GRCh38
rs755228726 Chr.16: 68228836-68228838 on Build GRCh38
rs772815082 Chr.16: 68228733-68228733 on Build GRCh38
rs918354070 Chr.16: 68228943-68228943 on Build GRCh38
rs766843163 Chr.16: 68228676-68228681 on Build GRCh38
rs1021451437 Chr.16: 68228809-68228809 on Build GRCh38
rs530589602 Chr.16: 68228972-68228972 on Build GRCh38
rs567403863 Chr.16: 68228975-68228975 on Build GRCh38
rs920671621 Chr.16: 68228748-68228748 on Build GRCh38
rs950513867 Chr.16: 68229140-68229140 on Build GRCh38
rs542658592 Chr.16: 68228651-68228651 on Build GRCh38
rs973449186 Chr.16: 68228713-68228713 on Build GRCh38
rs922186016 Chr.16: 68228930-68228930 on Build GRCh38
rs1045456114 Chr.16: 68228769-68228769 on Build GRCh38
rs920430905 Chr.16: 68228800-68228800 on Build GRCh38
rs891592656 Chr.16: 68228795-68228795 on Build GRCh38
rs183133847 Chr.16: 68228700-68228700 on Build GRCh38
rs562762314 Chr.16: 68228953-68228953 on Build GRCh38
rs908009966 Chr.16: 68228697-68228697 on Build GRCh38
rs865952018 Chr.16: 68228790-68228790 on Build GRCh38

View Full Product Details