Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 69121111-69121514 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CHTF8
Entrez Gene ID
54921
Gene Name
chromosome transmission fidelity factor 8
Gene Aliases
CTF8, DERPC
UniGene
Hs.85962
Gene Chromosome Location
Chr.16: 69118009-69132590 on Build GRCh38
Transcripts
NM_001039690.3 NM_001040146.3 NR_033227.1 XM_011523167.1 XM_011523168.1 XM_017023370.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5706188 c.3G>A p.M1I Chr.16: 69121456-69121456 on Build GRCh38
4062215 c.27G>A p.A9A Chr.16: 69121167-69121167 on Build GRCh38
4129243 c.25G>T p.A9S Chr.16: 69121169-69121169 on Build GRCh38
6629136 c.16A>G p.I6V Chr.16: 69121443-69121443 on Build GRCh38
6629138 c.25G>A p.A9T Chr.16: 69121169-69121169 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs924962213 Chr.16: 69121284-69121284 on Build GRCh38
rs773795101 Chr.16: 69121432-69121432 on Build GRCh38
rs761293566 Chr.16: 69121433-69121433 on Build GRCh38
rs1024642519 Chr.16: 69121235-69121235 on Build GRCh38
rs888034154 Chr.16: 69121155-69121155 on Build GRCh38
rs759713674 Chr.16: 69121317-69121317 on Build GRCh38
rs549595126 Chr.16: 69121239-69121239 on Build GRCh38
rs756442040 Chr.16: 69121209-69121209 on Build GRCh38
rs911158090 Chr.16: 69121355-69121355 on Build GRCh38
rs749496352 Chr.16: 69121387-69121387 on Build GRCh38
rs901315935 Chr.16: 69121244-69121244 on Build GRCh38
rs201301001 Chr.16: 69121390-69121390 on Build GRCh38
rs375312733 Chr.16: 69121255-69121255 on Build GRCh38
rs201210325 Chr.16: 69121388-69121388 on Build GRCh38
rs754165889 Chr.16: 69121467-69121467 on Build GRCh38
rs142784676 Chr.16: 69121388-69121388 on Build GRCh38
rs755213875 Chr.16: 69121474-69121474 on Build GRCh38
rs531988610 Chr.16: 69121433-69121434 on Build GRCh38
rs745758551 Chr.16: 69121414-69121414 on Build GRCh38
rs776877950 Chr.16: 69121283-69121283 on Build GRCh38
rs768055082 Chr.16: 69121435-69121435 on Build GRCh38
rs150188271 Chr.16: 69121189-69121190 on Build GRCh38
rs779173707 Chr.16: 69121478-69121478 on Build GRCh38
rs750070477 Chr.16: 69121462-69121465 on Build GRCh38
rs761123744 Chr.16: 69121455-69121455 on Build GRCh38
rs769730716 Chr.16: 69121427-69121427 on Build GRCh38
rs771175928 Chr.16: 69121260-69121260 on Build GRCh38
rs752863103 Chr.16: 69121479-69121479 on Build GRCh38
rs751019271 Chr.16: 69121197-69121197 on Build GRCh38
rs775487827 Chr.16: 69121431-69121431 on Build GRCh38
rs953968974 Chr.16: 69121351-69121351 on Build GRCh38
rs776627380 Chr.16: 69121156-69121156 on Build GRCh38
rs201600961 Chr.16: 69121167-69121167 on Build GRCh38
rs977723849 Chr.16: 69121240-69121240 on Build GRCh38
rs777948178 Chr.16: 69121485-69121485 on Build GRCh38
rs192664244 Chr.16: 69121389-69121389 on Build GRCh38
rs200097399 Chr.16: 69121207-69121207 on Build GRCh38
rs762512928 Chr.16: 69121428-69121428 on Build GRCh38
rs772574013 Chr.16: 69121190-69121190 on Build GRCh38
rs376900622 Chr.16: 69121195-69121195 on Build GRCh38
rs1048366961 Chr.16: 69121483-69121483 on Build GRCh38
rs138743702 Chr.16: 69121477-69121480 on Build GRCh38
rs780427524 Chr.16: 69121211-69121211 on Build GRCh38
rs957968029 Chr.16: 69121304-69121304 on Build GRCh38
rs748245611 Chr.16: 69121392-69121392 on Build GRCh38
rs376615755 Chr.16: 69121294-69121294 on Build GRCh38
rs1032708037 Chr.16: 69121374-69121374 on Build GRCh38
rs766667272 Chr.16: 69121462-69121462 on Build GRCh38
rs758579769 Chr.16: 69121481-69121481 on Build GRCh38
rs757823397 Chr.16: 69121203-69121203 on Build GRCh38
rs980374974 Chr.16: 69121449-69121449 on Build GRCh38
rs118093016 Chr.16: 69121406-69121406 on Build GRCh38
rs770988516 Chr.16: 69121142-69121142 on Build GRCh38
rs767995655 Chr.16: 69121206-69121206 on Build GRCh38
rs377631210 Chr.16: 69121373-69121373 on Build GRCh38
rs534770175 Chr.16: 69121242-69121242 on Build GRCh38
rs762620178 Chr.16: 69121170-69121170 on Build GRCh38
rs778057586 Chr.16: 69121364-69121364 on Build GRCh38
rs373936693 Chr.16: 69121168-69121168 on Build GRCh38
rs771039848 Chr.16: 69121393-69121393 on Build GRCh38
rs141542657 Chr.16: 69121436-69121436 on Build GRCh38
rs985752428 Chr.16: 69121307-69121307 on Build GRCh38
rs765029061 Chr.16: 69121435-69121435 on Build GRCh38

View Full Product Details