Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 55079779-55080267 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EPS8L1
Entrez Gene ID
54869
Gene Name
EPS8 like 1
Gene Aliases
DRC3, EPS8R1, PP10566
UniGene
Hs.438862
Gene Chromosome Location
Chr.19: 55075853-55087923 on Build GRCh38
Transcripts
NM_133180.2 XM_005259020.1 XM_011527051.2 XM_011527052.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7177127 c.209G>C p.R70P Chr.19: 55079781-55079781 on Build GRCh38
4127889 c.411C>T p.F137F Chr.19: 55080260-55080260 on Build GRCh38
4540242 c.226G>A p.E76K Chr.19: 55080129-55080129 on Build GRCh38
4485835 c.243C>T p.Y81Y Chr.19: 55080146-55080146 on Build GRCh38
6862997 c.242C>A p.S81Y Chr.19: 55079814-55079814 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs563334472 Chr.19: 55079932-55079932 on Build GRCh38
rs536448589 Chr.19: 55079836-55079836 on Build GRCh38
rs143589639 Chr.19: 55079834-55079834 on Build GRCh38
rs949228548 Chr.19: 55079892-55079892 on Build GRCh38
rs567891670 Chr.19: 55079807-55079807 on Build GRCh38
rs1009493951 Chr.19: 55080007-55080007 on Build GRCh38
rs764451379 Chr.19: 55079896-55079896 on Build GRCh38
rs899114735 Chr.19: 55080093-55080093 on Build GRCh38
rs543739662 Chr.19: 55079931-55079931 on Build GRCh38
rs755703244 Chr.19: 55079888-55079888 on Build GRCh38
rs371821193 Chr.19: 55080215-55080215 on Build GRCh38
rs575798648 Chr.19: 55079961-55079961 on Build GRCh38
rs748051097 Chr.19: 55080176-55080176 on Build GRCh38
rs761234909 Chr.19: 55079861-55079861 on Build GRCh38
rs780074272 Chr.19: 55080154-55080154 on Build GRCh38
rs200919752 Chr.19: 55080167-55080167 on Build GRCh38
rs905126817 Chr.19: 55079922-55079922 on Build GRCh38
rs766451385 Chr.19: 55080218-55080218 on Build GRCh38
rs775399659 Chr.19: 55079855-55079855 on Build GRCh38
rs367696708 Chr.19: 55079865-55079865 on Build GRCh38
rs577843967 Chr.19: 55079929-55079929 on Build GRCh38
rs200577842 Chr.19: 55080246-55080246 on Build GRCh38
rs760706669 Chr.19: 55080217-55080217 on Build GRCh38
rs988884364 Chr.19: 55080064-55080064 on Build GRCh38
rs755230546 Chr.19: 55079872-55079872 on Build GRCh38
rs56378581 Chr.19: 55080024-55080024 on Build GRCh38
rs993286721 Chr.19: 55079957-55079957 on Build GRCh38
rs1007496125 Chr.19: 55079813-55079813 on Build GRCh38
rs766823918 Chr.19: 55079868-55079868 on Build GRCh38
rs749255194 Chr.19: 55080162-55080162 on Build GRCh38
rs76480207 Chr.19: 55079945-55079945 on Build GRCh38
rs148068933 Chr.19: 55079837-55079837 on Build GRCh38
rs917345267 Chr.19: 55080124-55080124 on Build GRCh38
rs750883568 Chr.19: 55079829-55079829 on Build GRCh38
rs758163227 Chr.19: 55080063-55080063 on Build GRCh38
rs746677729 Chr.19: 55079841-55079841 on Build GRCh38
rs959187418 Chr.19: 55079926-55079926 on Build GRCh38
rs772137547 Chr.19: 55080233-55080234 on Build GRCh38
rs779706284 Chr.19: 55080156-55080156 on Build GRCh38
rs767652689 Chr.19: 55079819-55079819 on Build GRCh38
rs73932294 Chr.19: 55079870-55079870 on Build GRCh38
rs757246099 Chr.19: 55080031-55080031 on Build GRCh38
rs890221853 Chr.19: 55080243-55080243 on Build GRCh38
rs559851865 Chr.19: 55079959-55079959 on Build GRCh38
rs374610286 Chr.19: 55079810-55079810 on Build GRCh38
rs551779961 Chr.19: 55079990-55079990 on Build GRCh38
rs1051357052 Chr.19: 55079979-55079979 on Build GRCh38
rs557358947 Chr.19: 55079886-55079886 on Build GRCh38
rs769890008 Chr.19: 55079848-55079848 on Build GRCh38
rs761736038 Chr.19: 55079811-55079811 on Build GRCh38
rs774319978 Chr.19: 55079856-55079857 on Build GRCh38
rs1002975798 Chr.19: 55080165-55080165 on Build GRCh38
rs375136267 Chr.19: 55080216-55080216 on Build GRCh38
rs561472001 Chr.19: 55080245-55080245 on Build GRCh38
rs748289464 Chr.19: 55080188-55080188 on Build GRCh38
rs1017749644 Chr.19: 55080008-55080008 on Build GRCh38
rs756998203 Chr.19: 55079883-55079883 on Build GRCh38
rs774381920 Chr.19: 55079858-55079858 on Build GRCh38
rs745793450 Chr.19: 55079847-55079847 on Build GRCh38
rs898857694 Chr.19: 55080234-55080234 on Build GRCh38
rs376374139 Chr.19: 55079857-55079857 on Build GRCh38
rs1018261677 Chr.19: 55079831-55079831 on Build GRCh38
rs1037614617 Chr.19: 55080082-55080082 on Build GRCh38
rs781467148 Chr.19: 55079836-55079838 on Build GRCh38
rs914843446 Chr.19: 55079934-55079934 on Build GRCh38
rs1037620069 Chr.19: 55080233-55080233 on Build GRCh38
rs199714657 Chr.19: 55080166-55080166 on Build GRCh38
rs770985825 Chr.19: 55079799-55079801 on Build GRCh38
rs563946895 Chr.19: 55079947-55079947 on Build GRCh38
rs760145394 Chr.19: 55080232-55080232 on Build GRCh38
rs780533306 Chr.19: 55079885-55079885 on Build GRCh38
rs1046199723 Chr.19: 55079904-55079904 on Build GRCh38
rs370375170 Chr.19: 55079808-55079808 on Build GRCh38
rs201391252 Chr.19: 55079862-55079862 on Build GRCh38
rs1047401138 Chr.19: 55080239-55080239 on Build GRCh38
rs751157641 Chr.19: 55079881-55079881 on Build GRCh38
rs141622953 Chr.19: 55079842-55079842 on Build GRCh38
rs565316227 Chr.19: 55079999-55079999 on Build GRCh38
rs937959838 Chr.19: 55080174-55080174 on Build GRCh38
rs866935004 Chr.19: 55080139-55080139 on Build GRCh38
rs928945671 Chr.19: 55080235-55080235 on Build GRCh38
rs777063967 Chr.19: 55080231-55080231 on Build GRCh38
rs765513309 Chr.19: 55080237-55080237 on Build GRCh38
rs368298723 Chr.19: 55079857-55079857 on Build GRCh38
rs979027841 Chr.19: 55080025-55080025 on Build GRCh38
rs746129669 Chr.19: 55079854-55079855 on Build GRCh38
rs749038247 Chr.19: 55079856-55079856 on Build GRCh38
rs879185633 Chr.19: 55079924-55079924 on Build GRCh38

View Full Product Details