Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 48990444-48990941 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FSHR
Entrez Gene ID
2492
Gene Name
follicle stimulating hormone receptor
Gene Aliases
FSHR1, FSHRO, LGR1, ODG1
UniGene
Hs.1428
Gene Chromosome Location
Chr.2: 48953161-49154527 on Build GRCh38
Transcripts
NM_000145.3 NM_181446.2 XM_011532733.2 XM_011532734.2 XM_011532735.2 XM_011532736.2 XM_011532740.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
442990 c.406C>T p.P136S Chr.2: 48990606-48990606 on Build GRCh38
3962889 c.391G>T p.G131C Chr.2: 48990621-48990621 on Build GRCh38
6667491 c.431_432delTC p.L144fs*6 Chr.2: 48990580-48990581 on Build GRCh38
88106 c.382T>C p.S128P Chr.2: 48990630-48990630 on Build GRCh38
4094534 c.438A>T p.K146N Chr.2: 48990574-48990574 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1048715174 Chr.2: 48990500-48990500 on Build GRCh38
rs780619335 Chr.2: 48990548-48990548 on Build GRCh38
rs1048742477 Chr.2: 48990769-48990769 on Build GRCh38
rs1036457358 Chr.2: 48990474-48990474 on Build GRCh38
rs971807816 Chr.2: 48990705-48990705 on Build GRCh38
rs10644983 Chr.2: 48990892-48990893 on Build GRCh38
rs74526416 Chr.2: 48990682-48990682 on Build GRCh38
rs770728831 Chr.2: 48990561-48990561 on Build GRCh38
rs774182075 Chr.2: 48990601-48990601 on Build GRCh38
rs144458659 Chr.2: 48990508-48990508 on Build GRCh38
rs911708720 Chr.2: 48990659-48990659 on Build GRCh38
rs763846559 Chr.2: 48990600-48990600 on Build GRCh38
rs190202012 Chr.2: 48990798-48990798 on Build GRCh38
rs140415279 Chr.2: 48990617-48990617 on Build GRCh38
rs386390139 Chr.2: 48990893-48990894 on Build GRCh38
rs368775695 Chr.2: 48990901-48990902 on Build GRCh38
rs944456938 Chr.2: 48990472-48990472 on Build GRCh38
rs567725056 Chr.2: 48990800-48990800 on Build GRCh38
rs779456539 Chr.2: 48990555-48990555 on Build GRCh38
rs373439537 Chr.2: 48990535-48990535 on Build GRCh38
rs397813924 Chr.2: 48990902-48990903 on Build GRCh38
rs886154018 Chr.2: 48990737-48990737 on Build GRCh38
rs558872182 Chr.2: 48990903-48990903 on Build GRCh38
rs937389920 Chr.2: 48990838-48990838 on Build GRCh38
rs879339700 Chr.2: 48990504-48990504 on Build GRCh38
rs777165286 Chr.2: 48990615-48990616 on Build GRCh38
rs367688436 Chr.2: 48990906-48990906 on Build GRCh38
rs913411858 Chr.2: 48990804-48990804 on Build GRCh38
rs946116596 Chr.2: 48990641-48990641 on Build GRCh38
rs878983875 Chr.2: 48990731-48990731 on Build GRCh38
rs370728780 Chr.2: 48990654-48990654 on Build GRCh38
rs1015877896 Chr.2: 48990752-48990752 on Build GRCh38
rs375329429 Chr.2: 48990603-48990603 on Build GRCh38
rs2072486 Chr.2: 48990777-48990777 on Build GRCh38
rs200504733 Chr.2: 48990556-48990556 on Build GRCh38
rs753141201 Chr.2: 48990646-48990646 on Build GRCh38
rs762945198 Chr.2: 48990580-48990580 on Build GRCh38
rs368308512 Chr.2: 48990827-48990827 on Build GRCh38
rs759833133 Chr.2: 48990766-48990766 on Build GRCh38
rs200033776 Chr.2: 48990892-48990892 on Build GRCh38
rs933531596 Chr.2: 48990578-48990578 on Build GRCh38
rs745386710 Chr.2: 48990567-48990567 on Build GRCh38
rs1019203579 Chr.2: 48990854-48990854 on Build GRCh38
rs35605088 Chr.2: 48990554-48990555 on Build GRCh38
rs377222977 Chr.2: 48990650-48990650 on Build GRCh38
rs754335673 Chr.2: 48990619-48990619 on Build GRCh38
rs776251039 Chr.2: 48990519-48990519 on Build GRCh38
rs775484603 Chr.2: 48990579-48990579 on Build GRCh38
rs768605354 Chr.2: 48990680-48990680 on Build GRCh38
rs1055076327 Chr.2: 48990669-48990669 on Build GRCh38
rs750208579 Chr.2: 48990876-48990876 on Build GRCh38
rs367799616 Chr.2: 48990487-48990487 on Build GRCh38
rs963955470 Chr.2: 48990546-48990546 on Build GRCh38
rs148815172 Chr.2: 48990613-48990613 on Build GRCh38
rs886311092 Chr.2: 48990877-48990877 on Build GRCh38
rs181851787 Chr.2: 48990903-48990903 on Build GRCh38
rs755587702 Chr.2: 48990637-48990637 on Build GRCh38
rs371082462 Chr.2: 48990529-48990529 on Build GRCh38
rs775079724 Chr.2: 48990524-48990524 on Build GRCh38
rs751776651 Chr.2: 48990533-48990533 on Build GRCh38
rs916478839 Chr.2: 48990849-48990849 on Build GRCh38
rs767760320 Chr.2: 48990609-48990609 on Build GRCh38
rs775963095 Chr.2: 48990565-48990565 on Build GRCh38
rs574746126 Chr.2: 48990649-48990649 on Build GRCh38
rs146638732 Chr.2: 48990520-48990520 on Build GRCh38
rs756541653 Chr.2: 48990543-48990543 on Build GRCh38
rs1056214957 Chr.2: 48990839-48990839 on Build GRCh38
rs1013345621 Chr.2: 48990700-48990700 on Build GRCh38
rs139730582 Chr.2: 48990506-48990506 on Build GRCh38
rs930328553 Chr.2: 48990760-48990760 on Build GRCh38
rs530648020 Chr.2: 48990687-48990687 on Build GRCh38
rs749561916 Chr.2: 48990550-48990550 on Build GRCh38
rs895020904 Chr.2: 48990502-48990502 on Build GRCh38
rs184958361 Chr.2: 48990784-48990784 on Build GRCh38
rs374719504 Chr.2: 48990658-48990658 on Build GRCh38
rs535746659 Chr.2: 48990467-48990467 on Build GRCh38
rs13019186 Chr.2: 48990851-48990851 on Build GRCh38
rs768356076 Chr.2: 48990532-48990533 on Build GRCh38
rs2072485 Chr.2: 48990855-48990855 on Build GRCh38
rs529507576 Chr.2: 48990805-48990805 on Build GRCh38
rs868553921 Chr.2: 48990886-48990886 on Build GRCh38
rs769559580 Chr.2: 48990575-48990575 on Build GRCh38
rs978880963 Chr.2: 48990811-48990811 on Build GRCh38
rs963099792 Chr.2: 48990762-48990762 on Build GRCh38
rs1029217213 Chr.2: 48990771-48990771 on Build GRCh38
rs121909665 Chr.2: 48990629-48990629 on Build GRCh38
rs776844586 Chr.2: 48990633-48990633 on Build GRCh38
rs903730200 Chr.2: 48990478-48990478 on Build GRCh38
rs372434976 Chr.2: 48990522-48990522 on Build GRCh38
rs972299287 Chr.2: 48990887-48990887 on Build GRCh38
rs761976313 Chr.2: 48990602-48990602 on Build GRCh38
rs755386201 Chr.2: 48990552-48990552 on Build GRCh38
rs367722560 Chr.2: 48990678-48990678 on Build GRCh38
rs563446551 Chr.2: 48990657-48990657 on Build GRCh38
rs763934518 Chr.2: 48990530-48990530 on Build GRCh38
rs779369514 Chr.2: 48990643-48990643 on Build GRCh38
rs201387284 Chr.2: 48990604-48990604 on Build GRCh38
rs773478237 Chr.2: 48990476-48990476 on Build GRCh38
rs557996813 Chr.2: 48990509-48990509 on Build GRCh38
rs1033922918 Chr.2: 48990490-48990490 on Build GRCh38
rs1024764753 Chr.2: 48990732-48990732 on Build GRCh38
rs376735786 Chr.2: 48990536-48990536 on Build GRCh38
rs1004621751 Chr.2: 48990751-48990751 on Build GRCh38
rs74809436 Chr.2: 48990770-48990770 on Build GRCh38

View Full Product Details