Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 54253390-54253902 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ACYP2
Entrez Gene ID
98
Gene Name
acylphosphatase 2
Gene Aliases
ACYM, ACYP
UniGene
Hs.516173
Gene Chromosome Location
Chr.2: 53970838-54305300 on Build GRCh38
Transcripts
NM_001320586.1 NM_001320587.1 NM_001320588.1 NM_001320589.1 NM_138448.3 XM_017005411.1 XM_017005412.1 XM_017005413.1 NM_001003937.2

Gene Symbol
TSPYL6
Entrez Gene ID
388951
Gene Name
TSPY like 6
Gene Aliases
-
UniGene
Hs.620508
Gene Chromosome Location
Chr.2: 54253178-54256272 on Build GRCh38
Transcripts
NM_001320586.1 NM_001320587.1 NM_001320588.1 NM_001320589.1 NM_138448.3 XM_017005411.1 XM_017005412.1 XM_017005413.1 NM_001003937.2

SNP Detail

SNP ID Chromosome Location
rs141290724 Chr.2: 54253864-54253864 on Build GRCh38
rs534406022 Chr.2: 54253604-54253604 on Build GRCh38
rs749974559 Chr.2: 54253771-54253771 on Build GRCh38
rs141063292 Chr.2: 54253688-54253688 on Build GRCh38
rs1005443333 Chr.2: 54253581-54253581 on Build GRCh38
rs145203363 Chr.2: 54253449-54253449 on Build GRCh38
rs113162319 Chr.2: 54253566-54253566 on Build GRCh38
rs892537338 Chr.2: 54253624-54253624 on Build GRCh38
rs148244016 Chr.2: 54253834-54253834 on Build GRCh38
rs76327822 Chr.2: 54253667-54253667 on Build GRCh38
rs181518899 Chr.2: 54253734-54253734 on Build GRCh38
rs936264984 Chr.2: 54253437-54253437 on Build GRCh38
rs984587690 Chr.2: 54253728-54253728 on Build GRCh38
rs998184338 Chr.2: 54253609-54253609 on Build GRCh38
rs984914196 Chr.2: 54253497-54253497 on Build GRCh38
rs751255840 Chr.2: 54253709-54253709 on Build GRCh38
rs906322724 Chr.2: 54253551-54253551 on Build GRCh38
rs964021301 Chr.2: 54253585-54253587 on Build GRCh38
rs767036283 Chr.2: 54253757-54253757 on Build GRCh38
rs1030156377 Chr.2: 54253616-54253616 on Build GRCh38
rs548602692 Chr.2: 54253736-54253736 on Build GRCh38
rs947754076 Chr.2: 54253499-54253499 on Build GRCh38
rs903984769 Chr.2: 54253596-54253596 on Build GRCh38
rs531852261 Chr.2: 54253614-54253614 on Build GRCh38
rs1045235620 Chr.2: 54253594-54253594 on Build GRCh38
rs1018200380 Chr.2: 54253584-54253585 on Build GRCh38
rs1039599083 Chr.2: 54253825-54253825 on Build GRCh38
rs916213248 Chr.2: 54253705-54253705 on Build GRCh38
rs566668855 Chr.2: 54253419-54253424 on Build GRCh38
rs998132122 Chr.2: 54253859-54253859 on Build GRCh38
rs368513395 Chr.2: 54253600-54253600 on Build GRCh38
rs113556840 Chr.2: 54253554-54253554 on Build GRCh38
rs907623460 Chr.2: 54253516-54253516 on Build GRCh38
rs561415479 Chr.2: 54253426-54253426 on Build GRCh38
rs1047314291 Chr.2: 54253794-54253794 on Build GRCh38
rs977978972 Chr.2: 54253649-54253649 on Build GRCh38
rs868586921 Chr.2: 54253872-54253872 on Build GRCh38
rs948279831 Chr.2: 54253575-54253575 on Build GRCh38
rs1029689587 Chr.2: 54253875-54253875 on Build GRCh38
rs13005618 Chr.2: 54253745-54253745 on Build GRCh38
rs901780275 Chr.2: 54253645-54253645 on Build GRCh38
rs750281581 Chr.2: 54253504-54253504 on Build GRCh38
rs991772231 Chr.2: 54253556-54253556 on Build GRCh38
rs566190779 Chr.2: 54253706-54253706 on Build GRCh38
rs201962107 Chr.2: 54253423-54253426 on Build GRCh38
rs991750445 Chr.2: 54253694-54253694 on Build GRCh38
rs73934418 Chr.2: 54253636-54253636 on Build GRCh38
rs888292641 Chr.2: 54253800-54253800 on Build GRCh38
rs1019600534 Chr.2: 54253642-54253642 on Build GRCh38

View Full Product Details