Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 196584407-196584914 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FBXO45
Entrez Gene ID
200933
Gene Name
F-box protein 45
Gene Aliases
Fbx45
UniGene
Hs.169815
Gene Chromosome Location
Chr.3: 196568854-196589059 on Build GRCh38
Transcripts
NM_001105573.1

SNP Detail

SNP ID Chromosome Location
rs58204718 Chr.3: 196584453-196584456 on Build GRCh38
rs751329729 Chr.3: 196584463-196584463 on Build GRCh38
rs957789864 Chr.3: 196584719-196584719 on Build GRCh38
rs577930726 Chr.3: 196584558-196584558 on Build GRCh38
rs777172885 Chr.3: 196584733-196584733 on Build GRCh38
rs567298679 Chr.3: 196584476-196584476 on Build GRCh38
rs1022896508 Chr.3: 196584518-196584518 on Build GRCh38
rs538828421 Chr.3: 196584609-196584609 on Build GRCh38
rs920557445 Chr.3: 196584868-196584868 on Build GRCh38
rs879444743 Chr.3: 196584616-196584616 on Build GRCh38
rs899729563 Chr.3: 196584541-196584541 on Build GRCh38
rs1040995438 Chr.3: 196584535-196584535 on Build GRCh38
rs913880135 Chr.3: 196584710-196584710 on Build GRCh38
rs139927804 Chr.3: 196584719-196584719 on Build GRCh38
rs933335453 Chr.3: 196584711-196584712 on Build GRCh38
rs143653920 Chr.3: 196584622-196584622 on Build GRCh38
rs546690133 Chr.3: 196584475-196584475 on Build GRCh38
rs969878823 Chr.3: 196584554-196584554 on Build GRCh38
rs982672856 Chr.3: 196584776-196584776 on Build GRCh38
rs1008576296 Chr.3: 196584510-196584510 on Build GRCh38
rs1021557925 Chr.3: 196584724-196584724 on Build GRCh38
rs973411411 Chr.3: 196584846-196584846 on Build GRCh38
rs895451055 Chr.3: 196584888-196584889 on Build GRCh38
rs553208888 Chr.3: 196584522-196584522 on Build GRCh38
rs534323236 Chr.3: 196584511-196584511 on Build GRCh38
rs11298661 Chr.3: 196584875-196584875 on Build GRCh38
rs397877731 Chr.3: 196584886-196584886 on Build GRCh38
rs926937536 Chr.3: 196584787-196584787 on Build GRCh38
rs978999668 Chr.3: 196584594-196584594 on Build GRCh38
rs988207828 Chr.3: 196584671-196584671 on Build GRCh38
rs1026084403 Chr.3: 196584886-196584886 on Build GRCh38
rs754270345 Chr.3: 196584452-196584453 on Build GRCh38
rs929396013 Chr.3: 196584873-196584873 on Build GRCh38
rs748915848 Chr.3: 196584838-196584838 on Build GRCh38
rs993277088 Chr.3: 196584453-196584453 on Build GRCh38
rs140139122 Chr.3: 196584437-196584437 on Build GRCh38
rs567933145 Chr.3: 196584482-196584482 on Build GRCh38
rs150852815 Chr.3: 196584721-196584721 on Build GRCh38
rs1047342171 Chr.3: 196584462-196584462 on Build GRCh38
rs200261609 Chr.3: 196584887-196584887 on Build GRCh38
rs959606291 Chr.3: 196584791-196584791 on Build GRCh38
rs747630648 Chr.3: 196584610-196584610 on Build GRCh38
rs996508382 Chr.3: 196584582-196584582 on Build GRCh38
rs889366247 Chr.3: 196584753-196584760 on Build GRCh38
rs943670727 Chr.3: 196584794-196584794 on Build GRCh38

View Full Product Details