Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 76214231-76214736 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FAM47E
Entrez Gene ID
100129583
Gene Name
family with sequence similarity 47 member E
Gene Aliases
-
UniGene
Hs.720509
Gene Chromosome Location
Chr.4: 76214040-76283783 on Build GRCh38
Transcripts
NM_001242936.1

SNP Detail

SNP ID Chromosome Location
rs938541924 Chr.4: 76214712-76214712 on Build GRCh38
rs890597778 Chr.4: 76214488-76214488 on Build GRCh38
rs991636636 Chr.4: 76214462-76214462 on Build GRCh38
rs948756040 Chr.4: 76214392-76214392 on Build GRCh38
rs888883924 Chr.4: 76214686-76214686 on Build GRCh38
rs775418437 Chr.4: 76214291-76214291 on Build GRCh38
rs965970319 Chr.4: 76214317-76214317 on Build GRCh38
rs769224839 Chr.4: 76214278-76214278 on Build GRCh38
rs567058172 Chr.4: 76214441-76214441 on Build GRCh38
rs1054175085 Chr.4: 76214659-76214659 on Build GRCh38
rs993956008 Chr.4: 76214461-76214461 on Build GRCh38
rs575560014 Chr.4: 76214522-76214522 on Build GRCh38
rs1026497558 Chr.4: 76214482-76214482 on Build GRCh38
rs763823249 Chr.4: 76214352-76214352 on Build GRCh38
rs964953364 Chr.4: 76214520-76214520 on Build GRCh38
rs938629746 Chr.4: 76214335-76214335 on Build GRCh38
rs909978400 Chr.4: 76214371-76214371 on Build GRCh38
rs1017900843 Chr.4: 76214512-76214512 on Build GRCh38
rs924366457 Chr.4: 76214710-76214710 on Build GRCh38
rs116533523 Chr.4: 76214634-76214634 on Build GRCh38
rs369710301 Chr.4: 76214365-76214365 on Build GRCh38
rs531439252 Chr.4: 76214492-76214492 on Build GRCh38
rs536634073 Chr.4: 76214568-76214568 on Build GRCh38
rs993849535 Chr.4: 76214322-76214322 on Build GRCh38
rs924301108 Chr.4: 76214511-76214511 on Build GRCh38
rs368794524 Chr.4: 76214427-76214427 on Build GRCh38
rs975075672 Chr.4: 76214521-76214521 on Build GRCh38
rs183858810 Chr.4: 76214367-76214367 on Build GRCh38
rs866676793 Chr.4: 76214359-76214359 on Build GRCh38
rs567640375 Chr.4: 76214354-76214354 on Build GRCh38
rs977550889 Chr.4: 76214701-76214701 on Build GRCh38
rs899251491 Chr.4: 76214459-76214459 on Build GRCh38
rs191623430 Chr.4: 76214650-76214650 on Build GRCh38
rs551025314 Chr.4: 76214293-76214293 on Build GRCh38
rs188825460 Chr.4: 76214513-76214513 on Build GRCh38
rs142796106 Chr.4: 76214490-76214490 on Build GRCh38

View Full Product Details