Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 29723539-29724078 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HLA-F
Entrez Gene ID
3134
Gene Name
major histocompatibility complex, class I, F
Gene Aliases
CDA12, HLA-5.4, HLA-CDA12, HLAF
UniGene
Hs.519972
Gene Chromosome Location
Chr.6: 29723340-29740355 on Build GRCh38
Transcripts
NM_001098478.1 NM_001098479.1 NM_018950.2 XM_011514564.1 XM_017010810.1 XM_017010811.1 XM_017010812.1 XM_017010813.1 XM_017010814.1 XM_017010815.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4006245 c.287G>C p.R96P Chr.6: 29723880-29723880 on Build GRCh38
5029995 c.148G>C p.D50H Chr.6: 29723741-29723741 on Build GRCh38
6425467 c.94G>A p.A32T Chr.6: 29723687-29723687 on Build GRCh38
3861313 c.166C>T p.R56W Chr.6: 29723759-29723759 on Build GRCh38
6404061 c.175A>G p.S59G Chr.6: 29723768-29723768 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs769737712 Chr.6: 29723840-29723840 on Build GRCh38
rs751837865 Chr.6: 29723901-29723901 on Build GRCh38
rs2076178 Chr.6: 29723997-29723997 on Build GRCh38
rs386698419 Chr.6: 29723613-29723614 on Build GRCh38
rs547518914 Chr.6: 29723966-29723966 on Build GRCh38
rs767978557 Chr.6: 29723672-29723672 on Build GRCh38
rs776120457 Chr.6: 29723694-29723704 on Build GRCh38
rs753820152 Chr.6: 29723670-29723670 on Build GRCh38
rs1044172482 Chr.6: 29723982-29723982 on Build GRCh38
rs373876297 Chr.6: 29723608-29723608 on Build GRCh38
rs775877655 Chr.6: 29723968-29723968 on Build GRCh38
rs61739958 Chr.6: 29723668-29723668 on Build GRCh38
rs112396609 Chr.6: 29723610-29723611 on Build GRCh38
rs948413381 Chr.6: 29723744-29723744 on Build GRCh38
rs771497766 Chr.6: 29723743-29723743 on Build GRCh38
rs776996820 Chr.6: 29723827-29723827 on Build GRCh38
rs748689916 Chr.6: 29723715-29723715 on Build GRCh38
rs938308518 Chr.6: 29723993-29723993 on Build GRCh38
rs779226984 Chr.6: 29723577-29723577 on Build GRCh38
rs1040621783 Chr.6: 29723649-29723649 on Build GRCh38
rs746722149 Chr.6: 29723961-29723961 on Build GRCh38
rs757973145 Chr.6: 29724005-29724005 on Build GRCh38
rs752186602 Chr.6: 29723940-29723940 on Build GRCh38
rs201474292 Chr.6: 29723781-29723781 on Build GRCh38
rs781002327 Chr.6: 29723701-29723701 on Build GRCh38
rs761311405 Chr.6: 29723665-29723665 on Build GRCh38
rs774066513 Chr.6: 29723632-29723632 on Build GRCh38
rs748864874 Chr.6: 29723624-29723624 on Build GRCh38
rs868002997 Chr.6: 29724056-29724056 on Build GRCh38
rs764335226 Chr.6: 29723765-29723765 on Build GRCh38
rs755105818 Chr.6: 29723571-29723571 on Build GRCh38
rs756258830 Chr.6: 29723562-29723562 on Build GRCh38
rs916875032 Chr.6: 29724026-29724026 on Build GRCh38
rs760126750 Chr.6: 29723832-29723832 on Build GRCh38
rs939621676 Chr.6: 29723711-29723711 on Build GRCh38
rs774935556 Chr.6: 29723623-29723623 on Build GRCh38
rs753017432 Chr.6: 29723687-29723687 on Build GRCh38
rs386698420 Chr.6: 29723995-29723997 on Build GRCh38
rs1043946815 Chr.6: 29724042-29724042 on Build GRCh38
rs759253001 Chr.6: 29723994-29723994 on Build GRCh38
rs778745328 Chr.6: 29723938-29723938 on Build GRCh38
rs768270857 Chr.6: 29723716-29723716 on Build GRCh38
rs750155724 Chr.6: 29723773-29723773 on Build GRCh38
rs569508739 Chr.6: 29723746-29723746 on Build GRCh38
rs202160506 Chr.6: 29723790-29723790 on Build GRCh38
rs17875380 Chr.6: 29723805-29723805 on Build GRCh38
rs1021267082 Chr.6: 29723565-29723565 on Build GRCh38
rs778536918 Chr.6: 29723797-29723797 on Build GRCh38
rs117540842 Chr.6: 29723857-29723857 on Build GRCh38
rs766604838 Chr.6: 29723567-29723567 on Build GRCh38
rs1042826143 Chr.6: 29723783-29723783 on Build GRCh38
rs906871831 Chr.6: 29723804-29723804 on Build GRCh38
rs753698900 Chr.6: 29723924-29723924 on Build GRCh38
rs778428087 Chr.6: 29723717-29723717 on Build GRCh38
rs751031382 Chr.6: 29723561-29723561 on Build GRCh38
rs762071299 Chr.6: 29723637-29723637 on Build GRCh38
rs17875381 Chr.6: 29724023-29724023 on Build GRCh38
rs2076180 Chr.6: 29723975-29723975 on Build GRCh38
rs2076182 Chr.6: 29723936-29723936 on Build GRCh38
rs755090200 Chr.6: 29723674-29723674 on Build GRCh38
rs556086260 Chr.6: 29723978-29723978 on Build GRCh38
rs201075212 Chr.6: 29723752-29723752 on Build GRCh38
rs762075292 Chr.6: 29723617-29723619 on Build GRCh38
rs781655152 Chr.6: 29723965-29723965 on Build GRCh38
rs369106559 Chr.6: 29723630-29723630 on Build GRCh38
rs67064701 Chr.6: 29723614-29723614 on Build GRCh38
rs776444345 Chr.6: 29723992-29723992 on Build GRCh38
rs764247330 Chr.6: 29723894-29723894 on Build GRCh38
rs879786089 Chr.6: 29723613-29723614 on Build GRCh38
rs756008669 Chr.6: 29723705-29723705 on Build GRCh38
rs200319715 Chr.6: 29723759-29723759 on Build GRCh38
rs531915649 Chr.6: 29723764-29723764 on Build GRCh38
rs756994939 Chr.6: 29723688-29723688 on Build GRCh38
rs1049243043 Chr.6: 29723690-29723690 on Build GRCh38
rs561672947 Chr.6: 29723586-29723586 on Build GRCh38
rs759221105 Chr.6: 29723991-29723992 on Build GRCh38
rs1053224159 Chr.6: 29723849-29723849 on Build GRCh38
rs2076179 Chr.6: 29723995-29723995 on Build GRCh38
rs754416000 Chr.6: 29723796-29723796 on Build GRCh38
rs1632955 Chr.6: 29723613-29723613 on Build GRCh38
rs764490901 Chr.6: 29723944-29723944 on Build GRCh38
rs1021837809 Chr.6: 29724033-29724033 on Build GRCh38
rs772540636 Chr.6: 29723728-29723728 on Build GRCh38
rs2076181 Chr.6: 29723967-29723967 on Build GRCh38
rs948436969 Chr.6: 29723981-29723981 on Build GRCh38
rs1031579943 Chr.6: 29723611-29723611 on Build GRCh38
rs998967257 Chr.6: 29723584-29723584 on Build GRCh38
rs756883390 Chr.6: 29724036-29724036 on Build GRCh38
rs780715196 Chr.6: 29724046-29724046 on Build GRCh38
rs761151916 Chr.6: 29723733-29723733 on Build GRCh38
rs777549588 Chr.6: 29723958-29723958 on Build GRCh38
rs765314116 Chr.6: 29723680-29723680 on Build GRCh38
rs998525722 Chr.6: 29723862-29723862 on Build GRCh38
rs766238291 Chr.6: 29723923-29723923 on Build GRCh38
rs767657492 Chr.6: 29723991-29723991 on Build GRCh38
rs909845284 Chr.6: 29723669-29723669 on Build GRCh38
rs77329802 Chr.6: 29723799-29723799 on Build GRCh38
rs751231868 Chr.6: 29724027-29724027 on Build GRCh38
rs762072401 Chr.6: 29723990-29723990 on Build GRCh38
rs773827023 Chr.6: 29723647-29723647 on Build GRCh38
rs755920765 Chr.6: 29723785-29723785 on Build GRCh38
rs765422224 Chr.6: 29723749-29723749 on Build GRCh38
rs377421873 Chr.6: 29723788-29723788 on Build GRCh38
rs779975618 Chr.6: 29723706-29723706 on Build GRCh38
rs746189322 Chr.6: 29724047-29724047 on Build GRCh38
rs756496935 Chr.6: 29724057-29724057 on Build GRCh38
rs202014717 Chr.6: 29723756-29723756 on Build GRCh38
rs754089399 Chr.6: 29723568-29723568 on Build GRCh38
rs946859710 Chr.6: 29724039-29724039 on Build GRCh38
rs747319246 Chr.6: 29723816-29723816 on Build GRCh38
rs757662148 Chr.6: 29723963-29723963 on Build GRCh38
rs746711511 Chr.6: 29723578-29723578 on Build GRCh38
rs768063472 Chr.6: 29723641-29723641 on Build GRCh38
rs745687606 Chr.6: 29723702-29723702 on Build GRCh38
rs199658940 Chr.6: 29723910-29723910 on Build GRCh38
rs774707298 Chr.6: 29723613-29723614 on Build GRCh38
rs747877067 Chr.6: 29723718-29723718 on Build GRCh38
rs1632954 Chr.6: 29723614-29723614 on Build GRCh38
rs773661638 Chr.6: 29723729-29723729 on Build GRCh38
rs771477014 Chr.6: 29723825-29723825 on Build GRCh38

View Full Product Details