Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 31538685-31539191 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ATP6V1G2-DDX39B
Entrez Gene ID
100532737
Gene Name
ATP6V1G2-DDX39B readthrough (NMD candidate)
Gene Aliases
-
UniGene
Hs.254042
Gene Chromosome Location
Chr.6: 31530219-31546848 on Build GRCh38
Transcripts
NR_037853.1 NM_004640.6 NM_080598.5 NR_037852.1

Gene Symbol
DDX39B
Entrez Gene ID
7919
Gene Name
DEAD-box helicase 39B
Gene Aliases
BAT1, D6S81E, UAP56
UniGene
Hs.254042
Gene Chromosome Location
Chr.6: 31530219-31542475 on Build GRCh38
Transcripts
NR_037853.1 NM_004640.6 NM_080598.5 NR_037852.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7088910 c.437G>T p.R146L Chr.6: 31538827-31538827 on Build GRCh38
5357748 c.232T>C p.L78L Chr.6: 31538822-31538822 on Build GRCh38
7000210 c.365C>G p.T122S Chr.6: 31538830-31538830 on Build GRCh38
7088906 c.368G>T p.R123L Chr.6: 31538827-31538827 on Build GRCh38
5021518 c.432A>G p.V144V Chr.6: 31538847-31538847 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs747626075 Chr.6: 31538864-31538864 on Build GRCh38
rs774148327 Chr.6: 31538946-31538946 on Build GRCh38
rs749515605 Chr.6: 31539008-31539008 on Build GRCh38
rs776457628 Chr.6: 31539014-31539014 on Build GRCh38
rs774638800 Chr.6: 31538957-31538957 on Build GRCh38
rs531048281 Chr.6: 31538853-31538853 on Build GRCh38
rs541059804 Chr.6: 31539017-31539017 on Build GRCh38
rs775195232 Chr.6: 31539108-31539108 on Build GRCh38
rs112327138 Chr.6: 31539004-31539004 on Build GRCh38
rs754602970 Chr.6: 31538861-31538861 on Build GRCh38
rs372670994 Chr.6: 31538874-31538874 on Build GRCh38
rs770863639 Chr.6: 31539013-31539013 on Build GRCh38
rs760677256 Chr.6: 31538959-31538959 on Build GRCh38
rs758090991 Chr.6: 31538991-31538991 on Build GRCh38
rs2516393 Chr.6: 31538967-31538967 on Build GRCh38
rs145600883 Chr.6: 31539063-31539063 on Build GRCh38
rs759787531 Chr.6: 31538714-31538714 on Build GRCh38
rs1129640 Chr.6: 31538847-31538847 on Build GRCh38
rs756737471 Chr.6: 31539059-31539059 on Build GRCh38
rs553381734 Chr.6: 31538909-31538909 on Build GRCh38
rs766929998 Chr.6: 31539158-31539158 on Build GRCh38
rs751026616 Chr.6: 31539057-31539057 on Build GRCh38
rs2523511 Chr.6: 31539077-31539077 on Build GRCh38
rs779310325 Chr.6: 31539083-31539083 on Build GRCh38
rs533642852 Chr.6: 31538992-31538992 on Build GRCh38
rs745792250 Chr.6: 31539100-31539100 on Build GRCh38
rs575699700 Chr.6: 31538981-31538981 on Build GRCh38
rs753604795 Chr.6: 31538971-31538971 on Build GRCh38
rs781578661 Chr.6: 31539000-31539000 on Build GRCh38
rs763152729 Chr.6: 31538791-31538791 on Build GRCh38
rs536394576 Chr.6: 31539004-31539007 on Build GRCh38
rs763737896 Chr.6: 31539051-31539051 on Build GRCh38
rs563488082 Chr.6: 31539079-31539079 on Build GRCh38
rs913740038 Chr.6: 31538950-31538950 on Build GRCh38
rs568771188 Chr.6: 31538912-31538912 on Build GRCh38
rs763684827 Chr.6: 31539140-31539140 on Build GRCh38
rs139648127 Chr.6: 31538998-31538998 on Build GRCh38
rs150991114 Chr.6: 31539153-31539153 on Build GRCh38
rs887296773 Chr.6: 31539034-31539034 on Build GRCh38
rs748191884 Chr.6: 31539091-31539091 on Build GRCh38
rs762530649 Chr.6: 31539043-31539043 on Build GRCh38
rs770032337 Chr.6: 31539003-31539003 on Build GRCh38
rs770015146 Chr.6: 31538916-31538916 on Build GRCh38
rs755185820 Chr.6: 31539080-31539080 on Build GRCh38
rs535064717 Chr.6: 31538889-31538889 on Build GRCh38
rs879229025 Chr.6: 31539119-31539119 on Build GRCh38
rs750862925 Chr.6: 31539072-31539072 on Build GRCh38
rs765387543 Chr.6: 31538717-31538717 on Build GRCh38
rs987069638 Chr.6: 31538787-31538787 on Build GRCh38
rs775595502 Chr.6: 31538945-31538945 on Build GRCh38
rs746191618 Chr.6: 31539002-31539002 on Build GRCh38
rs757927750 Chr.6: 31538873-31538873 on Build GRCh38
rs933102807 Chr.6: 31538776-31538776 on Build GRCh38
rs752559880 Chr.6: 31539025-31539025 on Build GRCh38
rs905442565 Chr.6: 31539096-31539096 on Build GRCh38
rs1041720596 Chr.6: 31538960-31538960 on Build GRCh38
rs71563350 Chr.6: 31538997-31538997 on Build GRCh38
rs116766154 Chr.6: 31538868-31538868 on Build GRCh38
rs3219187 Chr.6: 31539098-31539098 on Build GRCh38
rs776904787 Chr.6: 31538712-31538712 on Build GRCh38
rs2523512 Chr.6: 31539024-31539024 on Build GRCh38
rs762764612 Chr.6: 31539112-31539112 on Build GRCh38
rs774516094 Chr.6: 31539012-31539012 on Build GRCh38
rs752036348 Chr.6: 31538947-31538947 on Build GRCh38
rs752326949 Chr.6: 31538983-31538983 on Build GRCh38
rs750362496 Chr.6: 31538822-31538822 on Build GRCh38
rs868060345 Chr.6: 31538890-31538890 on Build GRCh38
rs773967863 Chr.6: 31539142-31539142 on Build GRCh38
rs747487630 Chr.6: 31538893-31538893 on Build GRCh38
rs769831448 Chr.6: 31539103-31539103 on Build GRCh38
rs2071596 Chr.6: 31538914-31538914 on Build GRCh38
rs775570418 Chr.6: 31538777-31538777 on Build GRCh38
rs756160298 Chr.6: 31538830-31538830 on Build GRCh38
rs767511532 Chr.6: 31538802-31538802 on Build GRCh38
rs34985217 Chr.6: 31538732-31538733 on Build GRCh38
rs933208 Chr.6: 31538871-31538871 on Build GRCh38
rs750567223 Chr.6: 31539099-31539099 on Build GRCh38
rs756709512 Chr.6: 31539074-31539074 on Build GRCh38
rs750007290 Chr.6: 31539167-31539167 on Build GRCh38

View Full Product Details