Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 1092797-1093288 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C7orf50
Entrez Gene ID
84310
Gene Name
chromosome 7 open reading frame 50
Gene Aliases
YCR016W
UniGene
Hs.653258
Gene Chromosome Location
Chr.7: 978485-1138291 on Build GRCh38
Transcripts
NM_001134395.1 NM_001134396.1 NM_001318252.1 NM_032350.5 XM_011515581.2 XM_011515582.2 XM_011515583.2 XM_011515584.2 XM_017012720.1 XM_017012721.1 XM_017012722.1 NM_001039966.1 NM_001098201.1 NM_001505.2

Gene Symbol
GPER1
Entrez Gene ID
2852
Gene Name
G protein-coupled estrogen receptor 1
Gene Aliases
CEPR, CMKRL2, DRY12, FEG-1, GPCR-Br, GPER, GPR30, LERGU, LERGU2, LyGPR, mER
UniGene
Hs.20961
Gene Chromosome Location
Chr.7: 1086807-1093815 on Build GRCh38
Transcripts
NM_001134395.1 NM_001134396.1 NM_001318252.1 NM_032350.5 XM_011515581.2 XM_011515582.2 XM_011515583.2 XM_011515584.2 XM_017012720.1 XM_017012721.1 XM_017012722.1 NM_001039966.1 NM_001098201.1 NM_001505.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5871592 c.1108A>G p.R370G Chr.7: 1092836-1092836 on Build GRCh38
212413 c.1078A>G p.I360V Chr.7: 1092806-1092806 on Build GRCh38
7225906 c.1092C>T p.T364T Chr.7: 1092820-1092820 on Build GRCh38
4687797 c.1100C>T p.S367L Chr.7: 1092828-1092828 on Build GRCh38
4564225 c.1071_1072GG>AA p.A358T Chr.7: 1092799-1092800 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs897354566 Chr.7: 1093197-1093197 on Build GRCh38
rs754374497 Chr.7: 1093087-1093087 on Build GRCh38
rs764632912 Chr.7: 1092909-1092909 on Build GRCh38
rs757659970 Chr.7: 1092856-1092856 on Build GRCh38
rs73050449 Chr.7: 1093049-1093049 on Build GRCh38
rs781390250 Chr.7: 1092860-1092860 on Build GRCh38
rs766714154 Chr.7: 1093186-1093186 on Build GRCh38
rs888688395 Chr.7: 1092978-1092978 on Build GRCh38
rs879772687 Chr.7: 1093200-1093200 on Build GRCh38
rs549783931 Chr.7: 1093093-1093093 on Build GRCh38
rs985035078 Chr.7: 1092825-1092825 on Build GRCh38
rs750580587 Chr.7: 1092928-1092928 on Build GRCh38
rs764896213 Chr.7: 1092846-1092846 on Build GRCh38
rs746767126 Chr.7: 1092881-1092881 on Build GRCh38
rs752074660 Chr.7: 1092917-1092917 on Build GRCh38
rs989315448 Chr.7: 1092921-1092921 on Build GRCh38
rs147941530 Chr.7: 1092832-1092832 on Build GRCh38
rs764773374 Chr.7: 1092894-1092894 on Build GRCh38
rs201675059 Chr.7: 1093188-1093189 on Build GRCh38
rs532402280 Chr.7: 1093131-1093131 on Build GRCh38
rs768111607 Chr.7: 1092974-1092974 on Build GRCh38
rs762521942 Chr.7: 1092973-1092973 on Build GRCh38
rs757290518 Chr.7: 1093074-1093074 on Build GRCh38
rs759682528 Chr.7: 1093218-1093218 on Build GRCh38
rs758976064 Chr.7: 1092850-1092850 on Build GRCh38
rs546531324 Chr.7: 1093148-1093163 on Build GRCh38
rs777759506 Chr.7: 1092946-1092946 on Build GRCh38
rs941641938 Chr.7: 1093168-1093168 on Build GRCh38
rs775046934 Chr.7: 1092965-1092965 on Build GRCh38
rs958134041 Chr.7: 1092908-1092908 on Build GRCh38
rs537081392 Chr.7: 1093201-1093201 on Build GRCh38
rs774362025 Chr.7: 1092934-1092934 on Build GRCh38
rs10246354 Chr.7: 1092820-1092820 on Build GRCh38
rs4266553 Chr.7: 1093035-1093035 on Build GRCh38
rs758589940 Chr.7: 1093245-1093245 on Build GRCh38
rs978657584 Chr.7: 1093263-1093263 on Build GRCh38
rs12702047 Chr.7: 1093084-1093084 on Build GRCh38
rs938038882 Chr.7: 1093156-1093156 on Build GRCh38
rs764582351 Chr.7: 1093018-1093018 on Build GRCh38
rs982195204 Chr.7: 1093139-1093139 on Build GRCh38
rs766104171 Chr.7: 1092830-1092830 on Build GRCh38
rs879752490 Chr.7: 1092985-1092995 on Build GRCh38
rs546093461 Chr.7: 1092953-1092953 on Build GRCh38
rs138657567 Chr.7: 1092847-1092847 on Build GRCh38
rs565822980 Chr.7: 1093141-1093141 on Build GRCh38
rs778871670 Chr.7: 1093047-1093047 on Build GRCh38
rs746149188 Chr.7: 1092863-1092863 on Build GRCh38
rs1017561665 Chr.7: 1093088-1093088 on Build GRCh38
rs760193133 Chr.7: 1092829-1092829 on Build GRCh38
rs2070118 Chr.7: 1092869-1092869 on Build GRCh38
rs993626351 Chr.7: 1092871-1092871 on Build GRCh38
rs771701149 Chr.7: 1092823-1092823 on Build GRCh38
rs943638671 Chr.7: 1092838-1092838 on Build GRCh38
rs369412581 Chr.7: 1092872-1092872 on Build GRCh38
rs559867351 Chr.7: 1092958-1092958 on Build GRCh38
rs1055346026 Chr.7: 1092964-1092964 on Build GRCh38
rs776856922 Chr.7: 1093217-1093217 on Build GRCh38
rs543407810 Chr.7: 1092851-1092851 on Build GRCh38
rs750804723 Chr.7: 1093028-1093028 on Build GRCh38
rs990770893 Chr.7: 1093158-1093158 on Build GRCh38
rs10235056 Chr.7: 1093112-1093112 on Build GRCh38
rs941555697 Chr.7: 1092987-1092987 on Build GRCh38
rs776408692 Chr.7: 1092886-1092886 on Build GRCh38
rs895152896 Chr.7: 1093127-1093127 on Build GRCh38
rs887524150 Chr.7: 1093164-1093164 on Build GRCh38
rs10235195 Chr.7: 1093241-1093241 on Build GRCh38
rs762315966 Chr.7: 1093012-1093012 on Build GRCh38
rs746842139 Chr.7: 1093242-1093242 on Build GRCh38
rs928050287 Chr.7: 1093151-1093151 on Build GRCh38
rs767832155 Chr.7: 1093025-1093025 on Build GRCh38
rs568193210 Chr.7: 1093173-1093173 on Build GRCh38
rs55965425 Chr.7: 1093092-1093092 on Build GRCh38
rs1026498200 Chr.7: 1092997-1092997 on Build GRCh38
rs1133041 Chr.7: 1093055-1093055 on Build GRCh38
rs759110202 Chr.7: 1092833-1092833 on Build GRCh38
rs552109980 Chr.7: 1093136-1093136 on Build GRCh38
rs770914933 Chr.7: 1092885-1092885 on Build GRCh38
rs187422106 Chr.7: 1093159-1093159 on Build GRCh38
rs993230861 Chr.7: 1092996-1092996 on Build GRCh38
rs761072854 Chr.7: 1093030-1093030 on Build GRCh38
rs775089213 Chr.7: 1092895-1092895 on Build GRCh38
rs767885379 Chr.7: 1092926-1092926 on Build GRCh38
rs762420628 Chr.7: 1092905-1092905 on Build GRCh38
rs577487581 Chr.7: 1093230-1093230 on Build GRCh38
rs753928156 Chr.7: 1092935-1092935 on Build GRCh38
rs1037216416 Chr.7: 1093026-1093026 on Build GRCh38
rs747000051 Chr.7: 1092948-1092948 on Build GRCh38
rs113499788 Chr.7: 1092925-1092925 on Build GRCh38
rs951849992 Chr.7: 1093001-1093001 on Build GRCh38
rs774784557 Chr.7: 1092988-1092988 on Build GRCh38
rs759274564 Chr.7: 1092888-1092888 on Build GRCh38
rs773068978 Chr.7: 1092874-1092874 on Build GRCh38
rs765275169 Chr.7: 1092823-1092824 on Build GRCh38
rs755050548 Chr.7: 1092939-1092939 on Build GRCh38
rs777481200 Chr.7: 1093232-1093232 on Build GRCh38
rs375524389 Chr.7: 1092828-1092828 on Build GRCh38
rs3840681 Chr.7: 1093190-1093191 on Build GRCh38
rs967015911 Chr.7: 1093251-1093251 on Build GRCh38
rs762429776 Chr.7: 1092837-1092837 on Build GRCh38
rs191002661 Chr.7: 1093240-1093240 on Build GRCh38
rs534498546 Chr.7: 1093143-1093143 on Build GRCh38
rs549801191 Chr.7: 1092929-1092929 on Build GRCh38
rs923438554 Chr.7: 1092868-1092868 on Build GRCh38
rs773514947 Chr.7: 1093036-1093036 on Build GRCh38
rs76649173 Chr.7: 1093189-1093190 on Build GRCh38
rs1017201028 Chr.7: 1093146-1093146 on Build GRCh38
rs761099745 Chr.7: 1093177-1093177 on Build GRCh38

View Full Product Details