Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 91264388-91264884 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FZD1
Entrez Gene ID
8321
Gene Name
frizzled class receptor 1
Gene Aliases
-
UniGene
Hs.94234
Gene Chromosome Location
Chr.7: 91264468-91268817 on Build GRCh38
Transcripts
NM_003505.1

SNP Detail

SNP ID Chromosome Location
rs768617963 Chr.7: 91264847-91264847 on Build GRCh38
rs947303234 Chr.7: 91264597-91264597 on Build GRCh38
rs1012395241 Chr.7: 91264512-91264512 on Build GRCh38
rs747030550 Chr.7: 91264438-91264438 on Build GRCh38
rs1041386069 Chr.7: 91264604-91264604 on Build GRCh38
rs2232158 Chr.7: 91264787-91264787 on Build GRCh38
rs894187953 Chr.7: 91264671-91264671 on Build GRCh38
rs961171628 Chr.7: 91264508-91264508 on Build GRCh38
rs573089082 Chr.7: 91264526-91264527 on Build GRCh38
rs992942724 Chr.7: 91264605-91264605 on Build GRCh38
rs956282431 Chr.7: 91264786-91264786 on Build GRCh38
rs2232159 Chr.7: 91264830-91264830 on Build GRCh38
rs1012430153 Chr.7: 91264673-91264673 on Build GRCh38
rs983109527 Chr.7: 91264818-91264818 on Build GRCh38
rs754265840 Chr.7: 91264432-91264432 on Build GRCh38
rs916976962 Chr.7: 91264517-91264517 on Build GRCh38
rs540842487 Chr.7: 91264545-91264545 on Build GRCh38
rs985891281 Chr.7: 91264572-91264572 on Build GRCh38
rs944558120 Chr.7: 91264659-91264659 on Build GRCh38
rs969092383 Chr.7: 91264560-91264561 on Build GRCh38
rs951722140 Chr.7: 91264816-91264816 on Build GRCh38
rs996951107 Chr.7: 91264707-91264708 on Build GRCh38
rs2232155 Chr.7: 91264528-91264528 on Build GRCh38
rs917019621 Chr.7: 91264612-91264612 on Build GRCh38
rs930281904 Chr.7: 91264647-91264647 on Build GRCh38
rs920281577 Chr.7: 91264642-91264642 on Build GRCh38
rs920321475 Chr.7: 91264815-91264815 on Build GRCh38
rs918434172 Chr.7: 91264578-91264578 on Build GRCh38
rs139801070 Chr.7: 91264728-91264728 on Build GRCh38
rs563572082 Chr.7: 91264574-91264574 on Build GRCh38
rs552011271 Chr.7: 91264724-91264724 on Build GRCh38
rs1024479503 Chr.7: 91264806-91264806 on Build GRCh38
rs1035220898 Chr.7: 91264489-91264489 on Build GRCh38
rs902832393 Chr.7: 91264620-91264620 on Build GRCh38
rs112870214 Chr.7: 91264726-91264726 on Build GRCh38
rs901251939 Chr.7: 91264550-91264550 on Build GRCh38
rs952241666 Chr.7: 91264537-91264537 on Build GRCh38
rs1030838837 Chr.7: 91264802-91264802 on Build GRCh38
rs575983091 Chr.7: 91264418-91264418 on Build GRCh38
rs373542546 Chr.7: 91264532-91264532 on Build GRCh38
rs947332790 Chr.7: 91264854-91264854 on Build GRCh38
rs2232156 Chr.7: 91264656-91264656 on Build GRCh38
rs992553794 Chr.7: 91264509-91264509 on Build GRCh38
rs563899229 Chr.7: 91264546-91264546 on Build GRCh38
rs1006460425 Chr.7: 91264712-91264712 on Build GRCh38
rs535084233 Chr.7: 91264692-91264692 on Build GRCh38
rs550045839 Chr.7: 91264560-91264560 on Build GRCh38
rs902970464 Chr.7: 91264453-91264453 on Build GRCh38
rs914525071 Chr.7: 91264582-91264582 on Build GRCh38
rs1028330196 Chr.7: 91264556-91264556 on Build GRCh38
rs953480878 Chr.7: 91264807-91264807 on Build GRCh38
rs2232157 Chr.7: 91264657-91264657 on Build GRCh38
rs923353224 Chr.7: 91264555-91264555 on Build GRCh38
rs1050241155 Chr.7: 91264707-91264707 on Build GRCh38
rs907829569 Chr.7: 91264829-91264829 on Build GRCh38
rs1003015745 Chr.7: 91264478-91264478 on Build GRCh38
rs1004314143 Chr.7: 91264416-91264416 on Build GRCh38
rs984825040 Chr.7: 91264542-91264542 on Build GRCh38
rs1047649932 Chr.7: 91264541-91264541 on Build GRCh38
rs533080826 Chr.7: 91264547-91264547 on Build GRCh38
rs1019687422 Chr.7: 91264417-91264417 on Build GRCh38
rs985965497 Chr.7: 91264812-91264812 on Build GRCh38
rs931809919 Chr.7: 91264563-91264563 on Build GRCh38
rs1029029184 Chr.7: 91264437-91264437 on Build GRCh38
rs904538885 Chr.7: 91264723-91264723 on Build GRCh38
rs566197644 Chr.7: 91264680-91264680 on Build GRCh38
rs976368542 Chr.7: 91264849-91264849 on Build GRCh38
rs541597656 Chr.7: 91264452-91264452 on Build GRCh38
rs561427027 Chr.7: 91264515-91264515 on Build GRCh38

View Full Product Details