Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 135135339-135135851 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AGBL3
Entrez Gene ID
340351
Gene Name
ATP/GTP binding protein like 3
Gene Aliases
CCP3
UniGene
Hs.648616
Gene Chromosome Location
Chr.7: 134985169-135135782 on Build GRCh38
Transcripts
NM_178563.3 XM_017012136.1 XM_017012137.1 XM_017012138.1 XM_017012139.1 XM_017012140.1 XM_017012141.1 XM_017012142.1 XM_017012143.1 NM_001305630.1 XM_017012595.1

Gene Symbol
C7orf49
Entrez Gene ID
78996
Gene Name
chromosome 7 open reading frame 49
Gene Aliases
MRI
UniGene
Hs.643113
Gene Chromosome Location
Chr.7: 135092303-135170826 on Build GRCh38
Transcripts
NM_178563.3 XM_017012136.1 XM_017012137.1 XM_017012138.1 XM_017012139.1 XM_017012140.1 XM_017012141.1 XM_017012142.1 XM_017012143.1 NM_001305630.1 XM_017012595.1

SNP Detail

SNP ID Chromosome Location
rs903666961 Chr.7: 135135788-135135788 on Build GRCh38
rs913812788 Chr.7: 135135707-135135707 on Build GRCh38
rs1007745569 Chr.7: 135135384-135135384 on Build GRCh38
rs758788204 Chr.7: 135135742-135135742 on Build GRCh38
rs144310396 Chr.7: 135135649-135135649 on Build GRCh38
rs992075412 Chr.7: 135135592-135135592 on Build GRCh38
rs997839471 Chr.7: 135135393-135135393 on Build GRCh38
rs1023024910 Chr.7: 135135638-135135638 on Build GRCh38
rs915149440 Chr.7: 135135779-135135779 on Build GRCh38
rs760079656 Chr.7: 135135547-135135547 on Build GRCh38
rs757332343 Chr.7: 135135777-135135777 on Build GRCh38
rs1056470826 Chr.7: 135135699-135135699 on Build GRCh38
rs118003802 Chr.7: 135135560-135135560 on Build GRCh38
rs529457071 Chr.7: 135135590-135135590 on Build GRCh38
rs946534278 Chr.7: 135135795-135135795 on Build GRCh38
rs61150470 Chr.7: 135135502-135135503 on Build GRCh38
rs899585620 Chr.7: 135135374-135135374 on Build GRCh38
rs538973057 Chr.7: 135135521-135135521 on Build GRCh38
rs1017373019 Chr.7: 135135467-135135467 on Build GRCh38
rs1043614632 Chr.7: 135135371-135135371 on Build GRCh38
rs939314200 Chr.7: 135135698-135135698 on Build GRCh38
rs1039419219 Chr.7: 135135368-135135368 on Build GRCh38
rs397720823 Chr.7: 135135503-135135504 on Build GRCh38
rs1011188747 Chr.7: 135135362-135135362 on Build GRCh38
rs1017236402 Chr.7: 135135566-135135566 on Build GRCh38
rs751721410 Chr.7: 135135752-135135752 on Build GRCh38
rs544919996 Chr.7: 135135408-135135408 on Build GRCh38
rs187406159 Chr.7: 135135533-135135533 on Build GRCh38
rs560571603 Chr.7: 135135563-135135563 on Build GRCh38
rs550872768 Chr.7: 135135652-135135652 on Build GRCh38
rs530692885 Chr.7: 135135651-135135651 on Build GRCh38
rs576823219 Chr.7: 135135650-135135650 on Build GRCh38
rs962903421 Chr.7: 135135573-135135573 on Build GRCh38
rs570277860 Chr.7: 135135723-135135723 on Build GRCh38
rs903593444 Chr.7: 135135380-135135380 on Build GRCh38
rs764321043 Chr.7: 135135741-135135741 on Build GRCh38
rs542646947 Chr.7: 135135635-135135635 on Build GRCh38
rs926739725 Chr.7: 135135687-135135687 on Build GRCh38

View Full Product Details