Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 42906120-42906560 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HOOK3
Entrez Gene ID
84376
Gene Name
hook microtubule-tethering protein 3
Gene Aliases
HK3
UniGene
Hs.162852
Gene Chromosome Location
Chr.8: 42896890-43030539 on Build GRCh38
Transcripts
NM_032410.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6681552 c.115G>A p.V39I Chr.8: 42906230-42906230 on Build GRCh38
4690945 c.87C>T p.C29C Chr.8: 42906202-42906202 on Build GRCh38
1254338 c.94G>A p.V32M Chr.8: 42906209-42906209 on Build GRCh38
4465163 c.136C>T p.Q46* Chr.8: 42906251-42906251 on Build GRCh38
176338 c.108G>A p.T36T Chr.8: 42906223-42906223 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs771885944 Chr.8: 42906150-42906151 on Build GRCh38
rs947284665 Chr.8: 42906422-42906422 on Build GRCh38
rs756183176 Chr.8: 42906252-42906252 on Build GRCh38
rs774064362 Chr.8: 42906195-42906195 on Build GRCh38
rs372716570 Chr.8: 42906209-42906209 on Build GRCh38
rs761755777 Chr.8: 42906295-42906295 on Build GRCh38
rs868370971 Chr.8: 42906205-42906205 on Build GRCh38
rs373940518 Chr.8: 42906152-42906152 on Build GRCh38
rs377485122 Chr.8: 42906288-42906288 on Build GRCh38
rs201879009 Chr.8: 42906150-42906150 on Build GRCh38
rs562485973 Chr.8: 42906349-42906349 on Build GRCh38
rs769810244 Chr.8: 42906182-42906182 on Build GRCh38
rs200599068 Chr.8: 42906146-42906146 on Build GRCh38
rs543280825 Chr.8: 42906344-42906344 on Build GRCh38
rs760175222 Chr.8: 42906298-42906298 on Build GRCh38
rs754269555 Chr.8: 42906212-42906212 on Build GRCh38
rs961891584 Chr.8: 42906329-42906329 on Build GRCh38
rs779934377 Chr.8: 42906173-42906173 on Build GRCh38
rs371480707 Chr.8: 42906148-42906148 on Build GRCh38
rs777837453 Chr.8: 42906261-42906261 on Build GRCh38
rs149930344 Chr.8: 42906164-42906165 on Build GRCh38
rs199518774 Chr.8: 42906156-42906156 on Build GRCh38
rs773176964 Chr.8: 42906183-42906183 on Build GRCh38
rs201927379 Chr.8: 42906154-42906154 on Build GRCh38
rs748151712 Chr.8: 42906178-42906178 on Build GRCh38
rs764677893 Chr.8: 42906459-42906459 on Build GRCh38
rs565224850 Chr.8: 42906374-42906374 on Build GRCh38
rs1042899713 Chr.8: 42906434-42906434 on Build GRCh38
rs200253742 Chr.8: 42906153-42906153 on Build GRCh38
rs946736371 Chr.8: 42906313-42906313 on Build GRCh38
rs766350098 Chr.8: 42906152-42906153 on Build GRCh38
rs369597018 Chr.8: 42906188-42906188 on Build GRCh38
rs745995976 Chr.8: 42906166-42906166 on Build GRCh38
rs938704339 Chr.8: 42906391-42906391 on Build GRCh38
rs201426292 Chr.8: 42906305-42906305 on Build GRCh38
rs530437477 Chr.8: 42906508-42906508 on Build GRCh38
rs747723958 Chr.8: 42906382-42906382 on Build GRCh38
rs983967793 Chr.8: 42906474-42906474 on Build GRCh38
rs770851876 Chr.8: 42906285-42906285 on Build GRCh38
rs764464349 Chr.8: 42906155-42906155 on Build GRCh38
rs751558260 Chr.8: 42906248-42906248 on Build GRCh38
rs771601050 Chr.8: 42906294-42906294 on Build GRCh38
rs757460486 Chr.8: 42906158-42906164 on Build GRCh38
rs750665963 Chr.8: 42906229-42906229 on Build GRCh38
rs547659688 Chr.8: 42906446-42906446 on Build GRCh38
rs765464370 Chr.8: 42906223-42906223 on Build GRCh38
rs531200065 Chr.8: 42906513-42906513 on Build GRCh38
rs913187430 Chr.8: 42906418-42906418 on Build GRCh38
rs989178654 Chr.8: 42906411-42906411 on Build GRCh38
rs780208796 Chr.8: 42906233-42906233 on Build GRCh38
rs80157780 Chr.8: 42906154-42906155 on Build GRCh38
rs780550607 Chr.8: 42906146-42906147 on Build GRCh38
rs6980873 Chr.8: 42906458-42906458 on Build GRCh38
rs760562693 Chr.8: 42906151-42906152 on Build GRCh38
rs201351323 Chr.8: 42906151-42906151 on Build GRCh38
rs764207823 Chr.8: 42906156-42906156 on Build GRCh38
rs140691200 Chr.8: 42906523-42906523 on Build GRCh38
rs764554162 Chr.8: 42906211-42906211 on Build GRCh38
rs759445264 Chr.8: 42906155-42906156 on Build GRCh38
rs575982507 Chr.8: 42906350-42906350 on Build GRCh38
rs1016557548 Chr.8: 42906483-42906483 on Build GRCh38
rs372731765 Chr.8: 42906208-42906208 on Build GRCh38
rs751630331 Chr.8: 42906157-42906158 on Build GRCh38
rs901032735 Chr.8: 42906373-42906373 on Build GRCh38
rs199564080 Chr.8: 42906144-42906145 on Build GRCh38
rs536191699 Chr.8: 42906197-42906197 on Build GRCh38
rs928674260 Chr.8: 42906377-42906377 on Build GRCh38
rs372977105 Chr.8: 42906269-42906269 on Build GRCh38
rs777137631 Chr.8: 42906308-42906308 on Build GRCh38
rs781718667 Chr.8: 42906147-42906147 on Build GRCh38
rs151329345 Chr.8: 42906215-42906215 on Build GRCh38
rs1015003519 Chr.8: 42906532-42906532 on Build GRCh38
rs545493717 Chr.8: 42906363-42906363 on Build GRCh38
rs968999395 Chr.8: 42906488-42906488 on Build GRCh38
rs745555738 Chr.8: 42906289-42906289 on Build GRCh38
rs369150233 Chr.8: 42906230-42906230 on Build GRCh38
rs371943831 Chr.8: 42906145-42906146 on Build GRCh38
rs527914119 Chr.8: 42906397-42906397 on Build GRCh38
rs554879439 Chr.8: 42906160-42906160 on Build GRCh38
rs757606567 Chr.8: 42906222-42906222 on Build GRCh38
rs554584504 Chr.8: 42906146-42906146 on Build GRCh38
rs1029977120 Chr.8: 42906265-42906265 on Build GRCh38
rs889083706 Chr.8: 42906357-42906357 on Build GRCh38
rs1019416192 Chr.8: 42906364-42906364 on Build GRCh38
rs758522063 Chr.8: 42906169-42906169 on Build GRCh38
rs200070946 Chr.8: 42906149-42906150 on Build GRCh38
rs749087990 Chr.8: 42906185-42906185 on Build GRCh38
rs779104416 Chr.8: 42906164-42906164 on Build GRCh38
rs757422029 Chr.8: 42906149-42906149 on Build GRCh38

View Full Product Details