Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 6519727-6520263 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ANGPT2
Entrez Gene ID
285
Gene Name
angiopoietin 2
Gene Aliases
AGPT2, ANG2
UniGene
Hs.583870
Gene Chromosome Location
Chr.8: 6499651-6563420 on Build GRCh38
Transcripts
NM_001118887.1 NM_001118888.1 NM_001147.2 XM_017013318.1 NM_001322042.1 NM_024596.4 XM_011534755.2 XM_011534756.2 XM_011534757.2 XM_011534759.2 XM_011534760.2 XM_017013829.1 XM_017013830.1 XM_017013831.1 XM_017013832.1 XM_017013833.1 XM_017013834.1

Gene Symbol
MCPH1
Entrez Gene ID
79648
Gene Name
microcephalin 1
Gene Aliases
BRIT1, MCT
UniGene
Hs.593807
Gene Chromosome Location
Chr.8: 6406592-6648505 on Build GRCh38
Transcripts
NM_001118887.1 NM_001118888.1 NM_001147.2 XM_017013318.1 NM_001322042.1 NM_024596.4 XM_011534755.2 XM_011534756.2 XM_011534757.2 XM_011534759.2 XM_011534760.2 XM_017013829.1 XM_017013830.1 XM_017013831.1 XM_017013832.1 XM_017013833.1 XM_017013834.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1100980 c.808G>A p.D270N Chr.8: 6519986-6519986 on Build GRCh38
1457779 c.813C>A p.P271P Chr.8: 6519981-6519981 on Build GRCh38
4163035 c.882G>A p.T294T Chr.8: 6519912-6519912 on Build GRCh38
6599619 c.854C>T p.A285V Chr.8: 6519940-6519940 on Build GRCh38
7111934 c.927C>G p.I309M Chr.8: 6519867-6519867 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs756054224 Chr.8: 6519832-6519832 on Build GRCh38
rs150416680 Chr.8: 6519798-6519798 on Build GRCh38
rs926744579 Chr.8: 6520142-6520142 on Build GRCh38
rs779486336 Chr.8: 6519834-6519834 on Build GRCh38
rs139074079 Chr.8: 6520088-6520088 on Build GRCh38
rs1046297922 Chr.8: 6520114-6520114 on Build GRCh38
rs760527678 Chr.8: 6519926-6519926 on Build GRCh38
rs935360315 Chr.8: 6519793-6519793 on Build GRCh38
rs899558026 Chr.8: 6520081-6520081 on Build GRCh38
rs922502949 Chr.8: 6519838-6519838 on Build GRCh38
rs752192087 Chr.8: 6519947-6519947 on Build GRCh38
rs753844136 Chr.8: 6519939-6519939 on Build GRCh38
rs567105951 Chr.8: 6520136-6520136 on Build GRCh38
rs929443659 Chr.8: 6520184-6520187 on Build GRCh38
rs1018422444 Chr.8: 6519875-6519875 on Build GRCh38
rs147113668 Chr.8: 6519986-6519986 on Build GRCh38
rs772416353 Chr.8: 6519923-6519923 on Build GRCh38
rs368790940 Chr.8: 6519852-6519852 on Build GRCh38
rs376445795 Chr.8: 6520008-6520008 on Build GRCh38
rs968048456 Chr.8: 6520130-6520130 on Build GRCh38
rs980843322 Chr.8: 6519936-6519936 on Build GRCh38
rs1055832214 Chr.8: 6520067-6520067 on Build GRCh38
rs748239199 Chr.8: 6520013-6520013 on Build GRCh38
rs1024974015 Chr.8: 6519769-6519769 on Build GRCh38
rs945918287 Chr.8: 6520031-6520031 on Build GRCh38
rs765837596 Chr.8: 6520098-6520098 on Build GRCh38
rs935996442 Chr.8: 6520059-6520059 on Build GRCh38
rs755522191 Chr.8: 6519999-6519999 on Build GRCh38
rs141170504 Chr.8: 6519974-6519974 on Build GRCh38
rs138224215 Chr.8: 6519859-6519859 on Build GRCh38
rs962302409 Chr.8: 6519791-6519791 on Build GRCh38
rs775503402 Chr.8: 6519858-6519858 on Build GRCh38
rs536740263 Chr.8: 6520072-6520072 on Build GRCh38
rs867286365 Chr.8: 6520152-6520152 on Build GRCh38
rs2442622 Chr.8: 6519809-6519809 on Build GRCh38
rs144222266 Chr.8: 6519909-6519909 on Build GRCh38
rs766344540 Chr.8: 6519893-6519893 on Build GRCh38
rs992249280 Chr.8: 6520171-6520171 on Build GRCh38
rs774737058 Chr.8: 6519870-6519870 on Build GRCh38
rs376681192 Chr.8: 6519815-6519815 on Build GRCh38
rs758025831 Chr.8: 6519903-6519903 on Build GRCh38
rs1022147371 Chr.8: 6520128-6520128 on Build GRCh38
rs919460984 Chr.8: 6520182-6520183 on Build GRCh38
rs926685890 Chr.8: 6519977-6519977 on Build GRCh38
rs942531218 Chr.8: 6519822-6519822 on Build GRCh38
rs946768350 Chr.8: 6519805-6519805 on Build GRCh38
rs532699897 Chr.8: 6520022-6520022 on Build GRCh38
rs749224375 Chr.8: 6519753-6519753 on Build GRCh38
rs1961222 Chr.8: 6519912-6519912 on Build GRCh38
rs758115390 Chr.8: 6519948-6519948 on Build GRCh38
rs1057009494 Chr.8: 6520014-6520014 on Build GRCh38
rs774544582 Chr.8: 6519922-6519922 on Build GRCh38
rs201656196 Chr.8: 6519913-6519913 on Build GRCh38
rs778546365 Chr.8: 6519843-6519843 on Build GRCh38
rs760319549 Chr.8: 6520095-6520095 on Build GRCh38
rs1005623000 Chr.8: 6520213-6520213 on Build GRCh38
rs556677528 Chr.8: 6519800-6519800 on Build GRCh38
rs1009478658 Chr.8: 6520127-6520127 on Build GRCh38
rs759437348 Chr.8: 6519940-6519940 on Build GRCh38
rs1029874891 Chr.8: 6520111-6520111 on Build GRCh38
rs984292094 Chr.8: 6519806-6519806 on Build GRCh38
rs764774322 Chr.8: 6519943-6519943 on Build GRCh38
rs751114936 Chr.8: 6519964-6519964 on Build GRCh38
rs182334057 Chr.8: 6519872-6519872 on Build GRCh38
rs1049340408 Chr.8: 6520035-6520035 on Build GRCh38
rs776386762 Chr.8: 6520039-6520039 on Build GRCh38
rs750361458 Chr.8: 6519829-6519829 on Build GRCh38
rs753656054 Chr.8: 6519897-6519897 on Build GRCh38
rs963863238 Chr.8: 6520012-6520012 on Build GRCh38
rs749767790 Chr.8: 6519918-6519918 on Build GRCh38
rs1017968843 Chr.8: 6520215-6520215 on Build GRCh38
rs983849219 Chr.8: 6519970-6519970 on Build GRCh38
rs763812248 Chr.8: 6519952-6519952 on Build GRCh38
rs757132726 Chr.8: 6519823-6519823 on Build GRCh38
rs144393044 Chr.8: 6520142-6520146 on Build GRCh38
rs111537897 Chr.8: 6519915-6519915 on Build GRCh38
rs767372535 Chr.8: 6519827-6519827 on Build GRCh38
rs373059384 Chr.8: 6520043-6520043 on Build GRCh38
rs964111015 Chr.8: 6520229-6520229 on Build GRCh38
rs993678517 Chr.8: 6520121-6520121 on Build GRCh38
rs956790865 Chr.8: 6519905-6519905 on Build GRCh38
rs773505391 Chr.8: 6520023-6520023 on Build GRCh38
rs766039126 Chr.8: 6519935-6519935 on Build GRCh38
rs751765666 Chr.8: 6519820-6519820 on Build GRCh38
rs1027865511 Chr.8: 6520148-6520148 on Build GRCh38
rs750894272 Chr.8: 6520110-6520110 on Build GRCh38
rs952443080 Chr.8: 6519804-6519804 on Build GRCh38
rs561085589 Chr.8: 6519911-6519911 on Build GRCh38
rs373427487 Chr.8: 6519853-6519853 on Build GRCh38
rs945968971 Chr.8: 6519845-6519845 on Build GRCh38
rs61733318 Chr.8: 6519898-6519898 on Build GRCh38
rs767462360 Chr.8: 6519882-6519882 on Build GRCh38
rs2922898 Chr.8: 6520058-6520058 on Build GRCh38
rs562204725 Chr.8: 6519930-6519930 on Build GRCh38
rs771313941 Chr.8: 6519847-6519847 on Build GRCh38
rs760595816 Chr.8: 6519887-6519887 on Build GRCh38
rs2922899 Chr.8: 6520037-6520037 on Build GRCh38
rs964220157 Chr.8: 6519774-6519774 on Build GRCh38
rs577701612 Chr.8: 6520028-6520028 on Build GRCh38
rs145451529 Chr.8: 6519920-6519920 on Build GRCh38
rs753383272 Chr.8: 6519836-6519836 on Build GRCh38
rs961042581 Chr.8: 6520157-6520157 on Build GRCh38
rs770576887 Chr.8: 6519766-6519766 on Build GRCh38
rs898013435 Chr.8: 6520118-6520118 on Build GRCh38
rs750217936 Chr.8: 6519885-6519885 on Build GRCh38
rs747746759 Chr.8: 6519846-6519846 on Build GRCh38
rs939495421 Chr.8: 6519816-6519816 on Build GRCh38
rs149555402 Chr.8: 6519867-6519867 on Build GRCh38
rs976720019 Chr.8: 6519777-6519777 on Build GRCh38

View Full Product Details