Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 25886677-25887185 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EBF2
Entrez Gene ID
64641
Gene Name
early B-cell factor 2
Gene Aliases
COE2, EBF-2, O/E-3, OE-3
UniGene
Hs.491292
Gene Chromosome Location
Chr.8: 25841730-26045124 on Build GRCh38
Transcripts
NM_022659.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5544881 c.932C>A p.P311Q Chr.8: 25886832-25886832 on Build GRCh38
1456283 c.893C>A p.P298H Chr.8: 25886871-25886871 on Build GRCh38
5874443 c.982G>A p.G328R Chr.8: 25886782-25886782 on Build GRCh38
486337 c.970C>G p.Q324E Chr.8: 25886794-25886794 on Build GRCh38
749989 c.999C>T p.F333F Chr.8: 25886765-25886765 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs59529004 Chr.8: 25887136-25887137 on Build GRCh38
rs752076528 Chr.8: 25886819-25886819 on Build GRCh38
rs796069070 Chr.8: 25887136-25887137 on Build GRCh38
rs765464688 Chr.8: 25886769-25886769 on Build GRCh38
rs904264581 Chr.8: 25887141-25887141 on Build GRCh38
rs202043511 Chr.8: 25886828-25886828 on Build GRCh38
rs908515182 Chr.8: 25887051-25887051 on Build GRCh38
rs74875427 Chr.8: 25887094-25887094 on Build GRCh38
rs369023999 Chr.8: 25887113-25887113 on Build GRCh38
rs375086316 Chr.8: 25886778-25886778 on Build GRCh38
rs771408774 Chr.8: 25887098-25887098 on Build GRCh38
rs760033612 Chr.8: 25886898-25886898 on Build GRCh38
rs958180206 Chr.8: 25887016-25887016 on Build GRCh38
rs759001146 Chr.8: 25886734-25886734 on Build GRCh38
rs745510571 Chr.8: 25886872-25886872 on Build GRCh38
rs11135900 Chr.8: 25886939-25886939 on Build GRCh38
rs937716365 Chr.8: 25887148-25887148 on Build GRCh38
rs376101300 Chr.8: 25886859-25886859 on Build GRCh38
rs372836065 Chr.8: 25886704-25886704 on Build GRCh38
rs67623458 Chr.8: 25887118-25887119 on Build GRCh38
rs758458636 Chr.8: 25886787-25886787 on Build GRCh38
rs975493299 Chr.8: 25886995-25886995 on Build GRCh38
rs542514301 Chr.8: 25886964-25886964 on Build GRCh38
rs77426684 Chr.8: 25887100-25887100 on Build GRCh38
rs899800777 Chr.8: 25887106-25887109 on Build GRCh38
rs746233424 Chr.8: 25887101-25887101 on Build GRCh38
rs186402048 Chr.8: 25886970-25886970 on Build GRCh38
rs770721077 Chr.8: 25886723-25886723 on Build GRCh38
rs368733185 Chr.8: 25887062-25887062 on Build GRCh38
rs376263965 Chr.8: 25887135-25887136 on Build GRCh38
rs954629037 Chr.8: 25886944-25886944 on Build GRCh38
rs750138922 Chr.8: 25886887-25886887 on Build GRCh38
rs372685484 Chr.8: 25887117-25887117 on Build GRCh38
rs748547353 Chr.8: 25886831-25886831 on Build GRCh38
rs1046042291 Chr.8: 25886821-25886821 on Build GRCh38
rs776501042 Chr.8: 25886914-25886914 on Build GRCh38
rs770102146 Chr.8: 25887102-25887102 on Build GRCh38
rs1032462575 Chr.8: 25887011-25887011 on Build GRCh38
rs75327259 Chr.8: 25887118-25887118 on Build GRCh38
rs1047485984 Chr.8: 25887044-25887044 on Build GRCh38
rs988350589 Chr.8: 25887086-25887086 on Build GRCh38
rs770244798 Chr.8: 25886842-25886842 on Build GRCh38
rs529186189 Chr.8: 25886940-25886940 on Build GRCh38
rs760647893 Chr.8: 25886742-25886742 on Build GRCh38
rs1031854153 Chr.8: 25887145-25887145 on Build GRCh38
rs753630189 Chr.8: 25886752-25886752 on Build GRCh38
rs775798896 Chr.8: 25887103-25887103 on Build GRCh38
rs1037403610 Chr.8: 25887149-25887149 on Build GRCh38
rs919999263 Chr.8: 25886980-25886980 on Build GRCh38
rs570396891 Chr.8: 25887119-25887120 on Build GRCh38
rs914074208 Chr.8: 25887095-25887095 on Build GRCh38
rs192519568 Chr.8: 25887137-25887137 on Build GRCh38
rs757905310 Chr.8: 25887154-25887154 on Build GRCh38
rs926250261 Chr.8: 25887123-25887123 on Build GRCh38
rs761631326 Chr.8: 25887101-25887102 on Build GRCh38
rs944049715 Chr.8: 25887091-25887091 on Build GRCh38
rs774098760 Chr.8: 25886728-25886728 on Build GRCh38
rs543590179 Chr.8: 25886902-25886902 on Build GRCh38
rs182170287 Chr.8: 25886705-25886705 on Build GRCh38
rs1021327200 Chr.8: 25886706-25886706 on Build GRCh38
rs1042901172 Chr.8: 25887114-25887137 on Build GRCh38
rs571691529 Chr.8: 25887099-25887099 on Build GRCh38
rs898999838 Chr.8: 25886893-25886893 on Build GRCh38
rs1037413614 Chr.8: 25886844-25886844 on Build GRCh38
rs6557865 Chr.8: 25886741-25886741 on Build GRCh38
rs768304832 Chr.8: 25886967-25886967 on Build GRCh38
rs557856249 Chr.8: 25886768-25886768 on Build GRCh38
rs201815898 Chr.8: 25887135-25887135 on Build GRCh38
rs767021050 Chr.8: 25886735-25886735 on Build GRCh38
rs774150131 Chr.8: 25886775-25886777 on Build GRCh38
rs763942684 Chr.8: 25886743-25886743 on Build GRCh38
rs528480379 Chr.8: 25886971-25886971 on Build GRCh38
rs771539401 Chr.8: 25886884-25886884 on Build GRCh38
rs369397370 Chr.8: 25886897-25886897 on Build GRCh38
rs12675104 Chr.8: 25887109-25887109 on Build GRCh38
rs200807889 Chr.8: 25886777-25886777 on Build GRCh38
rs368351856 Chr.8: 25886827-25886827 on Build GRCh38
rs780171954 Chr.8: 25886801-25886801 on Build GRCh38
rs138654893 Chr.8: 25886996-25886996 on Build GRCh38

View Full Product Details