Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 1056414-1056923 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DMRT2
Entrez Gene ID
10655
Gene Name
doublesex and mab-3 related transcription factor 2
Gene Aliases
-
UniGene
Hs.59506
Gene Chromosome Location
Chr.9: 1003237-1057554 on Build GRCh38
Transcripts
NM_001130865.2 NM_006557.6 NM_181872.4 XM_011517687.1 XM_011517690.2 XM_011517694.2 XM_017014213.1 XM_017014214.1 XM_017014215.1 XM_017014216.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5606477 c.1308C>T p.F436F Chr.9: 1056895-1056895 on Build GRCh38
6645676 c.922G>T p.D308Y Chr.9: 1056509-1056509 on Build GRCh38
4678139 c.1043G>A p.G348D Chr.9: 1056630-1056630 on Build GRCh38
5559334 c.1009T>G p.L337V Chr.9: 1056596-1056596 on Build GRCh38
4600730 c.1099G>T p.V367F Chr.9: 1056686-1056686 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs754599241 Chr.9: 1056888-1056888 on Build GRCh38
rs527258854 Chr.9: 1056547-1056547 on Build GRCh38
rs16928356 Chr.9: 1056517-1056517 on Build GRCh38
rs762838054 Chr.9: 1056653-1056653 on Build GRCh38
rs140337454 Chr.9: 1056868-1056868 on Build GRCh38
rs746515910 Chr.9: 1056704-1056704 on Build GRCh38
rs144648877 Chr.9: 1056471-1056471 on Build GRCh38
rs772030456 Chr.9: 1056869-1056869 on Build GRCh38
rs770167252 Chr.9: 1056519-1056519 on Build GRCh38
rs762928577 Chr.9: 1056726-1056726 on Build GRCh38
rs779154249 Chr.9: 1056777-1056778 on Build GRCh38
rs922923589 Chr.9: 1056809-1056809 on Build GRCh38
rs748889790 Chr.9: 1056858-1056858 on Build GRCh38
rs371929280 Chr.9: 1056700-1056700 on Build GRCh38
rs780616964 Chr.9: 1056851-1056851 on Build GRCh38
rs1006425526 Chr.9: 1056601-1056601 on Build GRCh38
rs564464685 Chr.9: 1056646-1056646 on Build GRCh38
rs752041820 Chr.9: 1056826-1056826 on Build GRCh38
rs753096035 Chr.9: 1056749-1056749 on Build GRCh38
rs780242958 Chr.9: 1056524-1056524 on Build GRCh38
rs41311430 Chr.9: 1056870-1056870 on Build GRCh38
rs777960815 Chr.9: 1056504-1056504 on Build GRCh38
rs202027041 Chr.9: 1056724-1056724 on Build GRCh38
rs770480487 Chr.9: 1056641-1056641 on Build GRCh38
rs767625504 Chr.9: 1056663-1056663 on Build GRCh38
rs370513290 Chr.9: 1056884-1056884 on Build GRCh38
rs760360224 Chr.9: 1056567-1056567 on Build GRCh38
rs746875160 Chr.9: 1056902-1056902 on Build GRCh38
rs759258940 Chr.9: 1056644-1056644 on Build GRCh38
rs777782068 Chr.9: 1056900-1056900 on Build GRCh38
rs770081495 Chr.9: 1056466-1056466 on Build GRCh38
rs762285885 Chr.9: 1056882-1056882 on Build GRCh38
rs140701396 Chr.9: 1056516-1056516 on Build GRCh38
rs758059465 Chr.9: 1056897-1056897 on Build GRCh38
rs944994027 Chr.9: 1056654-1056654 on Build GRCh38
rs150989542 Chr.9: 1056775-1056775 on Build GRCh38
rs763320879 Chr.9: 1056470-1056470 on Build GRCh38
rs142906421 Chr.9: 1056572-1056572 on Build GRCh38
rs143092642 Chr.9: 1056852-1056852 on Build GRCh38
rs372603448 Chr.9: 1056810-1056810 on Build GRCh38
rs563777846 Chr.9: 1056484-1056484 on Build GRCh38
rs775881772 Chr.9: 1056874-1056874 on Build GRCh38
rs140004479 Chr.9: 1056569-1056569 on Build GRCh38
rs779299775 Chr.9: 1056625-1056625 on Build GRCh38
rs780144126 Chr.9: 1056442-1056442 on Build GRCh38
rs749085023 Chr.9: 1056782-1056782 on Build GRCh38
rs139912017 Chr.9: 1056675-1056675 on Build GRCh38
rs779378470 Chr.9: 1056899-1056899 on Build GRCh38
rs755320432 Chr.9: 1056690-1056690 on Build GRCh38
rs779292473 Chr.9: 1056699-1056699 on Build GRCh38
rs141302557 Chr.9: 1056548-1056548 on Build GRCh38
rs773977303 Chr.9: 1056555-1056555 on Build GRCh38
rs761770101 Chr.9: 1056798-1056798 on Build GRCh38
rs754836991 Chr.9: 1056500-1056500 on Build GRCh38
rs575076415 Chr.9: 1056779-1056779 on Build GRCh38
rs775428927 Chr.9: 1056647-1056647 on Build GRCh38
rs138207989 Chr.9: 1056871-1056871 on Build GRCh38
rs752860436 Chr.9: 1056680-1056680 on Build GRCh38
rs147461872 Chr.9: 1056887-1056887 on Build GRCh38
rs779693419 Chr.9: 1056856-1056856 on Build GRCh38
rs764593509 Chr.9: 1056879-1056879 on Build GRCh38
rs914360965 Chr.9: 1056468-1056468 on Build GRCh38
rs772814583 Chr.9: 1056636-1056636 on Build GRCh38
rs369891962 Chr.9: 1056707-1056707 on Build GRCh38
rs148245031 Chr.9: 1056864-1056864 on Build GRCh38
rs750029316 Chr.9: 1056481-1056481 on Build GRCh38
rs773704993 Chr.9: 1056474-1056474 on Build GRCh38
rs1038715336 Chr.9: 1056487-1056487 on Build GRCh38
rs895237252 Chr.9: 1056709-1056709 on Build GRCh38
rs778628435 Chr.9: 1056895-1056895 on Build GRCh38
rs1046103823 Chr.9: 1056678-1056678 on Build GRCh38
rs765548746 Chr.9: 1056806-1056806 on Build GRCh38
rs879139735 Chr.9: 1056876-1056876 on Build GRCh38
rs759044350 Chr.9: 1056755-1056755 on Build GRCh38
rs771122278 Chr.9: 1056453-1056453 on Build GRCh38
rs140844621 Chr.9: 1056803-1056803 on Build GRCh38
rs199502026 Chr.9: 1056582-1056582 on Build GRCh38
rs151086071 Chr.9: 1056590-1056590 on Build GRCh38
rs368560020 Chr.9: 1056450-1056450 on Build GRCh38
rs377182125 Chr.9: 1056863-1056863 on Build GRCh38
rs147471809 Chr.9: 1056532-1056532 on Build GRCh38
rs757751363 Chr.9: 1056830-1056830 on Build GRCh38
rs756393184 Chr.9: 1056676-1056676 on Build GRCh38
rs749610895 Chr.9: 1056624-1056624 on Build GRCh38
rs766223609 Chr.9: 1056486-1056486 on Build GRCh38
rs521525 Chr.9: 1056787-1056787 on Build GRCh38
rs753842208 Chr.9: 1056499-1056499 on Build GRCh38
rs746033739 Chr.9: 1056518-1056518 on Build GRCh38
rs72703237 Chr.9: 1056732-1056732 on Build GRCh38
rs113111105 Chr.9: 1056877-1056877 on Build GRCh38
rs761412181 Chr.9: 1056561-1056561 on Build GRCh38
rs536600958 Chr.9: 1056799-1056799 on Build GRCh38
rs755790680 Chr.9: 1056853-1056853 on Build GRCh38
rs182646996 Chr.9: 1056562-1056562 on Build GRCh38
rs750675828 Chr.9: 1056588-1056588 on Build GRCh38
rs747967718 Chr.9: 1056896-1056896 on Build GRCh38
rs769466615 Chr.9: 1056777-1056777 on Build GRCh38
rs372851397 Chr.9: 1056458-1056458 on Build GRCh38
rs757702014 Chr.9: 1056508-1056508 on Build GRCh38
rs375812912 Chr.9: 1056763-1056763 on Build GRCh38
rs747291755 Chr.9: 1056449-1056449 on Build GRCh38
rs113748895 Chr.9: 1056640-1056640 on Build GRCh38
rs543802092 Chr.9: 1056793-1056793 on Build GRCh38
rs749118381 Chr.9: 1056443-1056443 on Build GRCh38
rs756745369 Chr.9: 1056769-1056769 on Build GRCh38
rs371164792 Chr.9: 1056477-1056477 on Build GRCh38
rs1031631742 Chr.9: 1056761-1056761 on Build GRCh38
rs777481724 Chr.9: 1056703-1056703 on Build GRCh38
rs142308383 Chr.9: 1056774-1056774 on Build GRCh38
rs3824419 Chr.9: 1056728-1056728 on Build GRCh38
rs748613087 Chr.9: 1056626-1056626 on Build GRCh38
rs200683258 Chr.9: 1056635-1056635 on Build GRCh38
rs778431616 Chr.9: 1056759-1056759 on Build GRCh38
rs377059611 Chr.9: 1056469-1056469 on Build GRCh38
rs777041286 Chr.9: 1056455-1056455 on Build GRCh38
rs749842545 Chr.9: 1056530-1056530 on Build GRCh38
rs776165447 Chr.9: 1056711-1056711 on Build GRCh38
rs773017461 Chr.9: 1056565-1056565 on Build GRCh38
rs774294154 Chr.9: 1056796-1056796 on Build GRCh38
rs745479643 Chr.9: 1056717-1056717 on Build GRCh38
rs750836199 Chr.9: 1056885-1056885 on Build GRCh38
rs375071878 Chr.9: 1056701-1056701 on Build GRCh38
rs781201005 Chr.9: 1056583-1056583 on Build GRCh38
rs376262081 Chr.9: 1056579-1056579 on Build GRCh38
rs777672832 Chr.9: 1056444-1056444 on Build GRCh38
rs758428225 Chr.9: 1056534-1056534 on Build GRCh38
rs763426492 Chr.9: 1056820-1056820 on Build GRCh38
rs748491009 Chr.9: 1056878-1056879 on Build GRCh38
rs754033173 Chr.9: 1056684-1056684 on Build GRCh38
rs564642993 Chr.9: 1056546-1056546 on Build GRCh38
rs776746176 Chr.9: 1056800-1056800 on Build GRCh38
rs750482012 Chr.9: 1056664-1056664 on Build GRCh38
rs371344054 Chr.9: 1056568-1056568 on Build GRCh38
rs191456726 Chr.9: 1056751-1056751 on Build GRCh38
rs765743642 Chr.9: 1056743-1056743 on Build GRCh38
rs780517526 Chr.9: 1056605-1056605 on Build GRCh38
rs760681935 Chr.9: 1056731-1056731 on Build GRCh38
rs556954799 Chr.9: 1056821-1056821 on Build GRCh38
rs112701754 Chr.9: 1056559-1056559 on Build GRCh38
rs769507886 Chr.9: 1056719-1056719 on Build GRCh38
rs149054600 Chr.9: 1056825-1056825 on Build GRCh38
rs765028262 Chr.9: 1056645-1056645 on Build GRCh38
rs1026674641 Chr.9: 1056451-1056451 on Build GRCh38

View Full Product Details